Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118544366-118544908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT46
Entrez Gene ID
56912
Gene Name
intraflagellar transport 46
Gene Aliases
C11orf2, C11orf60, CFAP32
UniGene
Hs.533738
Gene Chromosome Location
Chr.11: 118544528-118575624 on Build GRCh38
Transcripts
NM_001168618.1 NM_020153.3 XM_011542905.2 XM_011542906.2 XM_017018017.1 XM_017018018.1 NM_001144037.1 NM_001144038.1 XM_017018431.1

Gene Symbol
TMEM25
Entrez Gene ID
84866
Gene Name
transmembrane protein 25
Gene Aliases
-
UniGene
Hs.564188
Gene Chromosome Location
Chr.11: 118531088-118546670 on Build GRCh38
Transcripts
NM_001168618.1 NM_020153.3 XM_011542905.2 XM_011542906.2 XM_017018017.1 XM_017018018.1 NM_001144037.1 NM_001144038.1 XM_017018431.1

SNP Detail

SNP ID Chromosome Location
rs188920459 Chr.11: 118544533-118544533 on Build GRCh38
rs560192814 Chr.11: 118544548-118544548 on Build GRCh38
rs141881724 Chr.11: 118544529-118544529 on Build GRCh38
rs76143861 Chr.11: 118544714-118544714 on Build GRCh38
rs1030468213 Chr.11: 118544723-118544723 on Build GRCh38
rs763367139 Chr.11: 118544684-118544684 on Build GRCh38
rs958309478 Chr.11: 118544792-118544792 on Build GRCh38
rs953682010 Chr.11: 118544475-118544475 on Build GRCh38
rs752001311 Chr.11: 118544716-118544716 on Build GRCh38
rs997677934 Chr.11: 118544455-118544455 on Build GRCh38
rs562277331 Chr.11: 118544476-118544476 on Build GRCh38
rs782208580 Chr.11: 118544699-118544699 on Build GRCh38
rs75320610 Chr.11: 118544391-118544391 on Build GRCh38
rs949115116 Chr.11: 118544546-118544546 on Build GRCh38
rs897905537 Chr.11: 118544446-118544446 on Build GRCh38
rs887746739 Chr.11: 118544398-118544398 on Build GRCh38
rs1020615853 Chr.11: 118544807-118544807 on Build GRCh38
rs549745827 Chr.11: 118544675-118544675 on Build GRCh38
rs116622289 Chr.11: 118544655-118544655 on Build GRCh38
rs80130455 Chr.11: 118544431-118544431 on Build GRCh38
rs782055525 Chr.11: 118544872-118544872 on Build GRCh38
rs375236450 Chr.11: 118544530-118544530 on Build GRCh38
rs112051935 Chr.11: 118544645-118544645 on Build GRCh38
rs937956179 Chr.11: 118544636-118544636 on Build GRCh38
rs781792454 Chr.11: 118544466-118544466 on Build GRCh38
rs782740216 Chr.11: 118544692-118544692 on Build GRCh38
rs1032022943 Chr.11: 118544490-118544490 on Build GRCh38
rs782765387 Chr.11: 118544414-118544414 on Build GRCh38
rs781866641 Chr.11: 118544881-118544881 on Build GRCh38
rs557806358 Chr.11: 118544859-118544859 on Build GRCh38
rs79627484 Chr.11: 118544768-118544768 on Build GRCh38
rs889258303 Chr.11: 118544748-118544748 on Build GRCh38
rs527340652 Chr.11: 118544577-118544577 on Build GRCh38
rs782782431 Chr.11: 118544880-118544880 on Build GRCh38
rs546689601 Chr.11: 118544666-118544666 on Build GRCh38
rs782488831 Chr.11: 118544884-118544884 on Build GRCh38
rs1048765946 Chr.11: 118544644-118544644 on Build GRCh38
rs913689905 Chr.11: 118544536-118544536 on Build GRCh38
rs987846410 Chr.11: 118544799-118544799 on Build GRCh38
rs782798375 Chr.11: 118544521-118544521 on Build GRCh38
rs782193181 Chr.11: 118544809-118544809 on Build GRCh38
rs937804156 Chr.11: 118544874-118544874 on Build GRCh38
rs116534318 Chr.11: 118544779-118544779 on Build GRCh38
rs781944908 Chr.11: 118544871-118544871 on Build GRCh38
rs782513108 Chr.11: 118544585-118544585 on Build GRCh38
rs181628467 Chr.11: 118544868-118544868 on Build GRCh38
rs535408373 Chr.11: 118544742-118544742 on Build GRCh38
rs981966829 Chr.11: 118544576-118544576 on Build GRCh38
rs139719866 Chr.11: 118544850-118544850 on Build GRCh38

View Full Product Details