Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101396216-101396760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL1
Entrez Gene ID
400
Gene Name
ADP ribosylation factor like GTPase 1
Gene Aliases
ARFL1
UniGene
Hs.372616
Gene Chromosome Location
Chr.12: 101393120-101407820 on Build GRCh38
Transcripts
NM_001177.5 NM_001301068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604776 c.451C>T p.R151* Chr.12: 101396463-101396463 on Build GRCh38
2063365 c.452G>A p.R151Q Chr.12: 101396462-101396462 on Build GRCh38
430163 c.508A>G p.M170V Chr.12: 101396406-101396406 on Build GRCh38
2063364 c.492C>T p.T164T Chr.12: 101396422-101396422 on Build GRCh38
127077 c.337-1G>C p.? Chr.12: 101396578-101396578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773359188 Chr.12: 101396609-101396609 on Build GRCh38
rs770515193 Chr.12: 101396561-101396561 on Build GRCh38
rs931359980 Chr.12: 101396620-101396620 on Build GRCh38
rs767710122 Chr.12: 101396692-101396692 on Build GRCh38
rs528202246 Chr.12: 101396627-101396627 on Build GRCh38
rs768976320 Chr.12: 101396340-101396340 on Build GRCh38
rs965204214 Chr.12: 101396548-101396548 on Build GRCh38
rs144056647 Chr.12: 101396362-101396362 on Build GRCh38
rs759390187 Chr.12: 101396314-101396314 on Build GRCh38
rs780485078 Chr.12: 101396303-101396303 on Build GRCh38
rs757378518 Chr.12: 101396320-101396320 on Build GRCh38
rs955965029 Chr.12: 101396575-101396575 on Build GRCh38
rs1021684257 Chr.12: 101396437-101396437 on Build GRCh38
rs758738882 Chr.12: 101396334-101396334 on Build GRCh38
rs551626425 Chr.12: 101396333-101396333 on Build GRCh38
rs944131091 Chr.12: 101396252-101396252 on Build GRCh38
rs370117976 Chr.12: 101396262-101396262 on Build GRCh38
rs766628640 Chr.12: 101396478-101396478 on Build GRCh38
rs78306115 Chr.12: 101396675-101396678 on Build GRCh38
rs775194143 Chr.12: 101396593-101396593 on Build GRCh38
rs1039693507 Chr.12: 101396279-101396279 on Build GRCh38
rs764742890 Chr.12: 101396385-101396385 on Build GRCh38
rs1019490890 Chr.12: 101396532-101396532 on Build GRCh38
rs375864214 Chr.12: 101396422-101396422 on Build GRCh38
rs73378795 Chr.12: 101396341-101396341 on Build GRCh38
rs199895049 Chr.12: 101396497-101396497 on Build GRCh38
rs759889739 Chr.12: 101396294-101396294 on Build GRCh38
rs777077559 Chr.12: 101396388-101396388 on Build GRCh38
rs940069119 Chr.12: 101396715-101396715 on Build GRCh38
rs745606476 Chr.12: 101396567-101396567 on Build GRCh38
rs900804598 Chr.12: 101396697-101396697 on Build GRCh38
rs777262033 Chr.12: 101396526-101396526 on Build GRCh38
rs748806030 Chr.12: 101396557-101396557 on Build GRCh38
rs940312563 Chr.12: 101396661-101396661 on Build GRCh38
rs760732358 Chr.12: 101396384-101396384 on Build GRCh38
rs767497521 Chr.12: 101396318-101396318 on Build GRCh38
rs573998462 Chr.12: 101396363-101396363 on Build GRCh38
rs199983546 Chr.12: 101396367-101396367 on Build GRCh38
rs755206038 Chr.12: 101396626-101396626 on Build GRCh38
rs200358299 Chr.12: 101396583-101396583 on Build GRCh38
rs769263709 Chr.12: 101396345-101396345 on Build GRCh38
rs552404672 Chr.12: 101396353-101396353 on Build GRCh38
rs536959046 Chr.12: 101396346-101396346 on Build GRCh38
rs751776862 Chr.12: 101396617-101396617 on Build GRCh38
rs977321395 Chr.12: 101396505-101396505 on Build GRCh38
rs756672768 Chr.12: 101396519-101396519 on Build GRCh38
rs544756180 Chr.12: 101396663-101396663 on Build GRCh38
rs373870337 Chr.12: 101396515-101396515 on Build GRCh38
rs763465995 Chr.12: 101396595-101396598 on Build GRCh38
rs11110778 Chr.12: 101396319-101396319 on Build GRCh38
rs747472901 Chr.12: 101396335-101396335 on Build GRCh38
rs752189207 Chr.12: 101396285-101396287 on Build GRCh38
rs11110777 Chr.12: 101396246-101396246 on Build GRCh38
rs748926794 Chr.12: 101396360-101396360 on Build GRCh38
rs751433188 Chr.12: 101396311-101396311 on Build GRCh38
rs898758644 Chr.12: 101396372-101396372 on Build GRCh38
rs202231699 Chr.12: 101396521-101396521 on Build GRCh38
rs144514450 Chr.12: 101396671-101396674 on Build GRCh38
rs369693988 Chr.12: 101396612-101396612 on Build GRCh38
rs73159756 Chr.12: 101396595-101396595 on Build GRCh38
rs906757817 Chr.12: 101396687-101396687 on Build GRCh38
rs763526739 Chr.12: 101396371-101396371 on Build GRCh38
rs750585373 Chr.12: 101396409-101396409 on Build GRCh38
rs766462105 Chr.12: 101396615-101396615 on Build GRCh38
rs981097493 Chr.12: 101396433-101396433 on Build GRCh38
rs750622693 Chr.12: 101396700-101396700 on Build GRCh38
rs747572309 Chr.12: 101396344-101396344 on Build GRCh38
rs760673081 Chr.12: 101396605-101396605 on Build GRCh38
rs116437821 Chr.12: 101396375-101396375 on Build GRCh38
rs867593405 Chr.12: 101396496-101396496 on Build GRCh38
rs781451800 Chr.12: 101396503-101396503 on Build GRCh38
rs753829623 Chr.12: 101396596-101396600 on Build GRCh38
rs751810522 Chr.12: 101396493-101396493 on Build GRCh38
rs775525503 Chr.12: 101396380-101396380 on Build GRCh38
rs921366042 Chr.12: 101396600-101396600 on Build GRCh38
rs774078652 Chr.12: 101396566-101396566 on Build GRCh38
rs1055674671 Chr.12: 101396711-101396711 on Build GRCh38
rs766142106 Chr.12: 101396306-101396306 on Build GRCh38
rs372718102 Chr.12: 101396407-101396407 on Build GRCh38

View Full Product Details