Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167974839-167975283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF6
Entrez Gene ID
55827
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326
UniGene
Hs.435741
Gene Chromosome Location
Chr.1: 167936247-168075843 on Build GRCh38
Transcripts
NM_001017977.2 NM_001198956.1 NM_001198957.1 NM_018442.3 XM_005245331.4 XM_005245332.4 XM_005245333.4 XM_017001779.1 XM_017001780.1 XM_017001781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6059204 c.330G>C p.Q110H Chr.1: 167974907-167974907 on Build GRCh38
3477856 c.292T>G p.F98V Chr.1: 167974869-167974869 on Build GRCh38
366981 c.268C>T p.R90C Chr.1: 167974845-167974845 on Build GRCh38
4024840 c.293T>C p.F98S Chr.1: 167974870-167974870 on Build GRCh38
7257234 c.408A>G p.Q136Q Chr.1: 167974985-167974985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776707096 Chr.1: 167974945-167974945 on Build GRCh38
rs372615689 Chr.1: 167975197-167975197 on Build GRCh38
rs780456850 Chr.1: 167974932-167974932 on Build GRCh38
rs374946661 Chr.1: 167975027-167975027 on Build GRCh38
rs767000590 Chr.1: 167974887-167974887 on Build GRCh38
rs759166225 Chr.1: 167974960-167974960 on Build GRCh38
rs950620761 Chr.1: 167975106-167975106 on Build GRCh38
rs752775171 Chr.1: 167974977-167974978 on Build GRCh38
rs1010724393 Chr.1: 167975238-167975238 on Build GRCh38
rs947746363 Chr.1: 167975068-167975068 on Build GRCh38
rs765811124 Chr.1: 167974990-167974990 on Build GRCh38
rs561251027 Chr.1: 167975151-167975151 on Build GRCh38
rs369044688 Chr.1: 167975043-167975043 on Build GRCh38
rs1008964119 Chr.1: 167975018-167975018 on Build GRCh38
rs764803271 Chr.1: 167975048-167975048 on Build GRCh38
rs773037457 Chr.1: 167974950-167974950 on Build GRCh38
rs758857581 Chr.1: 167975019-167975019 on Build GRCh38
rs746215430 Chr.1: 167974913-167974913 on Build GRCh38
rs1004945000 Chr.1: 167975132-167975132 on Build GRCh38
rs777715641 Chr.1: 167975056-167975056 on Build GRCh38
rs764736643 Chr.1: 167975067-167975068 on Build GRCh38
rs562768510 Chr.1: 167974934-167974934 on Build GRCh38
rs1038748710 Chr.1: 167975044-167975044 on Build GRCh38
rs757838275 Chr.1: 167974900-167974900 on Build GRCh38
rs762462208 Chr.1: 167974982-167974982 on Build GRCh38
rs779660437 Chr.1: 167974979-167974979 on Build GRCh38
rs1044702685 Chr.1: 167975082-167975082 on Build GRCh38
rs762703198 Chr.1: 167974951-167974951 on Build GRCh38
rs757079968 Chr.1: 167975053-167975053 on Build GRCh38
rs934394930 Chr.1: 167974974-167974974 on Build GRCh38
rs371881262 Chr.1: 167974877-167974877 on Build GRCh38
rs200954948 Chr.1: 167974949-167974949 on Build GRCh38
rs148842529 Chr.1: 167975058-167975058 on Build GRCh38
rs549425657 Chr.1: 167975148-167975148 on Build GRCh38
rs375511404 Chr.1: 167974898-167974898 on Build GRCh38
rs541594026 Chr.1: 167975100-167975100 on Build GRCh38
rs750922643 Chr.1: 167974991-167974991 on Build GRCh38
rs1022258409 Chr.1: 167975247-167975247 on Build GRCh38
rs906278977 Chr.1: 167975120-167975120 on Build GRCh38
rs114094645 Chr.1: 167975237-167975237 on Build GRCh38
rs573827582 Chr.1: 167974969-167974969 on Build GRCh38
rs564781398 Chr.1: 167975202-167975202 on Build GRCh38
rs760814000 Chr.1: 167975022-167975024 on Build GRCh38
rs748099536 Chr.1: 167974948-167974948 on Build GRCh38
rs779481095 Chr.1: 167974911-167974911 on Build GRCh38
rs767007303 Chr.1: 167974977-167974977 on Build GRCh38
rs190047146 Chr.1: 167975159-167975159 on Build GRCh38
rs758736718 Chr.1: 167974919-167974919 on Build GRCh38
rs1003272989 Chr.1: 167975141-167975141 on Build GRCh38
rs200026976 Chr.1: 167975051-167975051 on Build GRCh38
rs773893367 Chr.1: 167974959-167974959 on Build GRCh38
rs527518461 Chr.1: 167975131-167975131 on Build GRCh38
rs768632730 Chr.1: 167974941-167974941 on Build GRCh38
rs770670114 Chr.1: 167974955-167974955 on Build GRCh38
rs916618754 Chr.1: 167975154-167975154 on Build GRCh38
rs751734893 Chr.1: 167975037-167975037 on Build GRCh38
rs549800934 Chr.1: 167975232-167975232 on Build GRCh38
rs866418838 Chr.1: 167975036-167975036 on Build GRCh38
rs776704522 Chr.1: 167975239-167975239 on Build GRCh38
rs1035497105 Chr.1: 167975134-167975134 on Build GRCh38

View Full Product Details