Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86604637-86605182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374845
Entrez Gene ID
105374845
Gene Name
uncharacterized LOC105374845
Gene Aliases
-
UniGene
Hs.741633
Gene Chromosome Location
Chr.2: 86562345-86618766 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103
Entrez Gene ID
7844
Gene Name
ring finger protein 103
Gene Aliases
HKF-1, KF-1, KF1, ZFP-103, ZFP103
UniGene
Hs.469199
Gene Chromosome Location
Chr.2: 86603393-86623877 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Gene Symbol
RNF103-CHMP3
Entrez Gene ID
100526767
Gene Name
RNF103-CHMP3 readthrough
Gene Aliases
RNF103-VPS24
UniGene
Hs.591582
Gene Chromosome Location
Chr.2: 86503430-86721122 on Build GRCh38
Transcripts
NM_001198951.1 NM_005667.3 NM_001198954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380284 c.1216G>A p.V406I Chr.2: 86604685-86604685 on Build GRCh38
5770905 c.1096C>T p.L366L Chr.2: 86604805-86604805 on Build GRCh38
1690764 c.1109C>T p.S370F Chr.2: 86604792-86604792 on Build GRCh38
3426743 c.1101A>C p.L367F Chr.2: 86604800-86604800 on Build GRCh38
4583684 c.1084A>G p.T362A Chr.2: 86604817-86604817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756884860 Chr.2: 86605115-86605115 on Build GRCh38
rs771790880 Chr.2: 86604764-86604764 on Build GRCh38
rs755509760 Chr.2: 86604995-86604995 on Build GRCh38
rs936164975 Chr.2: 86604773-86604773 on Build GRCh38
rs781026511 Chr.2: 86605120-86605120 on Build GRCh38
rs17857046 Chr.2: 86605150-86605150 on Build GRCh38
rs771983283 Chr.2: 86605061-86605061 on Build GRCh38
rs369237335 Chr.2: 86604796-86604796 on Build GRCh38
rs766889810 Chr.2: 86605078-86605078 on Build GRCh38
rs774155968 Chr.2: 86604726-86604726 on Build GRCh38
rs192990433 Chr.2: 86605091-86605091 on Build GRCh38
rs370678832 Chr.2: 86605014-86605014 on Build GRCh38
rs371225381 Chr.2: 86605082-86605082 on Build GRCh38
rs759429482 Chr.2: 86604950-86604950 on Build GRCh38
rs757585213 Chr.2: 86604660-86604660 on Build GRCh38
rs774394413 Chr.2: 86604961-86604961 on Build GRCh38
rs762137122 Chr.2: 86604930-86604930 on Build GRCh38
rs377682850 Chr.2: 86605046-86605046 on Build GRCh38
rs866109212 Chr.2: 86605160-86605160 on Build GRCh38
rs750543175 Chr.2: 86604986-86604986 on Build GRCh38
rs746828990 Chr.2: 86604908-86604908 on Build GRCh38
rs772950406 Chr.2: 86604885-86604885 on Build GRCh38
rs766796913 Chr.2: 86604989-86604989 on Build GRCh38
rs147773943 Chr.2: 86605004-86605004 on Build GRCh38
rs751811518 Chr.2: 86604813-86604813 on Build GRCh38
rs996207000 Chr.2: 86604871-86604871 on Build GRCh38
rs745656674 Chr.2: 86605127-86605127 on Build GRCh38
rs1025555780 Chr.2: 86604799-86604799 on Build GRCh38
rs754553526 Chr.2: 86604853-86604853 on Build GRCh38
rs778613314 Chr.2: 86604996-86604996 on Build GRCh38
rs779767603 Chr.2: 86604708-86604708 on Build GRCh38
rs564118278 Chr.2: 86604731-86604731 on Build GRCh38
rs539733260 Chr.2: 86605144-86605144 on Build GRCh38
rs373564785 Chr.2: 86604681-86604681 on Build GRCh38
rs375660475 Chr.2: 86604755-86604755 on Build GRCh38
rs766338643 Chr.2: 86604791-86604791 on Build GRCh38
rs757781314 Chr.2: 86605010-86605010 on Build GRCh38
rs747792563 Chr.2: 86604866-86604866 on Build GRCh38
rs1004153938 Chr.2: 86605117-86605117 on Build GRCh38
rs566408280 Chr.2: 86605139-86605139 on Build GRCh38
rs771744537 Chr.2: 86604874-86604874 on Build GRCh38
rs151125982 Chr.2: 86605057-86605057 on Build GRCh38
rs773505390 Chr.2: 86605073-86605073 on Build GRCh38
rs896869587 Chr.2: 86604772-86604772 on Build GRCh38
rs753088400 Chr.2: 86605097-86605097 on Build GRCh38
rs748802871 Chr.2: 86604718-86604718 on Build GRCh38
rs780937000 Chr.2: 86605036-86605036 on Build GRCh38
rs777111573 Chr.2: 86605102-86605102 on Build GRCh38
rs754426665 Chr.2: 86604992-86604992 on Build GRCh38
rs756645428 Chr.2: 86604842-86604842 on Build GRCh38
rs770659862 Chr.2: 86604938-86604938 on Build GRCh38
rs779542435 Chr.2: 86604796-86604799 on Build GRCh38
rs924812236 Chr.2: 86604713-86604713 on Build GRCh38
rs201780922 Chr.2: 86604958-86604958 on Build GRCh38
rs760589907 Chr.2: 86604786-86604786 on Build GRCh38
rs531471942 Chr.2: 86604941-86604941 on Build GRCh38
rs768214868 Chr.2: 86604721-86604721 on Build GRCh38
rs749749696 Chr.2: 86604851-86604851 on Build GRCh38
rs756620778 Chr.2: 86604689-86604689 on Build GRCh38
rs764111646 Chr.2: 86604812-86604812 on Build GRCh38
rs770748077 Chr.2: 86605026-86605026 on Build GRCh38
rs774359108 Chr.2: 86605055-86605055 on Build GRCh38
rs750784881 Chr.2: 86604841-86604841 on Build GRCh38
rs989413693 Chr.2: 86604952-86604952 on Build GRCh38
rs202140822 Chr.2: 86604863-86604863 on Build GRCh38
rs780710494 Chr.2: 86604846-86604846 on Build GRCh38
rs746404741 Chr.2: 86604683-86604683 on Build GRCh38
rs749126654 Chr.2: 86605145-86605145 on Build GRCh38
rs535409400 Chr.2: 86605040-86605040 on Build GRCh38
rs763038893 Chr.2: 86604800-86604800 on Build GRCh38
rs958293718 Chr.2: 86604892-86604892 on Build GRCh38
rs888098442 Chr.2: 86604662-86604662 on Build GRCh38
rs773026542 Chr.2: 86604779-86604779 on Build GRCh38
rs950323237 Chr.2: 86604678-86604678 on Build GRCh38
rs757472476 Chr.2: 86604818-86604818 on Build GRCh38
rs61760875 Chr.2: 86605009-86605009 on Build GRCh38
rs376272937 Chr.2: 86605100-86605100 on Build GRCh38
rs769708197 Chr.2: 86605048-86605048 on Build GRCh38
rs767823059 Chr.2: 86604830-86604830 on Build GRCh38
rs765845877 Chr.2: 86605095-86605095 on Build GRCh38
rs765625023 Chr.2: 86604937-86604937 on Build GRCh38
rs746593176 Chr.2: 86605025-86605025 on Build GRCh38
rs767773623 Chr.2: 86604981-86604981 on Build GRCh38
rs745639713 Chr.2: 86604720-86604720 on Build GRCh38
rs147608406 Chr.2: 86604980-86604980 on Build GRCh38
rs760768494 Chr.2: 86605076-86605076 on Build GRCh38
rs751269603 Chr.2: 86605111-86605111 on Build GRCh38
rs746438260 Chr.2: 86605080-86605082 on Build GRCh38

View Full Product Details