Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138557677-138558166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA9
Entrez Gene ID
3313
Gene Name
heat shock protein family A (Hsp70) member 9
Gene Aliases
CRP40, CSA, EVPLS, GRP-75, GRP75, HEL-S-124m, HSPA9B, MOT, MOT2, MTHSP75, PBP74, SAAN, SIDBA4
UniGene
Hs.184233
Gene Chromosome Location
Chr.5: 138554882-138575629 on Build GRCh38
Transcripts
NM_004134.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913181 c.1570A>G p.I524V Chr.5: 138557932-138557932 on Build GRCh38
3850464 c.1592A>G p.H531R Chr.5: 138557910-138557910 on Build GRCh38
6682670 c.1621C>T p.R541C Chr.5: 138557881-138557881 on Build GRCh38
3826851 c.1516-1G>C p.? Chr.5: 138557987-138557987 on Build GRCh38
4585429 c.1532C>A p.A511D Chr.5: 138557970-138557970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750461748 Chr.5: 138557930-138557930 on Build GRCh38
rs778715438 Chr.5: 138557826-138557826 on Build GRCh38
rs774577831 Chr.5: 138557881-138557881 on Build GRCh38
rs766249231 Chr.5: 138558086-138558086 on Build GRCh38
rs1041073492 Chr.5: 138558104-138558104 on Build GRCh38
rs765080012 Chr.5: 138557965-138557965 on Build GRCh38
rs768967239 Chr.5: 138557880-138557880 on Build GRCh38
rs975443706 Chr.5: 138557845-138557845 on Build GRCh38
rs772518104 Chr.5: 138558046-138558046 on Build GRCh38
rs113365132 Chr.5: 138557729-138557729 on Build GRCh38
rs746284977 Chr.5: 138558006-138558006 on Build GRCh38
rs746490648 Chr.5: 138557995-138557995 on Build GRCh38
rs547077078 Chr.5: 138558040-138558040 on Build GRCh38
rs530611408 Chr.5: 138557717-138557717 on Build GRCh38
rs562687071 Chr.5: 138557856-138557856 on Build GRCh38
rs779707180 Chr.5: 138557853-138557854 on Build GRCh38
rs897128920 Chr.5: 138558137-138558137 on Build GRCh38
rs746428354 Chr.5: 138557838-138557838 on Build GRCh38
rs547255093 Chr.5: 138558020-138558021 on Build GRCh38
rs370012281 Chr.5: 138558034-138558034 on Build GRCh38
rs566636367 Chr.5: 138557964-138557964 on Build GRCh38
rs551168802 Chr.5: 138557795-138557795 on Build GRCh38
rs1008219106 Chr.5: 138557698-138557698 on Build GRCh38
rs528617117 Chr.5: 138558039-138558039 on Build GRCh38
rs544185424 Chr.5: 138557824-138557824 on Build GRCh38
rs777400747 Chr.5: 138557835-138557835 on Build GRCh38
rs116771381 Chr.5: 138557805-138557805 on Build GRCh38
rs777494153 Chr.5: 138557981-138557981 on Build GRCh38
rs901762177 Chr.5: 138557733-138557733 on Build GRCh38
rs762000593 Chr.5: 138557889-138557889 on Build GRCh38
rs561206244 Chr.5: 138558108-138558108 on Build GRCh38
rs775983125 Chr.5: 138557844-138557844 on Build GRCh38
rs774116913 Chr.5: 138557986-138557986 on Build GRCh38
rs201894482 Chr.5: 138557977-138557977 on Build GRCh38
rs142315361 Chr.5: 138557748-138557748 on Build GRCh38
rs1044856235 Chr.5: 138557763-138557763 on Build GRCh38
rs368211786 Chr.5: 138557833-138557833 on Build GRCh38
rs1006411646 Chr.5: 138557791-138557791 on Build GRCh38
rs748126703 Chr.5: 138558038-138558040 on Build GRCh38
rs533231313 Chr.5: 138557921-138557921 on Build GRCh38
rs755652959 Chr.5: 138557723-138557723 on Build GRCh38
rs551634500 Chr.5: 138557954-138557954 on Build GRCh38
rs1026164403 Chr.5: 138557754-138557754 on Build GRCh38
rs752447855 Chr.5: 138557974-138557974 on Build GRCh38
rs936839133 Chr.5: 138558072-138558072 on Build GRCh38
rs1035262447 Chr.5: 138557790-138557790 on Build GRCh38
rs754940559 Chr.5: 138558006-138558007 on Build GRCh38
rs770774061 Chr.5: 138557908-138557908 on Build GRCh38
rs557916725 Chr.5: 138557735-138557735 on Build GRCh38
rs770198749 Chr.5: 138557843-138557843 on Build GRCh38
rs951890135 Chr.5: 138557781-138557781 on Build GRCh38
rs747766458 Chr.5: 138557832-138557832 on Build GRCh38
rs756717715 Chr.5: 138558028-138558028 on Build GRCh38
rs930299173 Chr.5: 138558101-138558101 on Build GRCh38
rs944079350 Chr.5: 138558091-138558091 on Build GRCh38
rs753722275 Chr.5: 138557855-138557856 on Build GRCh38
rs767619327 Chr.5: 138557910-138557910 on Build GRCh38
rs911306706 Chr.5: 138558089-138558089 on Build GRCh38
rs766237579 Chr.5: 138557939-138557939 on Build GRCh38
rs755793935 Chr.5: 138557830-138557841 on Build GRCh38
rs1022746390 Chr.5: 138557808-138557808 on Build GRCh38
rs905323182 Chr.5: 138557772-138557772 on Build GRCh38
rs1003722166 Chr.5: 138557782-138557782 on Build GRCh38
rs940367970 Chr.5: 138557704-138557704 on Build GRCh38
rs561726305 Chr.5: 138557769-138557769 on Build GRCh38
rs256001 Chr.5: 138557736-138557736 on Build GRCh38
rs925468967 Chr.5: 138558069-138558069 on Build GRCh38
rs972018925 Chr.5: 138558054-138558054 on Build GRCh38

View Full Product Details