Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63035670-63035934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGH
Entrez Gene ID
8836
Gene Name
gamma-glutamyl hydrolase
Gene Aliases
GH
UniGene
Hs.78619
Gene Chromosome Location
Chr.8: 63015079-63039051 on Build GRCh38
Transcripts
NM_003878.2 XM_011517623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599876 c.140T>C p.V47A Chr.8: 63035740-63035740 on Build GRCh38
6670692 c.128G>A p.C43Y Chr.8: 63035752-63035752 on Build GRCh38
6670691 c.131G>A p.R44H Chr.8: 63035749-63035749 on Build GRCh38
3650149 c.177C>T p.S59S Chr.8: 63035703-63035703 on Build GRCh38
3900936 c.173C>T p.A58V Chr.8: 63035707-63035707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77346539 Chr.8: 63035792-63035792 on Build GRCh38
rs761912749 Chr.8: 63035772-63035774 on Build GRCh38
rs754272209 Chr.8: 63035810-63035812 on Build GRCh38
rs761012028 Chr.8: 63035793-63035793 on Build GRCh38
rs768448681 Chr.8: 63035730-63035730 on Build GRCh38
rs377421757 Chr.8: 63035797-63035797 on Build GRCh38
rs142108552 Chr.8: 63035751-63035751 on Build GRCh38
rs11786893 Chr.8: 63035706-63035706 on Build GRCh38
rs767655894 Chr.8: 63035779-63035779 on Build GRCh38
rs902321577 Chr.8: 63035716-63035716 on Build GRCh38
rs916211005 Chr.8: 63035886-63035886 on Build GRCh38
rs754146055 Chr.8: 63035803-63035803 on Build GRCh38
rs775923482 Chr.8: 63035790-63035790 on Build GRCh38
rs1038179238 Chr.8: 63035765-63035765 on Build GRCh38
rs758036190 Chr.8: 63035709-63035709 on Build GRCh38
rs774365680 Chr.8: 63035742-63035744 on Build GRCh38
rs753468911 Chr.8: 63035704-63035704 on Build GRCh38
rs767556920 Chr.8: 63035914-63035914 on Build GRCh38
rs138112128 Chr.8: 63035750-63035750 on Build GRCh38
rs186979790 Chr.8: 63035806-63035806 on Build GRCh38
rs969135628 Chr.8: 63035864-63035864 on Build GRCh38
rs994062069 Chr.8: 63035778-63035778 on Build GRCh38
rs773316728 Chr.8: 63035778-63035779 on Build GRCh38
rs778696750 Chr.8: 63035707-63035707 on Build GRCh38
rs766646519 Chr.8: 63035797-63035797 on Build GRCh38
rs369357967 Chr.8: 63035701-63035701 on Build GRCh38
rs780864357 Chr.8: 63035738-63035738 on Build GRCh38
rs764384225 Chr.8: 63035690-63035690 on Build GRCh38
rs923240863 Chr.8: 63035694-63035694 on Build GRCh38
rs1032472551 Chr.8: 63035887-63035887 on Build GRCh38
rs772170413 Chr.8: 63035788-63035788 on Build GRCh38
rs112780270 Chr.8: 63035729-63035729 on Build GRCh38
rs192294785 Chr.8: 63035904-63035904 on Build GRCh38
rs746975684 Chr.8: 63035727-63035727 on Build GRCh38
rs200041062 Chr.8: 63035791-63035791 on Build GRCh38
rs979607594 Chr.8: 63035870-63035870 on Build GRCh38
rs780111805 Chr.8: 63035715-63035715 on Build GRCh38
rs939142948 Chr.8: 63035861-63035861 on Build GRCh38
rs116629513 Chr.8: 63035768-63035768 on Build GRCh38
rs146984806 Chr.8: 63035749-63035749 on Build GRCh38

View Full Product Details