Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6536089-6536566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367737 c.2683A>G p.M895V Chr.9: 6536219-6536219 on Build GRCh38
1285167 c.2797A>T p.I933F Chr.9: 6536105-6536105 on Build GRCh38
6861563 c.2705C>T p.T902I Chr.9: 6536197-6536197 on Build GRCh38
6183600 c.2711T>C p.M904T Chr.9: 6536191-6536191 on Build GRCh38
351884 c.2724T>A p.T908T Chr.9: 6536178-6536178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002972671 Chr.9: 6536420-6536420 on Build GRCh38
rs1039258333 Chr.9: 6536250-6536250 on Build GRCh38
rs995473768 Chr.9: 6536449-6536449 on Build GRCh38
rs773149955 Chr.9: 6536380-6536380 on Build GRCh38
rs576346137 Chr.9: 6536411-6536411 on Build GRCh38
rs541140925 Chr.9: 6536483-6536483 on Build GRCh38
rs374022098 Chr.9: 6536160-6536160 on Build GRCh38
rs751716055 Chr.9: 6536325-6536325 on Build GRCh38
rs760098112 Chr.9: 6536263-6536263 on Build GRCh38
rs555578628 Chr.9: 6536457-6536457 on Build GRCh38
rs111627722 Chr.9: 6536423-6536423 on Build GRCh38
rs748060681 Chr.9: 6536361-6536361 on Build GRCh38
rs753133816 Chr.9: 6536271-6536271 on Build GRCh38
rs144937031 Chr.9: 6536172-6536172 on Build GRCh38
rs912380232 Chr.9: 6536277-6536277 on Build GRCh38
rs751250959 Chr.9: 6536284-6536284 on Build GRCh38
rs188269735 Chr.9: 6536188-6536188 on Build GRCh38
rs1044568786 Chr.9: 6536386-6536386 on Build GRCh38
rs759933539 Chr.9: 6536173-6536173 on Build GRCh38
rs965560709 Chr.9: 6536523-6536524 on Build GRCh38
rs149860035 Chr.9: 6536370-6536370 on Build GRCh38
rs773430894 Chr.9: 6536156-6536156 on Build GRCh38
rs766077232 Chr.9: 6536373-6536377 on Build GRCh38
rs923646147 Chr.9: 6536351-6536351 on Build GRCh38
rs554367188 Chr.9: 6536306-6536306 on Build GRCh38
rs1007363577 Chr.9: 6536240-6536240 on Build GRCh38
rs535201181 Chr.9: 6536305-6536305 on Build GRCh38
rs1051347119 Chr.9: 6536448-6536448 on Build GRCh38
rs776573369 Chr.9: 6536245-6536245 on Build GRCh38
rs758046295 Chr.9: 6536198-6536198 on Build GRCh38
rs769670556 Chr.9: 6536143-6536143 on Build GRCh38
rs745769719 Chr.9: 6536129-6536129 on Build GRCh38
rs139982267 Chr.9: 6536154-6536154 on Build GRCh38
rs147762931 Chr.9: 6536208-6536208 on Build GRCh38
rs112921617 Chr.9: 6536392-6536392 on Build GRCh38
rs967408649 Chr.9: 6536330-6536330 on Build GRCh38
rs767463147 Chr.9: 6536481-6536481 on Build GRCh38
rs559884467 Chr.9: 6536534-6536534 on Build GRCh38
rs769606225 Chr.9: 6536218-6536218 on Build GRCh38
rs1026983267 Chr.9: 6536456-6536456 on Build GRCh38
rs765592910 Chr.9: 6536268-6536268 on Build GRCh38
rs781057291 Chr.9: 6536128-6536128 on Build GRCh38
rs770831522 Chr.9: 6536529-6536529 on Build GRCh38
rs867537291 Chr.9: 6536385-6536385 on Build GRCh38
rs1054610342 Chr.9: 6536124-6536124 on Build GRCh38
rs543842730 Chr.9: 6536427-6536427 on Build GRCh38
rs552072250 Chr.9: 6536123-6536123 on Build GRCh38
rs367759326 Chr.9: 6536109-6536109 on Build GRCh38
rs566293238 Chr.9: 6536360-6536360 on Build GRCh38
rs929003450 Chr.9: 6536435-6536435 on Build GRCh38
rs574033879 Chr.9: 6536482-6536482 on Build GRCh38
rs369767254 Chr.9: 6536199-6536199 on Build GRCh38
rs771178748 Chr.9: 6536244-6536244 on Build GRCh38
rs367987650 Chr.9: 6536224-6536224 on Build GRCh38
rs780535258 Chr.9: 6536205-6536205 on Build GRCh38
rs899919301 Chr.9: 6536259-6536259 on Build GRCh38
rs775143096 Chr.9: 6536151-6536151 on Build GRCh38
rs911830298 Chr.9: 6536469-6536469 on Build GRCh38
rs868422652 Chr.9: 6536162-6536162 on Build GRCh38
rs752943013 Chr.9: 6536196-6536196 on Build GRCh38
rs760961076 Chr.9: 6536232-6536232 on Build GRCh38
rs977869142 Chr.9: 6536532-6536532 on Build GRCh38
rs372072307 Chr.9: 6536278-6536278 on Build GRCh38
rs370297328 Chr.9: 6536153-6536153 on Build GRCh38
rs1026865807 Chr.9: 6536248-6536248 on Build GRCh38
rs756807420 Chr.9: 6536201-6536201 on Build GRCh38
rs144939090 Chr.9: 6536229-6536229 on Build GRCh38
rs767018561 Chr.9: 6536167-6536167 on Build GRCh38
rs764417358 Chr.9: 6536283-6536283 on Build GRCh38
rs146939116 Chr.9: 6536228-6536228 on Build GRCh38
rs201994873 Chr.9: 6536368-6536368 on Build GRCh38
rs751089321 Chr.9: 6536200-6536200 on Build GRCh38
rs141152043 Chr.9: 6536219-6536219 on Build GRCh38
rs934940369 Chr.9: 6536122-6536122 on Build GRCh38

View Full Product Details