Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23623013-23623181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALB2
Entrez Gene ID
79728
Gene Name
partner and localizer of BRCA2
Gene Aliases
FANCN, PNCA3
UniGene
Hs.444664
Gene Chromosome Location
Chr.16: 23603162-23641357 on Build GRCh38
Transcripts
NM_024675.3 XM_011545946.2 XM_011545947.2 XM_011545948.2 XM_017023671.1 XM_017023672.1 XM_017023673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6983911 c.2843T>G p.F948C Chr.16: 23623122-23623122 on Build GRCh38
1741986 c.2939G>A p.S980N Chr.16: 23623026-23623026 on Build GRCh38
3361689 c.2846G>C p.C949S Chr.16: 23623119-23623119 on Build GRCh38
4824477 c.2883G>A p.L961L Chr.16: 23623082-23623082 on Build GRCh38
163240 c.2946G>C p.G982G Chr.16: 23623019-23623019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149522412 Chr.16: 23623114-23623114 on Build GRCh38
rs876659945 Chr.16: 23623046-23623046 on Build GRCh38
rs876658154 Chr.16: 23623048-23623048 on Build GRCh38
rs515726103 Chr.16: 23623096-23623096 on Build GRCh38
rs878855113 Chr.16: 23623129-23623129 on Build GRCh38
rs369132015 Chr.16: 23623062-23623062 on Build GRCh38
rs180177126 Chr.16: 23623044-23623045 on Build GRCh38
rs587782889 Chr.16: 23623113-23623113 on Build GRCh38
rs762772267 Chr.16: 23623073-23623073 on Build GRCh38
rs786203248 Chr.16: 23623083-23623083 on Build GRCh38
rs45464500 Chr.16: 23623125-23623125 on Build GRCh38
rs515726100 Chr.16: 23623128-23623128 on Build GRCh38
rs786204248 Chr.16: 23623069-23623069 on Build GRCh38
rs587776421 Chr.16: 23623045-23623046 on Build GRCh38
rs879254015 Chr.16: 23623105-23623105 on Build GRCh38
rs730881892 Chr.16: 23623043-23623043 on Build GRCh38
rs587776420 Chr.16: 23623077-23623077 on Build GRCh38
rs876658818 Chr.16: 23623061-23623061 on Build GRCh38
rs515726102 Chr.16: 23623100-23623100 on Build GRCh38
rs786201384 Chr.16: 23623052-23623052 on Build GRCh38
rs515726101 Chr.16: 23623106-23623106 on Build GRCh38
rs587780214 Chr.16: 23623068-23623068 on Build GRCh38
rs876659693 Chr.16: 23623092-23623092 on Build GRCh38
rs747232850 Chr.16: 23623141-23623141 on Build GRCh38
rs587781280 Chr.16: 23623075-23623075 on Build GRCh38
rs786204122 Chr.16: 23623042-23623045 on Build GRCh38
rs515726099 Chr.16: 23623131-23623131 on Build GRCh38
rs886039613 Chr.16: 23623132-23623132 on Build GRCh38
rs876659983 Chr.16: 23623130-23623130 on Build GRCh38
rs876659160 Chr.16: 23623116-23623116 on Build GRCh38
rs375942535 Chr.16: 23623104-23623104 on Build GRCh38
rs587778588 Chr.16: 23623119-23623119 on Build GRCh38
rs786202813 Chr.16: 23623063-23623063 on Build GRCh38
rs61755166 Chr.16: 23623084-23623084 on Build GRCh38
rs864622525 Chr.16: 23623066-23623066 on Build GRCh38
rs876660352 Chr.16: 23623115-23623115 on Build GRCh38
rs45525135 Chr.16: 23623124-23623124 on Build GRCh38
rs373280855 Chr.16: 23623110-23623110 on Build GRCh38
rs762699642 Chr.16: 23623152-23623153 on Build GRCh38
rs878855115 Chr.16: 23623037-23623037 on Build GRCh38

View Full Product Details