Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7697994-7698396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6104737 c.982-1G>T p.? Chr.5: 7698246-7698246 on Build GRCh38
7354512 c.482A>T p.Y161F Chr.5: 7698287-7698287 on Build GRCh38
7354511 c.1022A>T p.Y341F Chr.5: 7698287-7698287 on Build GRCh38
6104738 c.442-1G>T p.? Chr.5: 7698246-7698246 on Build GRCh38
1244455 c.1067A>G p.K356R Chr.5: 7698332-7698332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038700487 Chr.5: 7698200-7698200 on Build GRCh38
rs756517626 Chr.5: 7698367-7698367 on Build GRCh38
rs775853444 Chr.5: 7698258-7698258 on Build GRCh38
rs749890914 Chr.5: 7698333-7698333 on Build GRCh38
rs969564938 Chr.5: 7698065-7698065 on Build GRCh38
rs776751628 Chr.5: 7698309-7698309 on Build GRCh38
rs960657903 Chr.5: 7698130-7698130 on Build GRCh38
rs1025339374 Chr.5: 7698241-7698241 on Build GRCh38
rs566862176 Chr.5: 7698067-7698067 on Build GRCh38
rs556723445 Chr.5: 7698180-7698180 on Build GRCh38
rs112411265 Chr.5: 7698128-7698128 on Build GRCh38
rs916621619 Chr.5: 7698064-7698064 on Build GRCh38
rs554867288 Chr.5: 7698119-7698119 on Build GRCh38
rs535441293 Chr.5: 7698116-7698116 on Build GRCh38
rs1042971813 Chr.5: 7698142-7698142 on Build GRCh38
rs924943133 Chr.5: 7698191-7698191 on Build GRCh38
rs60394679 Chr.5: 7698169-7698169 on Build GRCh38
rs552842052 Chr.5: 7698126-7698126 on Build GRCh38
rs774445847 Chr.5: 7698211-7698211 on Build GRCh38
rs760607597 Chr.5: 7698259-7698259 on Build GRCh38
rs947275706 Chr.5: 7698129-7698130 on Build GRCh38
rs757990046 Chr.5: 7698351-7698351 on Build GRCh38
rs916724032 Chr.5: 7698042-7698042 on Build GRCh38
rs746096168 Chr.5: 7698226-7698226 on Build GRCh38
rs764675866 Chr.5: 7698314-7698314 on Build GRCh38
rs545883987 Chr.5: 7698354-7698354 on Build GRCh38
rs778307357 Chr.5: 7698201-7698201 on Build GRCh38
rs900050321 Chr.5: 7698240-7698240 on Build GRCh38
rs992366192 Chr.5: 7698150-7698150 on Build GRCh38
rs576575409 Chr.5: 7698282-7698282 on Build GRCh38
rs772273842 Chr.5: 7698233-7698233 on Build GRCh38
rs945104629 Chr.5: 7698170-7698170 on Build GRCh38
rs751197755 Chr.5: 7698366-7698366 on Build GRCh38
rs761770444 Chr.5: 7698313-7698313 on Build GRCh38
rs943078091 Chr.5: 7698193-7698193 on Build GRCh38
rs928101770 Chr.5: 7698085-7698085 on Build GRCh38
rs371727394 Chr.5: 7698208-7698208 on Build GRCh38
rs988180530 Chr.5: 7698115-7698115 on Build GRCh38
rs1023898632 Chr.5: 7698361-7698361 on Build GRCh38
rs935325005 Chr.5: 7698113-7698113 on Build GRCh38

View Full Product Details