Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137174999-137175494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC2
Entrez Gene ID
29882
Gene Name
anaphase promoting complex subunit 2
Gene Aliases
APC2
UniGene
Hs.533262
Gene Chromosome Location
Chr.9: 137174784-137188605 on Build GRCh38
Transcripts
NM_013366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461332 c.2217C>T p.G739G Chr.9: 137175276-137175276 on Build GRCh38
5998400 c.2152G>T p.E718* Chr.9: 137175341-137175341 on Build GRCh38
6599534 c.2179A>G p.M727V Chr.9: 137175314-137175314 on Build GRCh38
7230838 c.2134A>T p.T712S Chr.9: 137175359-137175359 on Build GRCh38
6027732 c.2273T>C p.I758T Chr.9: 137175138-137175138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548010116 Chr.9: 137175378-137175378 on Build GRCh38
rs772081667 Chr.9: 137175332-137175332 on Build GRCh38
rs773256624 Chr.9: 137175334-137175334 on Build GRCh38
rs774223272 Chr.9: 137175284-137175284 on Build GRCh38
rs767854541 Chr.9: 137175472-137175472 on Build GRCh38
rs371949799 Chr.9: 137175406-137175406 on Build GRCh38
rs140009201 Chr.9: 137175402-137175402 on Build GRCh38
rs771525921 Chr.9: 137175197-137175198 on Build GRCh38
rs769457762 Chr.9: 137175260-137175260 on Build GRCh38
rs372115923 Chr.9: 137175279-137175279 on Build GRCh38
rs150004934 Chr.9: 137175372-137175372 on Build GRCh38
rs143591300 Chr.9: 137175423-137175423 on Build GRCh38
rs748662014 Chr.9: 137175078-137175078 on Build GRCh38
rs550173921 Chr.9: 137175429-137175429 on Build GRCh38
rs748020545 Chr.9: 137175416-137175416 on Build GRCh38
rs776318474 Chr.9: 137175143-137175143 on Build GRCh38
rs925783310 Chr.9: 137175444-137175444 on Build GRCh38
rs752606738 Chr.9: 137175029-137175029 on Build GRCh38
rs769365685 Chr.9: 137175212-137175212 on Build GRCh38
rs749120097 Chr.9: 137175265-137175265 on Build GRCh38
rs754682656 Chr.9: 137175410-137175410 on Build GRCh38
rs770523878 Chr.9: 137175135-137175135 on Build GRCh38
rs750489891 Chr.9: 137175177-137175177 on Build GRCh38
rs757792195 Chr.9: 137175305-137175305 on Build GRCh38
rs777919196 Chr.9: 137175424-137175424 on Build GRCh38
rs774404371 Chr.9: 137175442-137175442 on Build GRCh38
rs368688077 Chr.9: 137175476-137175476 on Build GRCh38
rs751800214 Chr.9: 137175303-137175303 on Build GRCh38
rs148909756 Chr.9: 137175198-137175198 on Build GRCh38
rs755587372 Chr.9: 137175316-137175316 on Build GRCh38
rs28617401 Chr.9: 137175239-137175239 on Build GRCh38
rs200409843 Chr.9: 137175246-137175246 on Build GRCh38
rs368445005 Chr.9: 137175399-137175399 on Build GRCh38
rs748469911 Chr.9: 137175197-137175197 on Build GRCh38
rs955216122 Chr.9: 137175367-137175367 on Build GRCh38
rs750906728 Chr.9: 137175387-137175387 on Build GRCh38
rs150560922 Chr.9: 137175362-137175362 on Build GRCh38
rs754079939 Chr.9: 137175186-137175186 on Build GRCh38
rs758808342 Chr.9: 137175233-137175233 on Build GRCh38
rs749274715 Chr.9: 137175438-137175438 on Build GRCh38
rs1000254588 Chr.9: 137175320-137175320 on Build GRCh38
rs764512353 Chr.9: 137175294-137175294 on Build GRCh38
rs775277404 Chr.9: 137175213-137175213 on Build GRCh38
rs775082651 Chr.9: 137175261-137175261 on Build GRCh38
rs770450491 Chr.9: 137175203-137175203 on Build GRCh38
rs762284857 Chr.9: 137175456-137175456 on Build GRCh38
rs574320106 Chr.9: 137175053-137175053 on Build GRCh38
rs143775204 Chr.9: 137175291-137175291 on Build GRCh38
rs998998041 Chr.9: 137175267-137175267 on Build GRCh38
rs768571820 Chr.9: 137175428-137175428 on Build GRCh38
rs756782133 Chr.9: 137175248-137175248 on Build GRCh38
rs376464638 Chr.9: 137175322-137175322 on Build GRCh38
rs868553428 Chr.9: 137175335-137175335 on Build GRCh38
rs765599515 Chr.9: 137175290-137175290 on Build GRCh38
rs1031147138 Chr.9: 137175136-137175136 on Build GRCh38
rs774311433 Chr.9: 137175217-137175217 on Build GRCh38
rs763283049 Chr.9: 137175293-137175293 on Build GRCh38
rs907354773 Chr.9: 137175079-137175079 on Build GRCh38
rs143593793 Chr.9: 137175441-137175441 on Build GRCh38
rs994184647 Chr.9: 137175381-137175381 on Build GRCh38
rs139583307 Chr.9: 137175369-137175369 on Build GRCh38
rs779425627 Chr.9: 137175196-137175196 on Build GRCh38
rs763968524 Chr.9: 137175215-137175215 on Build GRCh38
rs374867166 Chr.9: 137175415-137175415 on Build GRCh38
rs200617883 Chr.9: 137175318-137175318 on Build GRCh38
rs147666205 Chr.9: 137175175-137175175 on Build GRCh38
rs750450958 Chr.9: 137175042-137175042 on Build GRCh38
rs766934387 Chr.9: 137175159-137175159 on Build GRCh38
rs769984093 Chr.9: 137175363-137175363 on Build GRCh38
rs764160995 Chr.9: 137175371-137175371 on Build GRCh38
rs756177245 Chr.9: 137175183-137175183 on Build GRCh38
rs374020459 Chr.9: 137175195-137175195 on Build GRCh38
rs144294113 Chr.9: 137175376-137175376 on Build GRCh38
rs755689385 Chr.9: 137175401-137175401 on Build GRCh38
rs755461909 Chr.9: 137175193-137175193 on Build GRCh38
rs746283285 Chr.9: 137175336-137175338 on Build GRCh38
rs763960927 Chr.9: 137175176-137175176 on Build GRCh38
rs753904277 Chr.9: 137175228-137175228 on Build GRCh38
rs962251880 Chr.9: 137175377-137175377 on Build GRCh38
rs761053045 Chr.9: 137175120-137175120 on Build GRCh38
rs935835153 Chr.9: 137175468-137175468 on Build GRCh38
rs761815181 Chr.9: 137175285-137175285 on Build GRCh38
rs766560856 Chr.9: 137175185-137175185 on Build GRCh38
rs778782491 Chr.9: 137175324-137175324 on Build GRCh38
rs759373951 Chr.9: 137175158-137175158 on Build GRCh38
rs575734306 Chr.9: 137175080-137175080 on Build GRCh38
rs775469575 Chr.9: 137175162-137175162 on Build GRCh38
rs745511782 Chr.9: 137175209-137175209 on Build GRCh38
rs747849564 Chr.9: 137175331-137175331 on Build GRCh38
rs893296599 Chr.9: 137175469-137175469 on Build GRCh38
rs778393633 Chr.9: 137175414-137175414 on Build GRCh38
rs1022216479 Chr.9: 137175173-137175173 on Build GRCh38
rs780273811 Chr.9: 137175047-137175047 on Build GRCh38
rs201343294 Chr.9: 137175288-137175288 on Build GRCh38
rs757381501 Chr.9: 137175037-137175037 on Build GRCh38
rs781501202 Chr.9: 137175306-137175306 on Build GRCh38
rs931065684 Chr.9: 137175214-137175214 on Build GRCh38
rs776201074 Chr.9: 137175204-137175204 on Build GRCh38
rs778195347 Chr.9: 137175243-137175243 on Build GRCh38
rs377727702 Chr.9: 137175218-137175218 on Build GRCh38
rs531577315 Chr.9: 137175253-137175253 on Build GRCh38
rs138212843 Chr.9: 137175065-137175065 on Build GRCh38
rs1000709495 Chr.9: 137175103-137175103 on Build GRCh38
rs781241643 Chr.9: 137175041-137175041 on Build GRCh38
rs373296864 Chr.9: 137175046-137175046 on Build GRCh38
rs369056007 Chr.9: 137175230-137175230 on Build GRCh38
rs776876959 Chr.9: 137175286-137175286 on Build GRCh38
rs368206680 Chr.9: 137175231-137175231 on Build GRCh38
rs544435710 Chr.9: 137175209-137175209 on Build GRCh38
rs780754538 Chr.9: 137175249-137175249 on Build GRCh38
rs766477592 Chr.9: 137175227-137175227 on Build GRCh38
rs375108215 Chr.9: 137175435-137175435 on Build GRCh38
rs147046502 Chr.9: 137175050-137175050 on Build GRCh38
rs746543842 Chr.9: 137175202-137175202 on Build GRCh38

View Full Product Details