Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 29015216-29015751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
UniGene
Hs.175437
Gene Chromosome Location
Chr.1: 28887091-29120046 on Build GRCh38
Transcripts
NM_001166005.1 NM_001166006.1 NM_001166007.1 NM_004437.3 NM_203342.2 NM_203343.2 XM_005245753.1 XM_005245757.1 XM_005245760.1 XM_005245761.1 XM_005245763.1 XM_005245764.1 XM_005245765.1 XM_005245768.1 XM_005245769.1 XM_005245770.1 XM_005245772.4 XM_005245773.4 XM_005245774.1 XM_006710434.1 XM_006710439.1 XM_011540956.1 XM_011540957.1 XM_011540958.1 XM_011540959.1 XM_011540960.1 XM_011540961.1 XM_011540962.1 XM_011540963.2 XM_011540964.1 XM_011540965.2 XM_017000581.1 XM_017000582.1 XM_017000583.1 XM_017000584.1 XM_017000585.1 XM_017000586.1 XM_017000587.1 XM_017000588.1 XM_017000589.1 XM_017000590.1 XM_017000591.1 XM_017000592.1 XM_017000593.1 XM_017000594.1 XM_017000595.1 XM_017000596.1 XM_017000597.1 XM_017000598.1 XM_017000599.1 XM_017000600.1 XM_017000601.1 XM_017000602.1 XM_017000603.1 XM_017000604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5502193 c.835C>G p.P279A Chr.1: 29015697-29015697 on Build GRCh38
908011 c.237T>G p.F79L Chr.1: 29015726-29015726 on Build GRCh38
4031222 c.877C>T p.P293S Chr.1: 29015739-29015739 on Build GRCh38
3487833 c.836C>T p.P279L Chr.1: 29015698-29015698 on Build GRCh38
5502194 c.208C>G p.P70A Chr.1: 29015697-29015697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887563323 Chr.1: 29015329-29015329 on Build GRCh38
rs1030336461 Chr.1: 29015426-29015426 on Build GRCh38
rs878975790 Chr.1: 29015291-29015291 on Build GRCh38
rs397963452 Chr.1: 29015590-29015591 on Build GRCh38
rs941782938 Chr.1: 29015640-29015640 on Build GRCh38
rs776620068 Chr.1: 29015651-29015652 on Build GRCh38
rs373718459 Chr.1: 29015663-29015663 on Build GRCh38
rs574443336 Chr.1: 29015674-29015674 on Build GRCh38
rs754190005 Chr.1: 29015666-29015666 on Build GRCh38
rs926454642 Chr.1: 29015506-29015506 on Build GRCh38
rs1005713811 Chr.1: 29015337-29015337 on Build GRCh38
rs575827466 Chr.1: 29015413-29015413 on Build GRCh38
rs769763668 Chr.1: 29015716-29015716 on Build GRCh38
rs535588329 Chr.1: 29015472-29015472 on Build GRCh38
rs751123600 Chr.1: 29015653-29015653 on Build GRCh38
rs779040931 Chr.1: 29015688-29015688 on Build GRCh38
rs752509295 Chr.1: 29015524-29015524 on Build GRCh38
rs985725008 Chr.1: 29015391-29015391 on Build GRCh38
rs917561987 Chr.1: 29015310-29015310 on Build GRCh38
rs918262632 Chr.1: 29015514-29015514 on Build GRCh38
rs1001999542 Chr.1: 29015284-29015284 on Build GRCh38
rs113959725 Chr.1: 29015578-29015578 on Build GRCh38
rs909611742 Chr.1: 29015416-29015416 on Build GRCh38
rs369165636 Chr.1: 29015681-29015682 on Build GRCh38
rs1047461408 Chr.1: 29015614-29015614 on Build GRCh38
rs555280703 Chr.1: 29015505-29015505 on Build GRCh38
rs928963812 Chr.1: 29015312-29015312 on Build GRCh38
rs1029426770 Chr.1: 29015353-29015353 on Build GRCh38
rs189183599 Chr.1: 29015710-29015710 on Build GRCh38
rs118000762 Chr.1: 29015256-29015256 on Build GRCh38
rs917829274 Chr.1: 29015593-29015593 on Build GRCh38
rs577498012 Chr.1: 29015588-29015591 on Build GRCh38
rs928952472 Chr.1: 29015594-29015594 on Build GRCh38
rs748674481 Chr.1: 29015662-29015662 on Build GRCh38
rs774106664 Chr.1: 29015727-29015727 on Build GRCh38
rs748122076 Chr.1: 29015295-29015296 on Build GRCh38
rs745982851 Chr.1: 29015692-29015692 on Build GRCh38
rs562087940 Chr.1: 29015283-29015283 on Build GRCh38
rs968133195 Chr.1: 29015500-29015500 on Build GRCh38
rs1038576452 Chr.1: 29015341-29015341 on Build GRCh38
rs1010235473 Chr.1: 29015330-29015330 on Build GRCh38
rs559075977 Chr.1: 29015578-29015579 on Build GRCh38
rs1009563974 Chr.1: 29015440-29015440 on Build GRCh38
rs941144909 Chr.1: 29015460-29015460 on Build GRCh38
rs555878912 Chr.1: 29015323-29015323 on Build GRCh38
rs774775309 Chr.1: 29015632-29015632 on Build GRCh38
rs992054863 Chr.1: 29015536-29015536 on Build GRCh38
rs560486448 Chr.1: 29015531-29015531 on Build GRCh38
rs539128123 Chr.1: 29015305-29015305 on Build GRCh38
rs780163558 Chr.1: 29015697-29015697 on Build GRCh38
rs983568858 Chr.1: 29015332-29015332 on Build GRCh38
rs374474256 Chr.1: 29015521-29015521 on Build GRCh38
rs770712506 Chr.1: 29015719-29015719 on Build GRCh38
rs767175256 Chr.1: 29015464-29015464 on Build GRCh38
rs937967509 Chr.1: 29015296-29015296 on Build GRCh38
rs1045895923 Chr.1: 29015261-29015261 on Build GRCh38
rs570138067 Chr.1: 29015280-29015280 on Build GRCh38
rs900063320 Chr.1: 29015374-29015374 on Build GRCh38
rs773195319 Chr.1: 29015717-29015717 on Build GRCh38
rs754471476 Chr.1: 29015655-29015655 on Build GRCh38
rs1047474625 Chr.1: 29015326-29015326 on Build GRCh38
rs572269413 Chr.1: 29015513-29015513 on Build GRCh38
rs762774480 Chr.1: 29015718-29015718 on Build GRCh38
rs184474204 Chr.1: 29015434-29015434 on Build GRCh38
rs753094572 Chr.1: 29015660-29015660 on Build GRCh38
rs540790915 Chr.1: 29015456-29015456 on Build GRCh38
rs756020409 Chr.1: 29015535-29015535 on Build GRCh38
rs1020276844 Chr.1: 29015334-29015334 on Build GRCh38
rs137895267 Chr.1: 29015694-29015694 on Build GRCh38
rs11433402 Chr.1: 29015579-29015580 on Build GRCh38
rs768099144 Chr.1: 29015656-29015656 on Build GRCh38
rs376957446 Chr.1: 29015679-29015679 on Build GRCh38
rs761818534 Chr.1: 29015678-29015679 on Build GRCh38
rs540989021 Chr.1: 29015523-29015523 on Build GRCh38

View Full Product Details