Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14943611-14944156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD5-AS1
Entrez Gene ID
100505641
Gene Name
FGD5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.517821
Gene Chromosome Location
Chr.3: 14942779-14948441 on Build GRCh38
Transcripts
NR_046251.1 NR_046252.1 NR_046253.1 NR_046254.1 NR_046255.1

SNP Detail

SNP ID Chromosome Location
rs996219337 Chr.3: 14943965-14943965 on Build GRCh38
rs948649860 Chr.3: 14943697-14943697 on Build GRCh38
rs114076446 Chr.3: 14943935-14943935 on Build GRCh38
rs1009637188 Chr.3: 14944084-14944084 on Build GRCh38
rs892583409 Chr.3: 14944061-14944061 on Build GRCh38
rs11557128 Chr.3: 14943959-14943959 on Build GRCh38
rs578182964 Chr.3: 14944107-14944107 on Build GRCh38
rs746809471 Chr.3: 14943734-14943734 on Build GRCh38
rs1044329817 Chr.3: 14944016-14944016 on Build GRCh38
rs182494544 Chr.3: 14943720-14943720 on Build GRCh38
rs113490479 Chr.3: 14944001-14944001 on Build GRCh38
rs748179752 Chr.3: 14944095-14944095 on Build GRCh38
rs954502041 Chr.3: 14943767-14943767 on Build GRCh38
rs916871398 Chr.3: 14943816-14943816 on Build GRCh38
rs112786803 Chr.3: 14944056-14944056 on Build GRCh38
rs780823480 Chr.3: 14943806-14943806 on Build GRCh38
rs1016214926 Chr.3: 14943740-14943740 on Build GRCh38
rs543919051 Chr.3: 14944078-14944078 on Build GRCh38
rs116655051 Chr.3: 14943636-14943636 on Build GRCh38
rs540645609 Chr.3: 14944080-14944080 on Build GRCh38
rs771160392 Chr.3: 14943945-14943945 on Build GRCh38
rs777432004 Chr.3: 14943703-14943703 on Build GRCh38
rs547681280 Chr.3: 14943877-14943877 on Build GRCh38
rs900426341 Chr.3: 14943948-14943948 on Build GRCh38
rs149870479 Chr.3: 14943895-14943895 on Build GRCh38
rs114600921 Chr.3: 14943842-14943842 on Build GRCh38
rs574558199 Chr.3: 14943932-14943932 on Build GRCh38
rs984655778 Chr.3: 14943897-14943899 on Build GRCh38
rs974470959 Chr.3: 14943749-14943749 on Build GRCh38
rs368050479 Chr.3: 14944010-14944010 on Build GRCh38
rs999210899 Chr.3: 14944005-14944005 on Build GRCh38
rs961081209 Chr.3: 14943926-14943927 on Build GRCh38
rs116751060 Chr.3: 14944087-14944087 on Build GRCh38
rs908318546 Chr.3: 14943909-14943911 on Build GRCh38
rs954554552 Chr.3: 14943980-14943980 on Build GRCh38
rs549261347 Chr.3: 14943828-14943828 on Build GRCh38
rs939692907 Chr.3: 14943690-14943690 on Build GRCh38
rs936862909 Chr.3: 14943789-14943789 on Build GRCh38
rs187151423 Chr.3: 14944058-14944058 on Build GRCh38
rs1028942889 Chr.3: 14943878-14943878 on Build GRCh38
rs9663 Chr.3: 14943694-14943694 on Build GRCh38
rs147420360 Chr.3: 14943782-14943782 on Build GRCh38
rs1019613844 Chr.3: 14943870-14943870 on Build GRCh38
rs964731144 Chr.3: 14943741-14943741 on Build GRCh38
rs556308747 Chr.3: 14943930-14943930 on Build GRCh38
rs965643163 Chr.3: 14943872-14943872 on Build GRCh38
rs34831984 Chr.3: 14943872-14943873 on Build GRCh38
rs1032432916 Chr.3: 14943757-14943757 on Build GRCh38
rs56138742 Chr.3: 14943890-14943890 on Build GRCh38
rs546577110 Chr.3: 14943760-14943760 on Build GRCh38
rs61746969 Chr.3: 14943696-14943696 on Build GRCh38
rs564454197 Chr.3: 14943670-14943670 on Build GRCh38
rs901165147 Chr.3: 14944090-14944090 on Build GRCh38
rs886256555 Chr.3: 14943904-14943904 on Build GRCh38
rs1044642875 Chr.3: 14943707-14943707 on Build GRCh38
rs76192843 Chr.3: 14943847-14943847 on Build GRCh38
rs1030008645 Chr.3: 14943969-14943969 on Build GRCh38
rs958190921 Chr.3: 14944051-14944051 on Build GRCh38
rs978199634 Chr.3: 14943778-14943778 on Build GRCh38

View Full Product Details