Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 101871893-101872458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA1P5
Entrez Gene ID
94236
Gene Name
DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5
Gene Aliases
HEJ1
UniGene
Hs.606193
Gene Chromosome Location
Chr.1: 101872012-101894743 on Build GRCh38
Transcripts
NR_033424.2 NM_001288823.1 NM_058170.3 NR_110210.1 NR_110211.1

Gene Symbol
OLFM3
Entrez Gene ID
118427
Gene Name
olfactomedin 3
Gene Aliases
NOE3, NOELIN3, OPTIMEDIN
UniGene
Hs.484475
Gene Chromosome Location
Chr.1: 101800028-101997234 on Build GRCh38
Transcripts
NR_033424.2 NM_001288823.1 NM_058170.3 NR_110210.1 NR_110211.1

SNP Detail

SNP ID Chromosome Location
rs367872434 Chr.1: 101872223-101872223 on Build GRCh38
rs906548941 Chr.1: 101871970-101871970 on Build GRCh38
rs928083536 Chr.1: 101872347-101872347 on Build GRCh38
rs954590274 Chr.1: 101872091-101872091 on Build GRCh38
rs948168180 Chr.1: 101871938-101871938 on Build GRCh38
rs1005718492 Chr.1: 101871995-101871995 on Build GRCh38
rs151086958 Chr.1: 101872396-101872396 on Build GRCh38
rs1014561984 Chr.1: 101872007-101872007 on Build GRCh38
rs1045276243 Chr.1: 101872300-101872300 on Build GRCh38
rs10874522 Chr.1: 101872006-101872006 on Build GRCh38
rs973081203 Chr.1: 101872034-101872034 on Build GRCh38
rs1022901909 Chr.1: 101872018-101872018 on Build GRCh38
rs987170154 Chr.1: 101872114-101872114 on Build GRCh38
rs752897065 Chr.1: 101872200-101872200 on Build GRCh38
rs996357292 Chr.1: 101872072-101872072 on Build GRCh38
rs904607040 Chr.1: 101871969-101871969 on Build GRCh38
rs1018686526 Chr.1: 101872386-101872386 on Build GRCh38
rs765469214 Chr.1: 101872152-101872152 on Build GRCh38
rs904533349 Chr.1: 101872199-101872199 on Build GRCh38
rs142131546 Chr.1: 101871989-101871989 on Build GRCh38
rs765293420 Chr.1: 101872410-101872410 on Build GRCh38
rs563191179 Chr.1: 101872400-101872400 on Build GRCh38
rs763418821 Chr.1: 101872231-101872231 on Build GRCh38
rs75710751 Chr.1: 101872033-101872033 on Build GRCh38
rs1043478084 Chr.1: 101871955-101871955 on Build GRCh38
rs1041632496 Chr.1: 101872136-101872136 on Build GRCh38
rs908681172 Chr.1: 101872123-101872123 on Build GRCh38
rs976937409 Chr.1: 101872409-101872409 on Build GRCh38
rs949933264 Chr.1: 101872354-101872365 on Build GRCh38
rs768304033 Chr.1: 101871974-101871974 on Build GRCh38
rs765334728 Chr.1: 101872264-101872267 on Build GRCh38
rs372038730 Chr.1: 101872418-101872418 on Build GRCh38
rs1053147490 Chr.1: 101872202-101872202 on Build GRCh38
rs950445704 Chr.1: 101872099-101872099 on Build GRCh38
rs1028724575 Chr.1: 101872074-101872074 on Build GRCh38
rs1014071896 Chr.1: 101872206-101872206 on Build GRCh38
rs78785807 Chr.1: 101871993-101871993 on Build GRCh38
rs983175205 Chr.1: 101872342-101872342 on Build GRCh38
rs540931617 Chr.1: 101872101-101872101 on Build GRCh38
rs944237104 Chr.1: 101872129-101872129 on Build GRCh38
rs183647180 Chr.1: 101872203-101872203 on Build GRCh38
rs996838669 Chr.1: 101871935-101871935 on Build GRCh38
rs947892322 Chr.1: 101872208-101872208 on Build GRCh38
rs950375449 Chr.1: 101872341-101872341 on Build GRCh38
rs557529462 Chr.1: 101871917-101871917 on Build GRCh38
rs1043057181 Chr.1: 101872184-101872184 on Build GRCh38
rs983203179 Chr.1: 101872116-101872116 on Build GRCh38
rs776697731 Chr.1: 101872048-101872048 on Build GRCh38
rs905773429 Chr.1: 101872235-101872235 on Build GRCh38
rs1027284685 Chr.1: 101872338-101872338 on Build GRCh38
rs532934763 Chr.1: 101872251-101872251 on Build GRCh38
rs759572674 Chr.1: 101872088-101872088 on Build GRCh38
rs554525913 Chr.1: 101872377-101872377 on Build GRCh38
rs923569425 Chr.1: 101872132-101872132 on Build GRCh38
rs1003563897 Chr.1: 101871987-101871987 on Build GRCh38
rs921511674 Chr.1: 101872143-101872143 on Build GRCh38

View Full Product Details