Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46469424-46470022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
-
UniGene
Hs.97414
Gene Chromosome Location
Chr.16: 46469337-46569097 on Build GRCh38
Transcripts
NR_026556.1

SNP Detail

SNP ID Chromosome Location
rs926417409 Chr.16: 46469897-46469897 on Build GRCh38
rs887175743 Chr.16: 46469583-46469583 on Build GRCh38
rs1001537427 Chr.16: 46469481-46469481 on Build GRCh38
rs887194048 Chr.16: 46469973-46469973 on Build GRCh38
rs34571627 Chr.16: 46469582-46469583 on Build GRCh38
rs1025783599 Chr.16: 46469553-46469553 on Build GRCh38
rs1047120655 Chr.16: 46469732-46469732 on Build GRCh38
rs975654711 Chr.16: 46469649-46469649 on Build GRCh38
rs145942052 Chr.16: 46469558-46469558 on Build GRCh38
rs772239086 Chr.16: 46469584-46469584 on Build GRCh38
rs190400316 Chr.16: 46469714-46469714 on Build GRCh38
rs1020160900 Chr.16: 46469805-46469805 on Build GRCh38
rs1008347308 Chr.16: 46469794-46469794 on Build GRCh38
rs982473297 Chr.16: 46469658-46469658 on Build GRCh38
rs187675082 Chr.16: 46469528-46469528 on Build GRCh38
rs896384267 Chr.16: 46469896-46469896 on Build GRCh38
rs973707643 Chr.16: 46469901-46469901 on Build GRCh38
rs564573274 Chr.16: 46469909-46469909 on Build GRCh38
rs913647588 Chr.16: 46469523-46469523 on Build GRCh38
rs937887963 Chr.16: 46469675-46469675 on Build GRCh38
rs997477583 Chr.16: 46469819-46469819 on Build GRCh38
rs148401960 Chr.16: 46469554-46469554 on Build GRCh38
rs1055470302 Chr.16: 46469562-46469562 on Build GRCh38
rs925117096 Chr.16: 46469872-46469872 on Build GRCh38
rs772566346 Chr.16: 46469832-46469832 on Build GRCh38
rs545070185 Chr.16: 46469895-46469895 on Build GRCh38
rs559270780 Chr.16: 46469852-46469852 on Build GRCh38
rs76133883 Chr.16: 46469661-46469661 on Build GRCh38
rs879379258 Chr.16: 46469944-46469944 on Build GRCh38
rs575073011 Chr.16: 46469880-46469880 on Build GRCh38
rs550093389 Chr.16: 46469639-46469639 on Build GRCh38
rs894436808 Chr.16: 46469563-46469563 on Build GRCh38
rs997417065 Chr.16: 46469980-46469980 on Build GRCh38
rs898476422 Chr.16: 46469711-46469711 on Build GRCh38
rs986515879 Chr.16: 46469491-46469491 on Build GRCh38
rs756088486 Chr.16: 46469529-46469529 on Build GRCh38
rs931304842 Chr.16: 46469950-46469950 on Build GRCh38
rs182230817 Chr.16: 46469992-46469992 on Build GRCh38
rs1055016081 Chr.16: 46469888-46469888 on Build GRCh38
rs955685053 Chr.16: 46469729-46469729 on Build GRCh38
rs142393938 Chr.16: 46469556-46469556 on Build GRCh38
rs199773804 Chr.16: 46469826-46469826 on Build GRCh38
rs531948268 Chr.16: 46469935-46469935 on Build GRCh38
rs867035407 Chr.16: 46469542-46469542 on Build GRCh38
rs1032025931 Chr.16: 46469847-46469847 on Build GRCh38
rs887308884 Chr.16: 46469776-46469776 on Build GRCh38
rs1047172422 Chr.16: 46469953-46469953 on Build GRCh38
rs1004230462 Chr.16: 46469965-46469965 on Build GRCh38
rs970590480 Chr.16: 46469894-46469894 on Build GRCh38
rs1016979773 Chr.16: 46469975-46469975 on Build GRCh38
rs540638871 Chr.16: 46469772-46469772 on Build GRCh38
rs117849964 Chr.16: 46469582-46469582 on Build GRCh38
rs566065074 Chr.16: 46469570-46469570 on Build GRCh38
rs937959155 Chr.16: 46469898-46469898 on Build GRCh38
rs957787575 Chr.16: 46469593-46469593 on Build GRCh38
rs117381545 Chr.16: 46469979-46469979 on Build GRCh38
rs555243571 Chr.16: 46469699-46469699 on Build GRCh38
rs577689587 Chr.16: 46469881-46469881 on Build GRCh38
rs913591906 Chr.16: 46469825-46469825 on Build GRCh38
rs1055809942 Chr.16: 46469680-46469680 on Build GRCh38
rs1020709215 Chr.16: 46469883-46469883 on Build GRCh38
rs745871650 Chr.16: 46469718-46469718 on Build GRCh38
rs987865619 Chr.16: 46469622-46469622 on Build GRCh38
rs1004284214 Chr.16: 46469589-46469589 on Build GRCh38
rs963225003 Chr.16: 46469648-46469648 on Build GRCh38
rs1017028947 Chr.16: 46469636-46469636 on Build GRCh38
rs746418604 Chr.16: 46469755-46469755 on Build GRCh38
rs1045696519 Chr.16: 46469952-46469952 on Build GRCh38
rs539900149 Chr.16: 46469573-46469573 on Build GRCh38
rs913761874 Chr.16: 46469645-46469645 on Build GRCh38
rs779223190 Chr.16: 46469723-46469723 on Build GRCh38
rs986460338 Chr.16: 46469799-46469799 on Build GRCh38
rs887123409 Chr.16: 46469962-46469962 on Build GRCh38

View Full Product Details