Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46470181-46470732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
-
UniGene
Hs.97414
Gene Chromosome Location
Chr.16: 46469337-46569097 on Build GRCh38
Transcripts
NR_026556.1

SNP Detail

SNP ID Chromosome Location
rs548061100 Chr.16: 46470580-46470580 on Build GRCh38
rs150004453 Chr.16: 46470614-46470614 on Build GRCh38
rs990599536 Chr.16: 46470484-46470484 on Build GRCh38
rs563408830 Chr.16: 46470531-46470531 on Build GRCh38
rs996962645 Chr.16: 46470286-46470286 on Build GRCh38
rs1000524250 Chr.16: 46470425-46470425 on Build GRCh38
rs866805930 Chr.16: 46470598-46470598 on Build GRCh38
rs139376632 Chr.16: 46470571-46470571 on Build GRCh38
rs891310829 Chr.16: 46470296-46470296 on Build GRCh38
rs555108343 Chr.16: 46470245-46470245 on Build GRCh38
rs908572879 Chr.16: 46470643-46470643 on Build GRCh38
rs995079647 Chr.16: 46470588-46470588 on Build GRCh38
rs959272859 Chr.16: 46470575-46470575 on Build GRCh38
rs921346366 Chr.16: 46470686-46470686 on Build GRCh38
rs932698623 Chr.16: 46470604-46470604 on Build GRCh38
rs1038760881 Chr.16: 46470590-46470590 on Build GRCh38
rs1042612673 Chr.16: 46470653-46470653 on Build GRCh38
rs534895225 Chr.16: 46470404-46470404 on Build GRCh38
rs983308283 Chr.16: 46470619-46470619 on Build GRCh38
rs1027409444 Chr.16: 46470606-46470606 on Build GRCh38
rs866295429 Chr.16: 46470668-46470668 on Build GRCh38
rs751395303 Chr.16: 46470233-46470233 on Build GRCh38
rs1042091180 Chr.16: 46470547-46470547 on Build GRCh38
rs546678001 Chr.16: 46470325-46470325 on Build GRCh38
rs898573881 Chr.16: 46470260-46470260 on Build GRCh38
rs746222514 Chr.16: 46470630-46470630 on Build GRCh38
rs143924899 Chr.16: 46470557-46470557 on Build GRCh38
rs533364312 Chr.16: 46470655-46470655 on Build GRCh38
rs1054081244 Chr.16: 46470447-46470447 on Build GRCh38
rs114196434 Chr.16: 46470439-46470439 on Build GRCh38
rs759605403 Chr.16: 46470374-46470374 on Build GRCh38
rs774103063 Chr.16: 46470454-46470454 on Build GRCh38
rs940035130 Chr.16: 46470236-46470236 on Build GRCh38
rs944195317 Chr.16: 46470651-46470651 on Build GRCh38
rs754135212 Chr.16: 46470561-46470561 on Build GRCh38
rs1032396220 Chr.16: 46470691-46470691 on Build GRCh38
rs1021219498 Chr.16: 46470369-46470369 on Build GRCh38
rs1050335331 Chr.16: 46470294-46470294 on Build GRCh38
rs966333332 Chr.16: 46470380-46470380 on Build GRCh38
rs973376231 Chr.16: 46470278-46470278 on Build GRCh38
rs552725256 Chr.16: 46470424-46470424 on Build GRCh38
rs200270015 Chr.16: 46470685-46470686 on Build GRCh38
rs756246659 Chr.16: 46470500-46470500 on Build GRCh38
rs908520920 Chr.16: 46470366-46470366 on Build GRCh38
rs536856523 Chr.16: 46470227-46470227 on Build GRCh38
rs1033539614 Chr.16: 46470206-46470206 on Build GRCh38
rs907791858 Chr.16: 46470568-46470568 on Build GRCh38
rs540533292 Chr.16: 46470494-46470494 on Build GRCh38
rs117220961 Chr.16: 46470555-46470555 on Build GRCh38
rs1049853837 Chr.16: 46470609-46470609 on Build GRCh38
rs950596888 Chr.16: 46470332-46470332 on Build GRCh38
rs573618054 Chr.16: 46470359-46470359 on Build GRCh38
rs982753814 Chr.16: 46470345-46470345 on Build GRCh38
rs146836676 Chr.16: 46470570-46470570 on Build GRCh38
rs920508151 Chr.16: 46470326-46470326 on Build GRCh38

View Full Product Details