Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46499784-46500335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
-
UniGene
Hs.97414
Gene Chromosome Location
Chr.16: 46469337-46569097 on Build GRCh38
Transcripts
NR_026556.1

SNP Detail

SNP ID Chromosome Location
rs767762706 Chr.16: 46500027-46500027 on Build GRCh38
rs900998013 Chr.16: 46500106-46500106 on Build GRCh38
rs1035568017 Chr.16: 46499899-46499899 on Build GRCh38
rs980181617 Chr.16: 46499853-46499853 on Build GRCh38
rs970155013 Chr.16: 46499833-46499833 on Build GRCh38
rs185069098 Chr.16: 46499819-46499819 on Build GRCh38
rs752816773 Chr.16: 46500063-46500063 on Build GRCh38
rs749309428 Chr.16: 46500300-46500300 on Build GRCh38
rs991266800 Chr.16: 46499966-46499966 on Build GRCh38
rs1021608991 Chr.16: 46500174-46500174 on Build GRCh38
rs577003004 Chr.16: 46499904-46499904 on Build GRCh38
rs932234543 Chr.16: 46500121-46500121 on Build GRCh38
rs756091892 Chr.16: 46500122-46500122 on Build GRCh38
rs569088205 Chr.16: 46500138-46500138 on Build GRCh38
rs536422637 Chr.16: 46500148-46500148 on Build GRCh38
rs571754091 Chr.16: 46499917-46499917 on Build GRCh38
rs181801809 Chr.16: 46499902-46499902 on Build GRCh38
rs1048069600 Chr.16: 46500075-46500075 on Build GRCh38
rs879895984 Chr.16: 46500158-46500158 on Build GRCh38
rs889475849 Chr.16: 46500081-46500081 on Build GRCh38
rs758573635 Chr.16: 46500240-46500240 on Build GRCh38
rs1037212735 Chr.16: 46500012-46500012 on Build GRCh38
rs894706190 Chr.16: 46499951-46499951 on Build GRCh38
rs756438398 Chr.16: 46500005-46500007 on Build GRCh38
rs539133539 Chr.16: 46500026-46500026 on Build GRCh38
rs914612659 Chr.16: 46499818-46499818 on Build GRCh38
rs762717042 Chr.16: 46500280-46500280 on Build GRCh38
rs75597957 Chr.16: 46500163-46500163 on Build GRCh38
rs773669814 Chr.16: 46500011-46500012 on Build GRCh38
rs894365990 Chr.16: 46500261-46500261 on Build GRCh38
rs1006625309 Chr.16: 46500083-46500083 on Build GRCh38
rs546663084 Chr.16: 46499877-46499877 on Build GRCh38
rs554877230 Chr.16: 46500193-46500193 on Build GRCh38
rs1035347877 Chr.16: 46500250-46500250 on Build GRCh38
rs1050593132 Chr.16: 46500151-46500151 on Build GRCh38
rs757085177 Chr.16: 46500195-46500195 on Build GRCh38
rs893297500 Chr.16: 46500164-46500164 on Build GRCh38
rs999367232 Chr.16: 46500111-46500111 on Build GRCh38
rs557444894 Chr.16: 46500135-46500135 on Build GRCh38
rs745581367 Chr.16: 46500259-46500259 on Build GRCh38
rs907988151 Chr.16: 46500002-46500002 on Build GRCh38
rs771710827 Chr.16: 46500268-46500268 on Build GRCh38
rs573038714 Chr.16: 46500286-46500286 on Build GRCh38
rs947123902 Chr.16: 46500175-46500175 on Build GRCh38
rs778660898 Chr.16: 46500244-46500244 on Build GRCh38
rs556346941 Chr.16: 46500287-46500287 on Build GRCh38
rs750348139 Chr.16: 46500099-46500099 on Build GRCh38
rs189484488 Chr.16: 46499872-46499872 on Build GRCh38
rs765272175 Chr.16: 46500067-46500067 on Build GRCh38
rs751871434 Chr.16: 46499976-46499976 on Build GRCh38
rs762398507 Chr.16: 46499974-46499974 on Build GRCh38
rs980150357 Chr.16: 46500242-46500242 on Build GRCh38
rs553425400 Chr.16: 46500116-46500116 on Build GRCh38
rs967458279 Chr.16: 46499825-46499825 on Build GRCh38
rs541299432 Chr.16: 46499973-46499974 on Build GRCh38

View Full Product Details