Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 168249379-168249958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFEM1P
Entrez Gene ID
93556
Gene Name
EGF like and EMI domain containing 1, pseudogene
Gene Aliases
C3orf50, NCRNA00259
UniGene
Hs.478158
Gene Chromosome Location
Chr.3: 168249522-168830599 on Build GRCh38
Transcripts
NR_021485.2

SNP Detail

SNP ID Chromosome Location
rs898090919 Chr.3: 168249916-168249916 on Build GRCh38
rs750352456 Chr.3: 168249623-168249623 on Build GRCh38
rs962198416 Chr.3: 168249555-168249555 on Build GRCh38
rs972245471 Chr.3: 168249731-168249731 on Build GRCh38
rs747714611 Chr.3: 168249670-168249670 on Build GRCh38
rs61740178 Chr.3: 168249703-168249703 on Build GRCh38
rs928008173 Chr.3: 168249719-168249719 on Build GRCh38
rs552616060 Chr.3: 168249701-168249701 on Build GRCh38
rs542511109 Chr.3: 168249899-168249899 on Build GRCh38
rs975068547 Chr.3: 168249466-168249466 on Build GRCh38
rs6769834 Chr.3: 168249776-168249776 on Build GRCh38
rs1041563103 Chr.3: 168249860-168249860 on Build GRCh38
rs377595712 Chr.3: 168249548-168249548 on Build GRCh38
rs1034402458 Chr.3: 168249512-168249512 on Build GRCh38
rs990765480 Chr.3: 168249746-168249746 on Build GRCh38
rs565845279 Chr.3: 168249420-168249420 on Build GRCh38
rs959016959 Chr.3: 168249522-168249522 on Build GRCh38
rs979492883 Chr.3: 168249417-168249417 on Build GRCh38
rs28395217 Chr.3: 168249429-168249429 on Build GRCh38
rs983860814 Chr.3: 168249664-168249664 on Build GRCh38
rs956524401 Chr.3: 168249622-168249623 on Build GRCh38
rs1019415006 Chr.3: 168249827-168249827 on Build GRCh38
rs954844594 Chr.3: 168249758-168249758 on Build GRCh38
rs940838727 Chr.3: 168249688-168249688 on Build GRCh38
rs1054778339 Chr.3: 168249658-168249658 on Build GRCh38
rs908211691 Chr.3: 168249914-168249914 on Build GRCh38
rs949605497 Chr.3: 168249934-168249934 on Build GRCh38
rs375093195 Chr.3: 168249498-168249498 on Build GRCh38
rs561247513 Chr.3: 168249909-168249909 on Build GRCh38
rs894193193 Chr.3: 168249897-168249897 on Build GRCh38
rs993736548 Chr.3: 168249923-168249923 on Build GRCh38
rs571787820 Chr.3: 168249460-168249460 on Build GRCh38
rs972194525 Chr.3: 168249574-168249574 on Build GRCh38
rs914492408 Chr.3: 168249852-168249852 on Build GRCh38
rs370532754 Chr.3: 168249587-168249587 on Build GRCh38
rs894131133 Chr.3: 168249714-168249714 on Build GRCh38
rs181761643 Chr.3: 168249873-168249873 on Build GRCh38
rs1019946323 Chr.3: 168249649-168249649 on Build GRCh38
rs557059509 Chr.3: 168249475-168249475 on Build GRCh38
rs920916505 Chr.3: 168249657-168249657 on Build GRCh38
rs975748965 Chr.3: 168249655-168249655 on Build GRCh38
rs962251175 Chr.3: 168249726-168249726 on Build GRCh38
rs910590625 Chr.3: 168249780-168249780 on Build GRCh38
rs546636435 Chr.3: 168249607-168249607 on Build GRCh38
rs544731273 Chr.3: 168249715-168249715 on Build GRCh38
rs9290335 Chr.3: 168249896-168249896 on Build GRCh38
rs1036610903 Chr.3: 168249566-168249567 on Build GRCh38
rs1054322392 Chr.3: 168249437-168249437 on Build GRCh38
rs905173237 Chr.3: 168249756-168249756 on Build GRCh38
rs892590122 Chr.3: 168249800-168249801 on Build GRCh38
rs928440686 Chr.3: 168249861-168249861 on Build GRCh38
rs959260001 Chr.3: 168249723-168249723 on Build GRCh38
rs989405447 Chr.3: 168249812-168249812 on Build GRCh38
rs892950481 Chr.3: 168249444-168249444 on Build GRCh38
rs539008788 Chr.3: 168249471-168249471 on Build GRCh38
rs1000849868 Chr.3: 168249775-168249775 on Build GRCh38
rs912508779 Chr.3: 168249454-168249454 on Build GRCh38
rs893005680 Chr.3: 168249659-168249659 on Build GRCh38
rs965290768 Chr.3: 168249652-168249652 on Build GRCh38
rs1036491065 Chr.3: 168249695-168249695 on Build GRCh38
rs762662768 Chr.3: 168249507-168249507 on Build GRCh38
rs933375741 Chr.3: 168249602-168249602 on Build GRCh38
rs7614524 Chr.3: 168249887-168249887 on Build GRCh38
rs945942355 Chr.3: 168249859-168249859 on Build GRCh38
rs1041491766 Chr.3: 168249693-168249693 on Build GRCh38
rs144847413 Chr.3: 168249497-168249497 on Build GRCh38
rs1037349603 Chr.3: 168249915-168249915 on Build GRCh38
rs534770593 Chr.3: 168249572-168249572 on Build GRCh38
rs1030473702 Chr.3: 168249744-168249744 on Build GRCh38
rs536533914 Chr.3: 168249543-168249543 on Build GRCh38
rs987998582 Chr.3: 168249449-168249449 on Build GRCh38
rs908328847 Chr.3: 168249678-168249678 on Build GRCh38
rs938484214 Chr.3: 168249877-168249877 on Build GRCh38
rs942654894 Chr.3: 168249656-168249656 on Build GRCh38
rs1009388917 Chr.3: 168249622-168249622 on Build GRCh38
rs532580435 Chr.3: 168249911-168249911 on Build GRCh38
rs965607611 Chr.3: 168249845-168249845 on Build GRCh38
rs905203466 Chr.3: 168249600-168249600 on Build GRCh38
rs986160417 Chr.3: 168249606-168249606 on Build GRCh38
rs1028689088 Chr.3: 168249883-168249883 on Build GRCh38
rs945889969 Chr.3: 168249671-168249671 on Build GRCh38
rs9990407 Chr.3: 168249592-168249592 on Build GRCh38
rs936659684 Chr.3: 168249806-168249806 on Build GRCh38
rs61749505 Chr.3: 168249700-168249700 on Build GRCh38
rs906926508 Chr.3: 168249487-168249487 on Build GRCh38

View Full Product Details