Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23622876-23623048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALB2
Entrez Gene ID
79728
Gene Name
partner and localizer of BRCA2
Gene Aliases
FANCN, PNCA3
UniGene
Hs.444664
Gene Chromosome Location
Chr.16: 23603162-23641357 on Build GRCh38
Transcripts
NM_024675.3 XM_011545946.2 XM_011545947.2 XM_011545948.2 XM_017023671.1 XM_017023672.1 XM_017023673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986919 c.2993G>A p.G998E Chr.16: 23622972-23622972 on Build GRCh38
1741986 c.2939G>A p.S980N Chr.16: 23623026-23623026 on Build GRCh38
5791641 c.2992G>C p.G998R Chr.16: 23622973-23622973 on Build GRCh38
3279827 c.2979G>A p.T993T Chr.16: 23622986-23622986 on Build GRCh38
5783740 c.2996+6A>T p.? Chr.16: 23622963-23622963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180177127 Chr.16: 23622982-23622983 on Build GRCh38
rs876659645 Chr.16: 23622984-23622984 on Build GRCh38
rs587781840 Chr.16: 23623001-23623001 on Build GRCh38
rs886039626 Chr.16: 23622968-23622968 on Build GRCh38
rs61756146 Chr.16: 23622987-23622987 on Build GRCh38
rs762445362 Chr.16: 23623014-23623014 on Build GRCh38
rs772357409 Chr.16: 23622986-23622986 on Build GRCh38
rs587782211 Chr.16: 23623018-23623018 on Build GRCh38
rs773023007 Chr.16: 23622959-23622959 on Build GRCh38
rs745984895 Chr.16: 23622910-23622910 on Build GRCh38
rs786203775 Chr.16: 23622979-23622979 on Build GRCh38
rs180177128 Chr.16: 23622952-23622952 on Build GRCh38
rs875989792 Chr.16: 23622998-23622999 on Build GRCh38
rs775585445 Chr.16: 23623022-23623022 on Build GRCh38
rs1039184417 Chr.16: 23622973-23622973 on Build GRCh38
rs876659931 Chr.16: 23622966-23622966 on Build GRCh38
rs876659036 Chr.16: 23622997-23622997 on Build GRCh38
rs765643734 Chr.16: 23623010-23623010 on Build GRCh38
rs769547308 Chr.16: 23622954-23622954 on Build GRCh38
rs180177129 Chr.16: 23622911-23622911 on Build GRCh38
rs879254193 Chr.16: 23622964-23622964 on Build GRCh38
rs764140747 Chr.16: 23623000-23623000 on Build GRCh38
rs765175665 Chr.16: 23622994-23622994 on Build GRCh38
rs913474031 Chr.16: 23622914-23622914 on Build GRCh38
rs118203999 Chr.16: 23623003-23623003 on Build GRCh38
rs762437762 Chr.16: 23623005-23623005 on Build GRCh38
rs781173873 Chr.16: 23622926-23622926 on Build GRCh38
rs777244673 Chr.16: 23623001-23623001 on Build GRCh38
rs769414858 Chr.16: 23622960-23622960 on Build GRCh38
rs786202892 Chr.16: 23622980-23622980 on Build GRCh38
rs139726856 Chr.16: 23623012-23623012 on Build GRCh38
rs879253986 Chr.16: 23623003-23623004 on Build GRCh38
rs45551636 Chr.16: 23622972-23622972 on Build GRCh38

View Full Product Details