Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208243459-208243627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH1
Entrez Gene ID
3417
Gene Name
isocitrate dehydrogenase (NADP(+)) 1, cytosolic
Gene Aliases
HEL-216, HEL-S-26, IDCD, IDH, IDP, IDPC, PICD
UniGene
Hs.593422
Gene Chromosome Location
Chr.2: 208236227-208255143 on Build GRCh38
Transcripts
NM_001282386.1 NM_001282387.1 NM_005896.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6931477 c.556G>C p.D186H Chr.2: 208243569-208243569 on Build GRCh38
6982566 c.524G>T p.G175V Chr.2: 208243601-208243601 on Build GRCh38
4653429 c.664C>T p.R222C Chr.2: 208243461-208243461 on Build GRCh38
97131 c.532G>A p.V178I Chr.2: 208243593-208243593 on Build GRCh38
6920565 c.577G>T p.A193S Chr.2: 208243548-208243548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772594297 Chr.2: 208243521-208243521 on Build GRCh38
rs34218846 Chr.2: 208243593-208243593 on Build GRCh38
rs748499158 Chr.2: 208243600-208243600 on Build GRCh38
rs186787509 Chr.2: 208243502-208243502 on Build GRCh38
rs182693596 Chr.2: 208243498-208243498 on Build GRCh38
rs370342586 Chr.2: 208243527-208243527 on Build GRCh38
rs374340555 Chr.2: 208243532-208243532 on Build GRCh38
rs374474539 Chr.2: 208243587-208243587 on Build GRCh38
rs951645776 Chr.2: 208243562-208243562 on Build GRCh38
rs781132701 Chr.2: 208243579-208243579 on Build GRCh38
rs748821411 Chr.2: 208243588-208243588 on Build GRCh38
rs781365220 Chr.2: 208243567-208243567 on Build GRCh38
rs757031960 Chr.2: 208243578-208243578 on Build GRCh38
rs62193615 Chr.2: 208243560-208243560 on Build GRCh38
rs750362869 Chr.2: 208243583-208243583 on Build GRCh38
rs762089749 Chr.2: 208243517-208243517 on Build GRCh38
rs773617210 Chr.2: 208243522-208243522 on Build GRCh38
rs756798008 Chr.2: 208243537-208243537 on Build GRCh38
rs772304188 Chr.2: 208243601-208243601 on Build GRCh38
rs778615377 Chr.2: 208243558-208243558 on Build GRCh38
rs587778402 Chr.2: 208243503-208243503 on Build GRCh38
rs768250871 Chr.2: 208243505-208243505 on Build GRCh38
rs753981193 Chr.2: 208243545-208243545 on Build GRCh38
rs773611901 Chr.2: 208243603-208243603 on Build GRCh38
rs774721532 Chr.2: 208243514-208243514 on Build GRCh38
rs34599179 Chr.2: 208243577-208243577 on Build GRCh38
rs149205273 Chr.2: 208243595-208243595 on Build GRCh38

View Full Product Details