Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23613979-23614137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALB2
Entrez Gene ID
79728
Gene Name
partner and localizer of BRCA2
Gene Aliases
FANCN, PNCA3
UniGene
Hs.444664
Gene Chromosome Location
Chr.16: 23603162-23641357 on Build GRCh38
Transcripts
NM_024675.3 XM_011545946.2 XM_011545947.2 XM_011545948.2 XM_017023671.1 XM_017023672.1 XM_017023673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6958329 c.3121A>G p.K1041E Chr.16: 23614084-23614084 on Build GRCh38
6909134 c.3144G>C p.K1048N Chr.16: 23614061-23614061 on Build GRCh38
3507835 c.3166C>T p.Q1056* Chr.16: 23614039-23614039 on Build GRCh38
5752156 c.3201_3201+2delGGT p.? Chr.16: 23614002-23614004 on Build GRCh38
968805 c.3200T>C p.M1067T Chr.16: 23614005-23614005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878855116 Chr.16: 23614051-23614051 on Build GRCh38
rs869312774 Chr.16: 23614019-23614019 on Build GRCh38
rs587780216 Chr.16: 23614073-23614073 on Build GRCh38
rs786203164 Chr.16: 23614001-23614002 on Build GRCh38
rs730881893 Chr.16: 23614014-23614014 on Build GRCh38
rs876658157 Chr.16: 23614040-23614040 on Build GRCh38
rs377713277 Chr.16: 23614077-23614077 on Build GRCh38
rs766394024 Chr.16: 23614075-23614075 on Build GRCh38
rs876660215 Chr.16: 23614001-23614001 on Build GRCh38
rs753307725 Chr.16: 23614096-23614096 on Build GRCh38
rs778244808 Chr.16: 23614107-23614107 on Build GRCh38
rs116967702 Chr.16: 23614026-23614026 on Build GRCh38
rs515726107 Chr.16: 23614062-23614062 on Build GRCh38
rs138273800 Chr.16: 23614059-23614059 on Build GRCh38
rs750579753 Chr.16: 23614069-23614069 on Build GRCh38
rs756415176 Chr.16: 23614101-23614101 on Build GRCh38
rs863224784 Chr.16: 23614033-23614033 on Build GRCh38
rs886039619 Chr.16: 23614092-23614092 on Build GRCh38
rs566549034 Chr.16: 23614079-23614079 on Build GRCh38
rs180177133 Chr.16: 23614089-23614089 on Build GRCh38
rs775290401 Chr.16: 23614050-23614050 on Build GRCh38
rs781663559 Chr.16: 23614083-23614083 on Build GRCh38
rs1021662947 Chr.16: 23614039-23614039 on Build GRCh38
rs587776423 Chr.16: 23614003-23614003 on Build GRCh38
rs587780215 Chr.16: 23614080-23614080 on Build GRCh38
rs587780217 Chr.16: 23614030-23614030 on Build GRCh38
rs967436311 Chr.16: 23614048-23614048 on Build GRCh38
rs778345761 Chr.16: 23614091-23614091 on Build GRCh38
rs760373091 Chr.16: 23614053-23614053 on Build GRCh38

View Full Product Details