Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 29197595-29197719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALK
Entrez Gene ID
238
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Gene Aliases
CD246, NBLST3
UniGene
Hs.654469
Gene Chromosome Location
Chr.2: 29192774-29921611 on Build GRCh38
Transcripts
NM_004304.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3839388 c.4019A>T p.E1340V Chr.2: 29197596-29197596 on Build GRCh38
4661134 c.3966C>A p.I1322I Chr.2: 29197649-29197649 on Build GRCh38
4492252 c.3942C>T p.S1314S Chr.2: 29197673-29197673 on Build GRCh38
6091354 c.3952C>A p.L1318M Chr.2: 29197663-29197663 on Build GRCh38
4419648 c.3939-8A>G p.? Chr.2: 29197684-29197684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771308439 Chr.2: 29197687-29197687 on Build GRCh38
rs540427775 Chr.2: 29197631-29197631 on Build GRCh38
rs3738869 Chr.2: 29197684-29197684 on Build GRCh38
rs770242459 Chr.2: 29197666-29197666 on Build GRCh38
rs200256900 Chr.2: 29197637-29197637 on Build GRCh38
rs370364694 Chr.2: 29197623-29197623 on Build GRCh38
rs749848775 Chr.2: 29197685-29197685 on Build GRCh38
rs779796155 Chr.2: 29197657-29197657 on Build GRCh38
rs748527047 Chr.2: 29197658-29197658 on Build GRCh38
rs758127441 Chr.2: 29197644-29197644 on Build GRCh38
rs867766429 Chr.2: 29197630-29197630 on Build GRCh38
rs56160491 Chr.2: 29197633-29197633 on Build GRCh38

View Full Product Details