Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75298678-75298911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_001286548.1 NM_001286549.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538890.1 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575741 c.320T>C p.V107A Chr.12: 75298772-75298772 on Build GRCh38
4514810 c.988C>T p.R330* Chr.12: 75298890-75298890 on Build GRCh38
6073566 c.1082G>T p.R361L Chr.12: 75298706-75298706 on Build GRCh38
6462953 c.368G>C p.R123T Chr.12: 75298724-75298724 on Build GRCh38
231948 c.325C>T p.P109S Chr.12: 75298767-75298767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759931490 Chr.12: 75298711-75298711 on Build GRCh38
rs753956471 Chr.12: 75298837-75298837 on Build GRCh38
rs752021004 Chr.12: 75298707-75298707 on Build GRCh38
rs1015171171 Chr.12: 75298826-75298826 on Build GRCh38
rs376920121 Chr.12: 75298772-75298772 on Build GRCh38
rs766873465 Chr.12: 75298706-75298706 on Build GRCh38
rs754077802 Chr.12: 75298735-75298735 on Build GRCh38
rs530022576 Chr.12: 75298755-75298755 on Build GRCh38
rs7485576 Chr.12: 75298827-75298827 on Build GRCh38
rs768127337 Chr.12: 75298714-75298714 on Build GRCh38
rs146917896 Chr.12: 75298763-75298763 on Build GRCh38
rs1014171956 Chr.12: 75298866-75298866 on Build GRCh38
rs575686639 Chr.12: 75298794-75298795 on Build GRCh38
rs758384371 Chr.12: 75298775-75298775 on Build GRCh38
rs746886254 Chr.12: 75298758-75298758 on Build GRCh38
rs752262856 Chr.12: 75298842-75298843 on Build GRCh38
rs1053046765 Chr.12: 75298743-75298743 on Build GRCh38
rs372993486 Chr.12: 75298779-75298779 on Build GRCh38
rs566263623 Chr.12: 75298808-75298808 on Build GRCh38
rs777930739 Chr.12: 75298728-75298728 on Build GRCh38
rs764132954 Chr.12: 75298828-75298828 on Build GRCh38
rs770749333 Chr.12: 75298802-75298802 on Build GRCh38
rs949262024 Chr.12: 75298851-75298851 on Build GRCh38
rs757463210 Chr.12: 75298863-75298863 on Build GRCh38
rs776095504 Chr.12: 75298816-75298816 on Build GRCh38
rs373523519 Chr.12: 75298727-75298727 on Build GRCh38
rs200532842 Chr.12: 75298801-75298801 on Build GRCh38
rs185985655 Chr.12: 75298773-75298773 on Build GRCh38
rs137928908 Chr.12: 75298872-75298872 on Build GRCh38
rs752659267 Chr.12: 75298720-75298720 on Build GRCh38
rs887809882 Chr.12: 75298803-75298803 on Build GRCh38
rs917312122 Chr.12: 75298842-75298842 on Build GRCh38
rs767009311 Chr.12: 75298795-75298795 on Build GRCh38
rs754861520 Chr.12: 75298766-75298766 on Build GRCh38
rs377661352 Chr.12: 75298770-75298770 on Build GRCh38
rs558788892 Chr.12: 75298804-75298804 on Build GRCh38
rs765504070 Chr.12: 75298871-75298871 on Build GRCh38
rs138090105 Chr.12: 75298745-75298745 on Build GRCh38
rs200814725 Chr.12: 75298749-75298750 on Build GRCh38

View Full Product Details