Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44331734-44331983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf2
Entrez Gene ID
755
Gene Name
chromosome 21 open reading frame 2
Gene Aliases
LRRC76, YF5/A2
UniGene
Hs.517331
Gene Chromosome Location
Chr.21: 44328944-44339417 on Build GRCh38
Transcripts
NM_001271440.1 NM_001271441.1 NM_001271442.1 NM_004928.2 XM_006724051.3 XM_006724053.3 XM_017028470.1 XM_017028471.1 XM_017028472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1031256 c.408T>C p.S136S Chr.21: 44331980-44331980 on Build GRCh38
4957294 c.427G>A p.A143T Chr.21: 44331961-44331961 on Build GRCh38
1031255 c.459C>T p.G153G Chr.21: 44331929-44331929 on Build GRCh38
1723036 c.443_444delAG p.E148fs*21 Chr.21: 44331944-44331945 on Build GRCh38
3799962 c.535G>C p.E179Q Chr.21: 44331853-44331853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753882358 Chr.21: 44331816-44331816 on Build GRCh38
rs547610646 Chr.21: 44331796-44331796 on Build GRCh38
rs994836989 Chr.21: 44331762-44331762 on Build GRCh38
rs372614395 Chr.21: 44331962-44331962 on Build GRCh38
rs766367624 Chr.21: 44331943-44331943 on Build GRCh38
rs761517826 Chr.21: 44331789-44331789 on Build GRCh38
rs763624843 Chr.21: 44331795-44331795 on Build GRCh38
rs11911832 Chr.21: 44331826-44331826 on Build GRCh38
rs748897535 Chr.21: 44331914-44331914 on Build GRCh38
rs142691279 Chr.21: 44331843-44331843 on Build GRCh38
rs2277809 Chr.21: 44331939-44331939 on Build GRCh38
rs906590447 Chr.21: 44331924-44331924 on Build GRCh38
rs369370304 Chr.21: 44331959-44331959 on Build GRCh38
rs751163294 Chr.21: 44331837-44331837 on Build GRCh38
rs773037448 Chr.21: 44331934-44331934 on Build GRCh38
rs756970705 Chr.21: 44331944-44331947 on Build GRCh38
rs755005560 Chr.21: 44331873-44331873 on Build GRCh38
rs752479202 Chr.21: 44331856-44331856 on Build GRCh38
rs746633371 Chr.21: 44331922-44331952 on Build GRCh38
rs535833529 Chr.21: 44331782-44331782 on Build GRCh38
rs750528434 Chr.21: 44331918-44331918 on Build GRCh38
rs1019407093 Chr.21: 44331810-44331810 on Build GRCh38
rs767299159 Chr.21: 44331844-44331844 on Build GRCh38
rs752395624 Chr.21: 44331895-44331895 on Build GRCh38
rs377579876 Chr.21: 44331928-44331928 on Build GRCh38
rs78738212 Chr.21: 44331883-44331883 on Build GRCh38
rs147363817 Chr.21: 44331860-44331860 on Build GRCh38
rs539517944 Chr.21: 44331808-44331808 on Build GRCh38
rs906734290 Chr.21: 44331768-44331768 on Build GRCh38
rs765226708 Chr.21: 44331846-44331847 on Build GRCh38
rs780666779 Chr.21: 44331944-44331945 on Build GRCh38
rs1017595293 Chr.21: 44331952-44331952 on Build GRCh38
rs9306099 Chr.21: 44331931-44331931 on Build GRCh38
rs139038672 Chr.21: 44331874-44331874 on Build GRCh38
rs773854721 Chr.21: 44331929-44331929 on Build GRCh38
rs568410064 Chr.21: 44331761-44331761 on Build GRCh38
rs767560634 Chr.21: 44331805-44331805 on Build GRCh38
rs377650325 Chr.21: 44331815-44331815 on Build GRCh38
rs747351700 Chr.21: 44331891-44331891 on Build GRCh38
rs781762035 Chr.21: 44331913-44331913 on Build GRCh38
rs777478813 Chr.21: 44331911-44331913 on Build GRCh38
rs764188617 Chr.21: 44331867-44331867 on Build GRCh38
rs550402758 Chr.21: 44331753-44331753 on Build GRCh38
rs761501026 Chr.21: 44331798-44331798 on Build GRCh38
rs186540315 Chr.21: 44331802-44331802 on Build GRCh38
rs1042497272 Chr.21: 44331903-44331903 on Build GRCh38
rs372721170 Chr.21: 44331866-44331866 on Build GRCh38
rs755996355 Chr.21: 44331813-44331813 on Build GRCh38
rs961329460 Chr.21: 44331823-44331823 on Build GRCh38
rs773872732 Chr.21: 44331857-44331857 on Build GRCh38
rs576589166 Chr.21: 44331824-44331824 on Build GRCh38
rs758738754 Chr.21: 44331960-44331960 on Build GRCh38
rs762514398 Chr.21: 44331783-44331783 on Build GRCh38
rs758235839 Chr.21: 44331901-44331912 on Build GRCh38
rs759456714 Chr.21: 44331946-44331946 on Build GRCh38
rs771681892 Chr.21: 44331932-44331932 on Build GRCh38
rs758252505 Chr.21: 44331779-44331779 on Build GRCh38
rs757891132 Chr.21: 44331912-44331912 on Build GRCh38
rs748728864 Chr.21: 44331853-44331853 on Build GRCh38
rs747205833 Chr.21: 44331944-44331944 on Build GRCh38
rs368429321 Chr.21: 44331941-44331941 on Build GRCh38
rs1006203906 Chr.21: 44331793-44331793 on Build GRCh38
rs369935156 Chr.21: 44331849-44331849 on Build GRCh38
rs778317077 Chr.21: 44331890-44331890 on Build GRCh38
rs945352722 Chr.21: 44331892-44331892 on Build GRCh38
rs537750706 Chr.21: 44331836-44331836 on Build GRCh38
rs1003673059 Chr.21: 44331772-44331772 on Build GRCh38
rs767314535 Chr.21: 44331862-44331862 on Build GRCh38
rs778986381 Chr.21: 44331825-44331825 on Build GRCh38
rs985237891 Chr.21: 44331864-44331864 on Build GRCh38
rs778222701 Chr.21: 44331842-44331842 on Build GRCh38
rs200411422 Chr.21: 44331889-44331889 on Build GRCh38

View Full Product Details