Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 13566930-13567191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf48
Entrez Gene ID
157773
Gene Name
chromosome 8 open reading frame 48
Gene Aliases
-
UniGene
Hs.104941
Gene Chromosome Location
Chr.8: 13566843-13568288 on Build GRCh38
Transcripts
NM_001007090.2 XM_017012951.1

Gene Symbol
DLC1
Entrez Gene ID
10395
Gene Name
DLC1 Rho GTPase activating protein
Gene Aliases
ARHGAP7, HP, STARD12, p122-RhoGAP
UniGene
Hs.134296
Gene Chromosome Location
Chr.8: 13083361-13604606 on Build GRCh38
Transcripts
NM_001007090.2 XM_017012951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1096356 c.89A>G p.D30G Chr.8: 13567080-13567080 on Build GRCh38
6223407 c.115A>G p.S39G Chr.8: 13567106-13567106 on Build GRCh38
1096355 c.80A>C p.N27T Chr.8: 13567071-13567071 on Build GRCh38
1455018 c.122C>A p.S41Y Chr.8: 13567113-13567113 on Build GRCh38
4532499 c.188G>A p.S63N Chr.8: 13567179-13567179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974385771 Chr.8: 13567114-13567114 on Build GRCh38
rs562740146 Chr.8: 13567107-13567107 on Build GRCh38
rs532978997 Chr.8: 13567018-13567018 on Build GRCh38
rs1030735153 Chr.8: 13566976-13566976 on Build GRCh38
rs751163792 Chr.8: 13567038-13567038 on Build GRCh38
rs191812233 Chr.8: 13567128-13567128 on Build GRCh38
rs895104102 Chr.8: 13567105-13567105 on Build GRCh38
rs756125886 Chr.8: 13567137-13567137 on Build GRCh38
rs920170626 Chr.8: 13567116-13567116 on Build GRCh38
rs1045157915 Chr.8: 13567151-13567151 on Build GRCh38
rs116418644 Chr.8: 13566977-13566977 on Build GRCh38
rs537888309 Chr.8: 13567020-13567020 on Build GRCh38
rs780137898 Chr.8: 13567143-13567144 on Build GRCh38
rs769191641 Chr.8: 13567154-13567154 on Build GRCh38
rs183770365 Chr.8: 13567025-13567025 on Build GRCh38
rs920136002 Chr.8: 13566978-13566978 on Build GRCh38
rs918024192 Chr.8: 13567135-13567135 on Build GRCh38
rs749094023 Chr.8: 13567122-13567122 on Build GRCh38
rs747398781 Chr.8: 13567141-13567141 on Build GRCh38
rs201250059 Chr.8: 13567056-13567056 on Build GRCh38
rs755998709 Chr.8: 13567090-13567090 on Build GRCh38
rs905173837 Chr.8: 13567040-13567040 on Build GRCh38
rs13273355 Chr.8: 13567074-13567074 on Build GRCh38
rs149488225 Chr.8: 13567093-13567093 on Build GRCh38
rs903538310 Chr.8: 13567151-13567155 on Build GRCh38
rs188494481 Chr.8: 13567091-13567091 on Build GRCh38
rs930128355 Chr.8: 13566979-13566979 on Build GRCh38
rs573686073 Chr.8: 13567092-13567092 on Build GRCh38
rs533439710 Chr.8: 13567120-13567120 on Build GRCh38
rs949471852 Chr.8: 13567143-13567143 on Build GRCh38
rs964306475 Chr.8: 13566974-13566974 on Build GRCh38
rs1047218482 Chr.8: 13566984-13566984 on Build GRCh38
rs772729057 Chr.8: 13567007-13567007 on Build GRCh38
rs542490022 Chr.8: 13567075-13567075 on Build GRCh38
rs775955635 Chr.8: 13567132-13567132 on Build GRCh38
rs553896592 Chr.8: 13567059-13567059 on Build GRCh38
rs767032573 Chr.8: 13567033-13567033 on Build GRCh38
rs577627190 Chr.8: 13567049-13567049 on Build GRCh38
rs964988020 Chr.8: 13567010-13567010 on Build GRCh38
rs1017919591 Chr.8: 13567099-13567099 on Build GRCh38
rs757281084 Chr.8: 13567098-13567098 on Build GRCh38
rs999267114 Chr.8: 13566954-13566954 on Build GRCh38

View Full Product Details