Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41230920-41231183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001278599.1 NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1481108 c.598A>G p.T200A Chr.19: 41230978-41230978 on Build GRCh38
4940365 c.626A>G p.N209S Chr.19: 41231006-41231006 on Build GRCh38
6410084 c.587G>A p.G196E Chr.19: 41230967-41230967 on Build GRCh38
4771125 c.589C>G p.L197V Chr.19: 41230969-41230969 on Build GRCh38
5807236 c.663C>T p.I221I Chr.19: 41231043-41231043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755470829 Chr.19: 41230985-41230985 on Build GRCh38
rs945530203 Chr.19: 41231116-41231116 on Build GRCh38
rs372987501 Chr.19: 41231148-41231148 on Build GRCh38
rs144284099 Chr.19: 41231016-41231016 on Build GRCh38
rs768501355 Chr.19: 41230953-41230953 on Build GRCh38
rs778617899 Chr.19: 41230960-41230960 on Build GRCh38
rs761727401 Chr.19: 41230957-41230957 on Build GRCh38
rs370473880 Chr.19: 41231003-41231003 on Build GRCh38
rs746497465 Chr.19: 41231151-41231151 on Build GRCh38
rs766652458 Chr.19: 41231078-41231078 on Build GRCh38
rs766459840 Chr.19: 41230969-41230969 on Build GRCh38
rs768067241 Chr.19: 41230988-41230988 on Build GRCh38
rs776286865 Chr.19: 41230955-41230955 on Build GRCh38
rs754575803 Chr.19: 41231152-41231152 on Build GRCh38
rs548297374 Chr.19: 41230944-41230944 on Build GRCh38
rs757927991 Chr.19: 41231007-41231007 on Build GRCh38
rs534638184 Chr.19: 41231103-41231103 on Build GRCh38
rs926104806 Chr.19: 41231158-41231158 on Build GRCh38
rs769662670 Chr.19: 41231053-41231053 on Build GRCh38
rs773248925 Chr.19: 41231076-41231076 on Build GRCh38
rs779718472 Chr.19: 41231008-41231008 on Build GRCh38
rs372829908 Chr.19: 41230949-41230949 on Build GRCh38
rs762892654 Chr.19: 41230968-41230968 on Build GRCh38
rs146774960 Chr.19: 41231101-41231101 on Build GRCh38
rs921250564 Chr.19: 41231072-41231072 on Build GRCh38
rs760704353 Chr.19: 41230950-41230950 on Build GRCh38
rs143934164 Chr.19: 41230995-41230995 on Build GRCh38
rs530626566 Chr.19: 41231000-41231000 on Build GRCh38
rs759658392 Chr.19: 41231081-41231081 on Build GRCh38
rs201799923 Chr.19: 41230996-41230996 on Build GRCh38
rs79266409 Chr.19: 41231112-41231112 on Build GRCh38
rs747864633 Chr.19: 41231040-41231040 on Build GRCh38
rs763166316 Chr.19: 41231077-41231077 on Build GRCh38
rs768702995 Chr.19: 41231029-41231029 on Build GRCh38
rs370295348 Chr.19: 41231143-41231143 on Build GRCh38
rs975407816 Chr.19: 41231036-41231036 on Build GRCh38
rs749981843 Chr.19: 41231147-41231147 on Build GRCh38
rs753274836 Chr.19: 41231090-41231090 on Build GRCh38
rs367872631 Chr.19: 41231150-41231150 on Build GRCh38
rs756382142 Chr.19: 41231091-41231091 on Build GRCh38
rs376082526 Chr.19: 41230961-41230961 on Build GRCh38
rs767979238 Chr.19: 41231083-41231083 on Build GRCh38
rs776682499 Chr.19: 41231030-41231030 on Build GRCh38
rs750698876 Chr.19: 41230967-41230967 on Build GRCh38
rs751995110 Chr.19: 41230984-41230984 on Build GRCh38
rs535396112 Chr.19: 41231121-41231121 on Build GRCh38
rs1026304928 Chr.19: 41231100-41231100 on Build GRCh38

View Full Product Details