Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7520675-7520948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850793 c.504C>T p.S168S Chr.5: 7520833-7520833 on Build GRCh38
4441518 c.523G>A p.V175I Chr.5: 7520852-7520852 on Build GRCh38
1717701 c.489C>G p.V163V Chr.5: 7520818-7520818 on Build GRCh38
739225 c.414G>T p.S138S Chr.5: 7520743-7520743 on Build GRCh38
593112 c.513C>T p.I171I Chr.5: 7520842-7520842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761634397 Chr.5: 7520744-7520744 on Build GRCh38
rs749714437 Chr.5: 7520706-7520706 on Build GRCh38
rs113594923 Chr.5: 7520766-7520766 on Build GRCh38
rs776936099 Chr.5: 7520798-7520798 on Build GRCh38
rs760357504 Chr.5: 7520843-7520843 on Build GRCh38
rs752355618 Chr.5: 7520769-7520769 on Build GRCh38
rs915868840 Chr.5: 7520925-7520925 on Build GRCh38
rs748404490 Chr.5: 7520800-7520800 on Build GRCh38
rs752304266 Chr.5: 7520842-7520842 on Build GRCh38
rs368607259 Chr.5: 7520715-7520715 on Build GRCh38
rs375438618 Chr.5: 7520731-7520731 on Build GRCh38
rs139006667 Chr.5: 7520743-7520743 on Build GRCh38
rs772001613 Chr.5: 7520733-7520733 on Build GRCh38
rs770193393 Chr.5: 7520871-7520871 on Build GRCh38
rs754703097 Chr.5: 7520698-7520698 on Build GRCh38
rs138436408 Chr.5: 7520796-7520796 on Build GRCh38
rs767032662 Chr.5: 7520838-7520838 on Build GRCh38
rs779329863 Chr.5: 7520720-7520720 on Build GRCh38
rs13166360 Chr.5: 7520768-7520768 on Build GRCh38
rs769674129 Chr.5: 7520801-7520801 on Build GRCh38
rs575107508 Chr.5: 7520821-7520821 on Build GRCh38
rs961317867 Chr.5: 7520864-7520864 on Build GRCh38
rs368864708 Chr.5: 7520734-7520734 on Build GRCh38
rs966689145 Chr.5: 7520716-7520716 on Build GRCh38
rs775027579 Chr.5: 7520917-7520917 on Build GRCh38
rs765137382 Chr.5: 7520755-7520755 on Build GRCh38
rs773733622 Chr.5: 7520831-7520831 on Build GRCh38
rs771536862 Chr.5: 7520890-7520890 on Build GRCh38
rs765495006 Chr.5: 7520847-7520847 on Build GRCh38
rs758990578 Chr.5: 7520793-7520793 on Build GRCh38
rs386352276 Chr.5: 7520818-7520818 on Build GRCh38
rs188943021 Chr.5: 7520885-7520885 on Build GRCh38
rs966803998 Chr.5: 7520903-7520903 on Build GRCh38
rs866772904 Chr.5: 7520742-7520742 on Build GRCh38
rs748351476 Chr.5: 7520870-7520870 on Build GRCh38
rs372114874 Chr.5: 7520727-7520727 on Build GRCh38
rs765777716 Chr.5: 7520764-7520764 on Build GRCh38
rs749178087 Chr.5: 7520875-7520875 on Build GRCh38
rs758825950 Chr.5: 7520851-7520851 on Build GRCh38
rs150675562 Chr.5: 7520867-7520867 on Build GRCh38
rs199572354 Chr.5: 7520756-7520756 on Build GRCh38
rs746327319 Chr.5: 7520792-7520792 on Build GRCh38
rs202247706 Chr.5: 7520836-7520836 on Build GRCh38
rs755061264 Chr.5: 7520865-7520865 on Build GRCh38
rs779363813 Chr.5: 7520785-7520785 on Build GRCh38
rs777420588 Chr.5: 7520872-7520872 on Build GRCh38
rs779983269 Chr.5: 7520794-7520794 on Build GRCh38
rs371456630 Chr.5: 7520761-7520761 on Build GRCh38
rs752126896 Chr.5: 7520854-7520854 on Build GRCh38
rs34043481 Chr.5: 7520816-7520816 on Build GRCh38
rs778092359 Chr.5: 7520703-7520703 on Build GRCh38
rs759496903 Chr.5: 7520888-7520888 on Build GRCh38
rs774292624 Chr.5: 7520886-7520886 on Build GRCh38
rs200996412 Chr.5: 7520815-7520815 on Build GRCh38
rs780543786 Chr.5: 7520852-7520852 on Build GRCh38
rs773475729 Chr.5: 7520718-7520720 on Build GRCh38
rs141561787 Chr.5: 7520765-7520765 on Build GRCh38
rs1000839964 Chr.5: 7520923-7520923 on Build GRCh38
rs200030249 Chr.5: 7520795-7520795 on Build GRCh38
rs750856333 Chr.5: 7520850-7520850 on Build GRCh38
rs767090582 Chr.5: 7520887-7520887 on Build GRCh38
rs774029455 Chr.5: 7520773-7520773 on Build GRCh38
rs763093770 Chr.5: 7520800-7520801 on Build GRCh38
rs539653924 Chr.5: 7520710-7520710 on Build GRCh38
rs776981954 Chr.5: 7520758-7520758 on Build GRCh38

View Full Product Details