Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67279312-67279585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
UniGene
Hs.657894
Gene Chromosome Location
Chr.15: 67254772-67501804 on Build GRCh38
Transcripts
NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 NM_022784.2 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521914.2 XM_011521915.2 XM_011521916.2 XM_011521917.2 XM_011521918.2 XM_011521922.2 XM_011521923.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4485984 c.299C>T p.S100L Chr.15: 67279424-67279424 on Build GRCh38
4827174 c.313C>G p.Q105E Chr.15: 67279438-67279438 on Build GRCh38
1147591 c.359C>T p.S120L Chr.15: 67279484-67279484 on Build GRCh38
5988263 c.336G>A p.W112* Chr.15: 67279461-67279461 on Build GRCh38
4827173 c.313C>G p.Q105E Chr.15: 67279438-67279438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769079574 Chr.15: 67279403-67279403 on Build GRCh38
rs907671250 Chr.15: 67279521-67279521 on Build GRCh38
rs562515115 Chr.15: 67279511-67279520 on Build GRCh38
rs147540367 Chr.15: 67279510-67279510 on Build GRCh38
rs767364145 Chr.15: 67279459-67279459 on Build GRCh38
rs761970153 Chr.15: 67279406-67279406 on Build GRCh38
rs538240661 Chr.15: 67279445-67279445 on Build GRCh38
rs1034142318 Chr.15: 67279357-67279357 on Build GRCh38
rs774587540 Chr.15: 67279520-67279520 on Build GRCh38
rs974452730 Chr.15: 67279543-67279543 on Build GRCh38
rs866417285 Chr.15: 67279509-67279510 on Build GRCh38
rs759497505 Chr.15: 67279527-67279527 on Build GRCh38
rs755684291 Chr.15: 67279465-67279465 on Build GRCh38
rs752562673 Chr.15: 67279461-67279461 on Build GRCh38
rs267604296 Chr.15: 67279434-67279434 on Build GRCh38
rs546852691 Chr.15: 67279350-67279350 on Build GRCh38
rs911820283 Chr.15: 67279505-67279505 on Build GRCh38
rs780424894 Chr.15: 67279482-67279482 on Build GRCh38
rs975956350 Chr.15: 67279563-67279563 on Build GRCh38
rs747452749 Chr.15: 67279492-67279492 on Build GRCh38
rs776990272 Chr.15: 67279405-67279405 on Build GRCh38
rs780662440 Chr.15: 67279386-67279386 on Build GRCh38
rs753268392 Chr.15: 67279469-67279469 on Build GRCh38
rs199754249 Chr.15: 67279507-67279507 on Build GRCh38
rs191801163 Chr.15: 67279558-67279558 on Build GRCh38
rs766210995 Chr.15: 67279435-67279435 on Build GRCh38
rs752511221 Chr.15: 67279551-67279551 on Build GRCh38
rs757136708 Chr.15: 67279532-67279532 on Build GRCh38
rs1024935412 Chr.15: 67279384-67279384 on Build GRCh38
rs762935869 Chr.15: 67279518-67279518 on Build GRCh38
rs376226738 Chr.15: 67279439-67279439 on Build GRCh38
rs769169643 Chr.15: 67279503-67279503 on Build GRCh38
rs773132185 Chr.15: 67279409-67279409 on Build GRCh38
rs148358395 Chr.15: 67279408-67279408 on Build GRCh38
rs147938779 Chr.15: 67279481-67279481 on Build GRCh38
rs747364778 Chr.15: 67279401-67279401 on Build GRCh38
rs980788759 Chr.15: 67279464-67279464 on Build GRCh38
rs369759374 Chr.15: 67279519-67279519 on Build GRCh38
rs773540039 Chr.15: 67279516-67279516 on Build GRCh38
rs777522555 Chr.15: 67279466-67279466 on Build GRCh38
rs1020731557 Chr.15: 67279426-67279426 on Build GRCh38
rs763141090 Chr.15: 67279429-67279429 on Build GRCh38
rs759550451 Chr.15: 67279458-67279458 on Build GRCh38

View Full Product Details