Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 25546317-25546588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD1
Entrez Gene ID
160492
Gene Name
lamin tail domain containing 1
Gene Aliases
IFLTD1, PAS1C1
UniGene
Hs.44647
Gene Chromosome Location
Chr.12: 25476082-25648604 on Build GRCh38
Transcripts
NM_001145727.2 NM_001145728.2 NM_001145729.1 NM_001256266.1 NM_152590.3 XM_011520575.1 XM_011520576.2 XM_011520577.2 XM_011520578.2 XM_011520579.2 XM_011520580.1 XM_011520582.1 XM_011520583.1 XM_011520584.1 XM_011520585.2 XM_011520587.1 XM_011520588.1 XM_017018891.1 XM_017018892.1 XM_017018893.1 XM_017018894.1 XM_017018895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3871317 c.377C>T p.T126I Chr.12: 25546425-25546425 on Build GRCh38
3459521 c.405G>A p.K135K Chr.12: 25546460-25546460 on Build GRCh38
1360944 c.333G>A p.P111P Chr.12: 25546532-25546532 on Build GRCh38
1360946 c.332C>T p.P111L Chr.12: 25546533-25546533 on Build GRCh38
7401305 c.325T>C p.S109P Chr.12: 25546540-25546540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956390768 Chr.12: 25546473-25546473 on Build GRCh38
rs138534907 Chr.12: 25546502-25546502 on Build GRCh38
rs373748835 Chr.12: 25546525-25546525 on Build GRCh38
rs927843798 Chr.12: 25546408-25546408 on Build GRCh38
rs746131184 Chr.12: 25546363-25546363 on Build GRCh38
rs763438924 Chr.12: 25546442-25546442 on Build GRCh38
rs982000753 Chr.12: 25546393-25546393 on Build GRCh38
rs753196943 Chr.12: 25546349-25546349 on Build GRCh38
rs757469715 Chr.12: 25546357-25546357 on Build GRCh38
rs775718016 Chr.12: 25546435-25546435 on Build GRCh38
rs750617933 Chr.12: 25546472-25546472 on Build GRCh38
rs748051448 Chr.12: 25546379-25546379 on Build GRCh38
rs866793098 Chr.12: 25546552-25546552 on Build GRCh38
rs183429873 Chr.12: 25546350-25546350 on Build GRCh38
rs546165183 Chr.12: 25546444-25546444 on Build GRCh38
rs142943069 Chr.12: 25546527-25546527 on Build GRCh38
rs781540395 Chr.12: 25546362-25546362 on Build GRCh38
rs144583764 Chr.12: 25546343-25546343 on Build GRCh38
rs146886635 Chr.12: 25546410-25546410 on Build GRCh38
rs773262508 Chr.12: 25546385-25546385 on Build GRCh38
rs747884066 Chr.12: 25546487-25546487 on Build GRCh38
rs199940998 Chr.12: 25546353-25546353 on Build GRCh38
rs757379588 Chr.12: 25546447-25546447 on Build GRCh38
rs759206369 Chr.12: 25546418-25546418 on Build GRCh38
rs767736035 Chr.12: 25546460-25546460 on Build GRCh38
rs370130318 Chr.12: 25546522-25546522 on Build GRCh38
rs752160102 Chr.12: 25546445-25546445 on Build GRCh38
rs778587870 Chr.12: 25546375-25546375 on Build GRCh38
rs765577984 Chr.12: 25546433-25546433 on Build GRCh38
rs745455370 Chr.12: 25546547-25546547 on Build GRCh38
rs1012648604 Chr.12: 25546517-25546517 on Build GRCh38
rs867523099 Chr.12: 25546529-25546529 on Build GRCh38
rs746504488 Chr.12: 25546392-25546392 on Build GRCh38
rs756296516 Chr.12: 25546475-25546475 on Build GRCh38
rs915003293 Chr.12: 25546509-25546509 on Build GRCh38
rs764147771 Chr.12: 25546475-25546475 on Build GRCh38
rs775267775 Chr.12: 25546556-25546556 on Build GRCh38
rs535247924 Chr.12: 25546463-25546463 on Build GRCh38
rs143922642 Chr.12: 25546382-25546382 on Build GRCh38
rs564830991 Chr.12: 25546481-25546481 on Build GRCh38
rs367548196 Chr.12: 25546533-25546533 on Build GRCh38
rs746982514 Chr.12: 25546506-25546506 on Build GRCh38
rs770470201 Chr.12: 25546397-25546397 on Build GRCh38
rs369328499 Chr.12: 25546371-25546371 on Build GRCh38
rs764164553 Chr.12: 25546351-25546351 on Build GRCh38
rs769323598 Chr.12: 25546554-25546554 on Build GRCh38
rs1048729578 Chr.12: 25546428-25546428 on Build GRCh38

View Full Product Details