Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16252211-16252480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO42
Entrez Gene ID
54455
Gene Name
F-box protein 42
Gene Aliases
Fbx42, JFK
UniGene
Hs.522384
Gene Chromosome Location
Chr.1: 16246840-16352491 on Build GRCh38
Transcripts
NM_018994.2 XM_006710698.3 XM_017001502.1 XM_017001503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4410099 c.1036C>T p.R346W Chr.1: 16252290-16252290 on Build GRCh38
4951580 c.1017G>A p.L339L Chr.1: 16252309-16252309 on Build GRCh38
377092 c.956C>T p.S319F Chr.1: 16252370-16252370 on Build GRCh38
6662295 c.1015C>T p.L339L Chr.1: 16252311-16252311 on Build GRCh38
4792675 c.1004G>T p.G335V Chr.1: 16252322-16252322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779475583 Chr.1: 16252367-16252367 on Build GRCh38
rs759463754 Chr.1: 16252416-16252421 on Build GRCh38
rs200591476 Chr.1: 16252333-16252333 on Build GRCh38
rs748543088 Chr.1: 16252368-16252368 on Build GRCh38
rs147075222 Chr.1: 16252289-16252289 on Build GRCh38
rs760926611 Chr.1: 16252280-16252280 on Build GRCh38
rs766594413 Chr.1: 16252281-16252281 on Build GRCh38
rs761088705 Chr.1: 16252397-16252397 on Build GRCh38
rs755029570 Chr.1: 16252457-16252458 on Build GRCh38
rs772504331 Chr.1: 16252376-16252376 on Build GRCh38
rs759666723 Chr.1: 16252290-16252290 on Build GRCh38
rs780795697 Chr.1: 16252357-16252357 on Build GRCh38
rs908490550 Chr.1: 16252445-16252445 on Build GRCh38
rs1033159143 Chr.1: 16252295-16252295 on Build GRCh38
rs753629631 Chr.1: 16252446-16252446 on Build GRCh38
rs373513903 Chr.1: 16252324-16252324 on Build GRCh38
rs370845560 Chr.1: 16252407-16252407 on Build GRCh38
rs922644791 Chr.1: 16252343-16252343 on Build GRCh38
rs776764421 Chr.1: 16252413-16252414 on Build GRCh38
rs572191716 Chr.1: 16252427-16252427 on Build GRCh38
rs541154894 Chr.1: 16252440-16252440 on Build GRCh38
rs763981751 Chr.1: 16252451-16252451 on Build GRCh38
rs964182993 Chr.1: 16252325-16252325 on Build GRCh38
rs775253468 Chr.1: 16252455-16252456 on Build GRCh38
rs756864521 Chr.1: 16252349-16252349 on Build GRCh38
rs758219133 Chr.1: 16252318-16252318 on Build GRCh38
rs754753250 Chr.1: 16252448-16252448 on Build GRCh38
rs376146288 Chr.1: 16252359-16252359 on Build GRCh38
rs370270945 Chr.1: 16252456-16252457 on Build GRCh38
rs956441720 Chr.1: 16252415-16252415 on Build GRCh38
rs773667927 Chr.1: 16252377-16252377 on Build GRCh38
rs373918193 Chr.1: 16252423-16252423 on Build GRCh38
rs914328678 Chr.1: 16252420-16252421 on Build GRCh38
rs975504910 Chr.1: 16252326-16252326 on Build GRCh38
rs752694573 Chr.1: 16252447-16252447 on Build GRCh38
rs771327863 Chr.1: 16252404-16252404 on Build GRCh38
rs773494900 Chr.1: 16252279-16252279 on Build GRCh38
rs762299136 Chr.1: 16252239-16252239 on Build GRCh38
rs752571195 Chr.1: 16252294-16252294 on Build GRCh38
rs751399888 Chr.1: 16252454-16252454 on Build GRCh38
rs769176769 Chr.1: 16252363-16252363 on Build GRCh38
rs751234655 Chr.1: 16252338-16252338 on Build GRCh38
rs772255020 Chr.1: 16252265-16252265 on Build GRCh38
rs926829804 Chr.1: 16252450-16252450 on Build GRCh38

View Full Product Details