Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28716384-28716657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874527 c.560G>A p.R187H Chr.8: 28716619-28716619 on Build GRCh38
6180717 c.416A>T p.E139V Chr.8: 28716475-28716475 on Build GRCh38
4162946 c.379G>A p.D127N Chr.8: 28716438-28716438 on Build GRCh38
1098915 c.533G>T p.R178L Chr.8: 28716592-28716592 on Build GRCh38
6657433 c.414C>T p.T138T Chr.8: 28716473-28716473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751265343 Chr.8: 28716543-28716543 on Build GRCh38
rs148681587 Chr.8: 28716582-28716582 on Build GRCh38
rs761359440 Chr.8: 28716428-28716428 on Build GRCh38
rs140029630 Chr.8: 28716515-28716515 on Build GRCh38
rs749286787 Chr.8: 28716571-28716571 on Build GRCh38
rs769580017 Chr.8: 28716592-28716592 on Build GRCh38
rs758041088 Chr.8: 28716434-28716434 on Build GRCh38
rs568445992 Chr.8: 28716446-28716446 on Build GRCh38
rs762558658 Chr.8: 28716536-28716536 on Build GRCh38
rs966098930 Chr.8: 28716526-28716526 on Build GRCh38
rs751884851 Chr.8: 28716447-28716447 on Build GRCh38
rs929998739 Chr.8: 28716549-28716549 on Build GRCh38
rs376071203 Chr.8: 28716529-28716529 on Build GRCh38
rs774744882 Chr.8: 28716495-28716495 on Build GRCh38
rs201375409 Chr.8: 28716619-28716619 on Build GRCh38
rs573052861 Chr.8: 28716532-28716532 on Build GRCh38
rs753849077 Chr.8: 28716412-28716412 on Build GRCh38
rs778345262 Chr.8: 28716560-28716560 on Build GRCh38
rs767173481 Chr.8: 28716626-28716626 on Build GRCh38
rs141908755 Chr.8: 28716557-28716557 on Build GRCh38
rs755362611 Chr.8: 28716449-28716449 on Build GRCh38
rs916779409 Chr.8: 28716435-28716435 on Build GRCh38
rs529663077 Chr.8: 28716590-28716590 on Build GRCh38
rs749842890 Chr.8: 28716533-28716533 on Build GRCh38
rs753235953 Chr.8: 28716556-28716556 on Build GRCh38
rs371856118 Chr.8: 28716551-28716551 on Build GRCh38
rs560576617 Chr.8: 28716583-28716583 on Build GRCh38
rs538938900 Chr.8: 28716585-28716585 on Build GRCh38
rs924846113 Chr.8: 28716530-28716530 on Build GRCh38
rs750083020 Chr.8: 28716433-28716433 on Build GRCh38
rs997984760 Chr.8: 28716586-28716586 on Build GRCh38
rs760497265 Chr.8: 28716496-28716496 on Build GRCh38
rs773992427 Chr.8: 28716601-28716601 on Build GRCh38
rs749401888 Chr.8: 28716474-28716474 on Build GRCh38
rs776674775 Chr.8: 28716527-28716527 on Build GRCh38
rs761680214 Chr.8: 28716591-28716591 on Build GRCh38
rs764690060 Chr.8: 28716429-28716429 on Build GRCh38
rs769913271 Chr.8: 28716473-28716473 on Build GRCh38
rs982329451 Chr.8: 28716497-28716497 on Build GRCh38
rs534156558 Chr.8: 28716452-28716452 on Build GRCh38
rs180762998 Chr.8: 28716430-28716430 on Build GRCh38
rs553322275 Chr.8: 28716486-28716486 on Build GRCh38
rs762349032 Chr.8: 28716598-28716598 on Build GRCh38
rs748643272 Chr.8: 28716456-28716456 on Build GRCh38
rs773224897 Chr.8: 28716597-28716597 on Build GRCh38
rs35677003 Chr.8: 28716572-28716572 on Build GRCh38
rs375243395 Chr.8: 28716552-28716552 on Build GRCh38
rs765913664 Chr.8: 28716599-28716599 on Build GRCh38

View Full Product Details