Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41986949-41987220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G4E
Entrez Gene ID
123745
Gene Name
phospholipase A2 group IVE
Gene Aliases
-
UniGene
Hs.668060
Gene Chromosome Location
Chr.15: 41981582-42050703 on Build GRCh38
Transcripts
NM_001206670.1 XM_011521237.2 XM_011521238.1 XM_011521239.1 NR_120334.1

Gene Symbol
PLA2G4E-AS1
Entrez Gene ID
101928388
Gene Name
PLA2G4E antisense RNA 1
Gene Aliases
-
UniGene
Hs.738043
Gene Chromosome Location
Chr.15: 41972763-41999094 on Build GRCh38
Transcripts
NM_001206670.1 XM_011521237.2 XM_011521238.1 XM_011521239.1 NR_120334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338466 c.1948+7C>A p.? Chr.15: 41987165-41987165 on Build GRCh38
5822829 c.1942T>C p.W648R Chr.15: 41987178-41987178 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs982152047 Chr.15: 41987050-41987050 on Build GRCh38
rs1023846717 Chr.15: 41986981-41986981 on Build GRCh38
rs1019364950 Chr.15: 41987132-41987132 on Build GRCh38
rs771630863 Chr.15: 41987123-41987123 on Build GRCh38
rs534176489 Chr.15: 41987055-41987055 on Build GRCh38
rs12913689 Chr.15: 41987183-41987183 on Build GRCh38
rs1001093928 Chr.15: 41987026-41987026 on Build GRCh38
rs577173962 Chr.15: 41987177-41987177 on Build GRCh38
rs75677999 Chr.15: 41987151-41987151 on Build GRCh38
rs774962061 Chr.15: 41987146-41987146 on Build GRCh38
rs770589897 Chr.15: 41987191-41987191 on Build GRCh38
rs866229200 Chr.15: 41987167-41987167 on Build GRCh38
rs969329044 Chr.15: 41987013-41987013 on Build GRCh38
rs1035495525 Chr.15: 41987060-41987060 on Build GRCh38
rs12913681 Chr.15: 41987157-41987157 on Build GRCh38
rs892594214 Chr.15: 41987080-41987080 on Build GRCh38
rs1009736436 Chr.15: 41987114-41987114 on Build GRCh38
rs1032940908 Chr.15: 41987069-41987069 on Build GRCh38
rs777487483 Chr.15: 41987012-41987012 on Build GRCh38
rs763699482 Chr.15: 41987172-41987172 on Build GRCh38
rs975079220 Chr.15: 41987144-41987144 on Build GRCh38
rs962179421 Chr.15: 41987067-41987067 on Build GRCh38
rs965081966 Chr.15: 41987137-41987137 on Build GRCh38

View Full Product Details