Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14752226-14752455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
UniGene
Hs.531619
Gene Chromosome Location
Chr.19: 14732697-14778541 on Build GRCh38
Transcripts
NM_001271052.1 NM_013447.3 XM_011527948.2 XM_011527949.2 XM_011527951.2 XM_011527952.2 XM_011527953.2 XM_011527954.2 XM_011527955.2 XM_017026726.1 XM_017026727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112890 c.1727C>T p.S576L Chr.19: 14752390-14752390 on Build GRCh38
7130107 c.1675C>T p.L559F Chr.19: 14752442-14752442 on Build GRCh38
4074869 c.1787A>C p.K596T Chr.19: 14752330-14752330 on Build GRCh38
302132 c.1742T>A p.L581Q Chr.19: 14752375-14752375 on Build GRCh38
6575074 c.1788+6G>A p.? Chr.19: 14752323-14752323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200405970 Chr.19: 14752317-14752317 on Build GRCh38
rs774185123 Chr.19: 14752386-14752386 on Build GRCh38
rs755340247 Chr.19: 14752346-14752346 on Build GRCh38
rs184841520 Chr.19: 14752318-14752318 on Build GRCh38
rs201874467 Chr.19: 14752390-14752390 on Build GRCh38
rs912771550 Chr.19: 14752260-14752260 on Build GRCh38
rs748886893 Chr.19: 14752365-14752365 on Build GRCh38
rs755213122 Chr.19: 14752412-14752412 on Build GRCh38
rs778688628 Chr.19: 14752353-14752353 on Build GRCh38
rs554888943 Chr.19: 14752308-14752308 on Build GRCh38
rs762866983 Chr.19: 14752299-14752299 on Build GRCh38
rs773946291 Chr.19: 14752261-14752261 on Build GRCh38
rs1000161792 Chr.19: 14752274-14752274 on Build GRCh38
rs766097193 Chr.19: 14752370-14752370 on Build GRCh38
rs760766279 Chr.19: 14752391-14752391 on Build GRCh38
rs765585011 Chr.19: 14752423-14752423 on Build GRCh38
rs148074899 Chr.19: 14752337-14752337 on Build GRCh38
rs769543174 Chr.19: 14752289-14752289 on Build GRCh38
rs116349625 Chr.19: 14752319-14752319 on Build GRCh38
rs62122600 Chr.19: 14752338-14752338 on Build GRCh38
rs766379780 Chr.19: 14752393-14752393 on Build GRCh38
rs572377645 Chr.19: 14752281-14752281 on Build GRCh38
rs768352866 Chr.19: 14752384-14752384 on Build GRCh38
rs1046566114 Chr.19: 14752250-14752250 on Build GRCh38
rs187229047 Chr.19: 14752408-14752408 on Build GRCh38
rs903730021 Chr.19: 14752331-14752331 on Build GRCh38
rs775317872 Chr.19: 14752358-14752358 on Build GRCh38
rs534018804 Chr.19: 14752361-14752362 on Build GRCh38
rs375427337 Chr.19: 14752408-14752424 on Build GRCh38
rs147337375 Chr.19: 14752348-14752348 on Build GRCh38
rs756366651 Chr.19: 14752332-14752332 on Build GRCh38
rs774156877 Chr.19: 14752316-14752316 on Build GRCh38
rs753072426 Chr.19: 14752425-14752425 on Build GRCh38
rs370041623 Chr.19: 14752435-14752435 on Build GRCh38
rs377307217 Chr.19: 14752355-14752355 on Build GRCh38
rs201378411 Chr.19: 14752290-14752290 on Build GRCh38
rs748659544 Chr.19: 14752350-14752350 on Build GRCh38
rs761725741 Chr.19: 14752389-14752389 on Build GRCh38
rs778412618 Chr.19: 14752426-14752427 on Build GRCh38
rs1030557530 Chr.19: 14752328-14752328 on Build GRCh38
rs758937302 Chr.19: 14752351-14752351 on Build GRCh38
rs776193709 Chr.19: 14752279-14752279 on Build GRCh38
rs969176734 Chr.19: 14752380-14752380 on Build GRCh38

View Full Product Details