Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41650672-41650945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC6
Entrez Gene ID
2547
Gene Name
X-ray repair cross complementing 6
Gene Aliases
CTC75, CTCBF, G22P1, KU70, ML8, TLAA
UniGene
Hs.292493
Gene Chromosome Location
Chr.22: 41621163-41664048 on Build GRCh38
Transcripts
NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3153559 c.973C>T p.R325C Chr.22: 41650735-41650735 on Build GRCh38
4606194 c.982A>G p.I328V Chr.22: 41650744-41650744 on Build GRCh38
1714428 c.1020T>A p.F340L Chr.22: 41650782-41650782 on Build GRCh38
5092229 c.1076A>G p.H359R Chr.22: 41650838-41650838 on Build GRCh38
6824152 c.1040T>G p.L347R Chr.22: 41650802-41650802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753708017 Chr.22: 41650908-41650908 on Build GRCh38
rs751455021 Chr.22: 41650771-41650771 on Build GRCh38
rs752593550 Chr.22: 41650904-41650904 on Build GRCh38
rs372832371 Chr.22: 41650920-41650921 on Build GRCh38
rs757863806 Chr.22: 41650747-41650747 on Build GRCh38
rs943053443 Chr.22: 41650783-41650783 on Build GRCh38
rs368334650 Chr.22: 41650863-41650863 on Build GRCh38
rs373277330 Chr.22: 41650895-41650895 on Build GRCh38
rs562297881 Chr.22: 41650880-41650880 on Build GRCh38
rs771094973 Chr.22: 41650817-41650817 on Build GRCh38
rs376188135 Chr.22: 41650803-41650803 on Build GRCh38
rs373247479 Chr.22: 41650896-41650896 on Build GRCh38
rs373679830 Chr.22: 41650901-41650901 on Build GRCh38
rs777033001 Chr.22: 41650731-41650731 on Build GRCh38
rs201816814 Chr.22: 41650857-41650857 on Build GRCh38
rs780551827 Chr.22: 41650920-41650920 on Build GRCh38
rs377078752 Chr.22: 41650903-41650903 on Build GRCh38
rs144313964 Chr.22: 41650806-41650806 on Build GRCh38
rs369839566 Chr.22: 41650762-41650762 on Build GRCh38
rs761413214 Chr.22: 41650864-41650864 on Build GRCh38
rs955740497 Chr.22: 41650691-41650691 on Build GRCh38
rs148749706 Chr.22: 41650837-41650837 on Build GRCh38
rs750047356 Chr.22: 41650725-41650725 on Build GRCh38
rs373568942 Chr.22: 41650798-41650798 on Build GRCh38
rs764936129 Chr.22: 41650708-41650708 on Build GRCh38
rs772870059 Chr.22: 41650881-41650881 on Build GRCh38
rs949957452 Chr.22: 41650717-41650717 on Build GRCh38
rs201453288 Chr.22: 41650906-41650906 on Build GRCh38
rs369620210 Chr.22: 41650892-41650892 on Build GRCh38
rs779334920 Chr.22: 41650912-41650912 on Build GRCh38
rs747490228 Chr.22: 41650923-41650923 on Build GRCh38
rs754875330 Chr.22: 41650778-41650778 on Build GRCh38
rs550596546 Chr.22: 41650818-41650818 on Build GRCh38
rs761032323 Chr.22: 41650866-41650866 on Build GRCh38
rs368293813 Chr.22: 41650701-41650701 on Build GRCh38
rs764939773 Chr.22: 41650879-41650879 on Build GRCh38
rs750011301 Chr.22: 41650909-41650909 on Build GRCh38
rs772280657 Chr.22: 41650919-41650919 on Build GRCh38
rs376623627 Chr.22: 41650707-41650707 on Build GRCh38
rs762466875 Chr.22: 41650709-41650711 on Build GRCh38
rs28384756 Chr.22: 41650922-41650923 on Build GRCh38
rs567784176 Chr.22: 41650918-41650918 on Build GRCh38
rs533107132 Chr.22: 41650900-41650900 on Build GRCh38
rs1041916235 Chr.22: 41650755-41650755 on Build GRCh38
rs79256654 Chr.22: 41650696-41650696 on Build GRCh38
rs781168212 Chr.22: 41650789-41650789 on Build GRCh38

View Full Product Details