Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201289492-201289739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CR12
Entrez Gene ID
130540
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 12
Gene Aliases
-
UniGene
Hs.107944
Gene Chromosome Location
Chr.2: 201288271-201357398 on Build GRCh38
Transcripts
NM_001127391.2 NM_001289993.1 NM_139163.3 NR_110620.1 XM_011510606.2 XM_011510610.2 XM_011510612.2 XM_017003360.1 XM_017003362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182646 c.1159G>T p.E387* Chr.2: 201289509-201289509 on Build GRCh38
5465359 c.1066A>T p.N356Y Chr.2: 201289731-201289731 on Build GRCh38
4788193 c.1062C>A p.N354K Chr.2: 201289735-201289735 on Build GRCh38
6283563 c.1086G>T p.L362F Chr.2: 201289711-201289711 on Build GRCh38
4661321 c.1170G>A p.K390K Chr.2: 201289498-201289498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779956947 Chr.2: 201289681-201289681 on Build GRCh38
rs776140847 Chr.2: 201289593-201289593 on Build GRCh38
rs1037238608 Chr.2: 201289698-201289698 on Build GRCh38
rs183137242 Chr.2: 201289553-201289553 on Build GRCh38
rs751547224 Chr.2: 201289653-201289653 on Build GRCh38
rs757422311 Chr.2: 201289549-201289549 on Build GRCh38
rs1050883768 Chr.2: 201289641-201289641 on Build GRCh38
rs772602053 Chr.2: 201289587-201289587 on Build GRCh38
rs749690770 Chr.2: 201289578-201289578 on Build GRCh38
rs6715284 Chr.2: 201289674-201289674 on Build GRCh38
rs762840366 Chr.2: 201289604-201289604 on Build GRCh38
rs370519404 Chr.2: 201289710-201289710 on Build GRCh38
rs1054329280 Chr.2: 201289600-201289600 on Build GRCh38
rs767415535 Chr.2: 201289672-201289672 on Build GRCh38
rs1053216005 Chr.2: 201289630-201289630 on Build GRCh38
rs769747993 Chr.2: 201289561-201289561 on Build GRCh38
rs780194249 Chr.2: 201289577-201289577 on Build GRCh38
rs568573975 Chr.2: 201289697-201289697 on Build GRCh38
rs764042410 Chr.2: 201289618-201289618 on Build GRCh38
rs746144112 Chr.2: 201289560-201289560 on Build GRCh38
rs1003863443 Chr.2: 201289696-201289696 on Build GRCh38
rs370544357 Chr.2: 201289657-201289657 on Build GRCh38
rs762241522 Chr.2: 201289584-201289584 on Build GRCh38
rs538408642 Chr.2: 201289559-201289559 on Build GRCh38
rs774829934 Chr.2: 201289667-201289667 on Build GRCh38
rs775074893 Chr.2: 201289603-201289603 on Build GRCh38
rs756361447 Chr.2: 201289676-201289676 on Build GRCh38
rs769316408 Chr.2: 201289579-201289579 on Build GRCh38
rs774804206 Chr.2: 201289582-201289582 on Build GRCh38
rs764596585 Chr.2: 201289602-201289602 on Build GRCh38
rs759293586 Chr.2: 201289594-201289594 on Build GRCh38
rs751390013 Chr.2: 201289531-201289531 on Build GRCh38
rs147923243 Chr.2: 201289707-201289707 on Build GRCh38
rs149151936 Chr.2: 201289524-201289524 on Build GRCh38

View Full Product Details