Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240596654-240596889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN10
Entrez Gene ID
11132
Gene Name
calpain 10
Gene Aliases
CANP10, NIDDM1
UniGene
Hs.728234
Gene Chromosome Location
Chr.2: 240586716-240599109 on Build GRCh38
Transcripts
NM_023083.3 NM_023085.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
720907 c.1671C>T p.V557V Chr.2: 240596870-240596870 on Build GRCh38
2909706 c.1618A>C p.T540P Chr.2: 240596817-240596817 on Build GRCh38
4965751 c.1643T>A p.V548D Chr.2: 240596842-240596842 on Build GRCh38
720908 c.1513C>T p.P505S Chr.2: 240596712-240596712 on Build GRCh38
4092877 c.1483G>A p.A495T Chr.2: 240596682-240596682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190581696 Chr.2: 240596810-240596810 on Build GRCh38
rs768253140 Chr.2: 240596769-240596769 on Build GRCh38
rs763737919 Chr.2: 240596712-240596712 on Build GRCh38
rs768412786 Chr.2: 240596861-240596861 on Build GRCh38
rs765667275 Chr.2: 240596790-240596790 on Build GRCh38
rs573291438 Chr.2: 240596714-240596714 on Build GRCh38
rs754341912 Chr.2: 240596717-240596717 on Build GRCh38
rs745937394 Chr.2: 240596840-240596840 on Build GRCh38
rs749003896 Chr.2: 240596714-240596715 on Build GRCh38
rs372483139 Chr.2: 240596747-240596747 on Build GRCh38
rs376990183 Chr.2: 240596775-240596775 on Build GRCh38
rs373937254 Chr.2: 240596819-240596819 on Build GRCh38
rs370128354 Chr.2: 240596732-240596732 on Build GRCh38
rs17846979 Chr.2: 240596846-240596846 on Build GRCh38
rs369630719 Chr.2: 240596715-240596715 on Build GRCh38
rs550974583 Chr.2: 240596766-240596766 on Build GRCh38
rs757476502 Chr.2: 240596745-240596745 on Build GRCh38
rs201157354 Chr.2: 240596862-240596862 on Build GRCh38
rs200361959 Chr.2: 240596730-240596730 on Build GRCh38
rs760085751 Chr.2: 240596786-240596786 on Build GRCh38
rs529342399 Chr.2: 240596751-240596751 on Build GRCh38
rs768321905 Chr.2: 240596708-240596708 on Build GRCh38
rs951438183 Chr.2: 240596755-240596755 on Build GRCh38
rs540417767 Chr.2: 240596728-240596728 on Build GRCh38
rs759430755 Chr.2: 240596682-240596682 on Build GRCh38
rs767033655 Chr.2: 240596782-240596782 on Build GRCh38
rs942942234 Chr.2: 240596774-240596774 on Build GRCh38
rs751806987 Chr.2: 240596813-240596813 on Build GRCh38
rs747787989 Chr.2: 240596865-240596865 on Build GRCh38
rs758863856 Chr.2: 240596798-240596798 on Build GRCh38
rs771103112 Chr.2: 240596740-240596740 on Build GRCh38
rs779173614 Chr.2: 240596785-240596786 on Build GRCh38
rs755489917 Chr.2: 240596719-240596719 on Build GRCh38
rs762746164 Chr.2: 240596758-240596758 on Build GRCh38
rs775284291 Chr.2: 240596757-240596757 on Build GRCh38
rs769607366 Chr.2: 240596685-240596685 on Build GRCh38
rs772712964 Chr.2: 240596871-240596871 on Build GRCh38
rs776661578 Chr.2: 240596681-240596681 on Build GRCh38
rs945698867 Chr.2: 240596698-240596698 on Build GRCh38
rs201527381 Chr.2: 240596697-240596697 on Build GRCh38
rs753054948 Chr.2: 240596796-240596796 on Build GRCh38
rs377630733 Chr.2: 240596839-240596839 on Build GRCh38
rs7607759 Chr.2: 240596709-240596709 on Build GRCh38
rs779325115 Chr.2: 240596726-240596726 on Build GRCh38
rs934112829 Chr.2: 240596792-240596792 on Build GRCh38
rs370493316 Chr.2: 240596783-240596783 on Build GRCh38
rs746271457 Chr.2: 240596869-240596869 on Build GRCh38
rs1042843275 Chr.2: 240596703-240596703 on Build GRCh38
rs868447092 Chr.2: 240596684-240596684 on Build GRCh38
rs761495953 Chr.2: 240596779-240596779 on Build GRCh38
rs747049929 Chr.2: 240596868-240596868 on Build GRCh38
rs779874859 Chr.2: 240596847-240596847 on Build GRCh38
rs577695842 Chr.2: 240596870-240596870 on Build GRCh38
rs945747188 Chr.2: 240596859-240596859 on Build GRCh38
rs778761844 Chr.2: 240596748-240596748 on Build GRCh38
rs921733409 Chr.2: 240596822-240596822 on Build GRCh38

View Full Product Details