Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 2752437-2752668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf38
Entrez Gene ID
153571
Gene Name
chromosome 5 open reading frame 38
Gene Aliases
CEI, IRX2NB
UniGene
Hs.148486
Gene Chromosome Location
Chr.5: 2751940-2758576 on Build GRCh38
Transcripts
NM_001294337.1 NM_001306149.1 NM_001306150.1 NM_178569.3 NR_122125.1 NR_122126.1 XM_005248256.3 XM_005248257.3 XM_011513975.2 XM_011513977.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1067200 c.178C>T p.R60* Chr.5: 2752442-2752442 on Build GRCh38
1067199 c.173G>T p.W58L Chr.5: 2752437-2752437 on Build GRCh38
1067202 c.205G>A p.V69M Chr.5: 2752626-2752626 on Build GRCh38
6616267 c.236A>G p.Y79C Chr.5: 2752657-2752657 on Build GRCh38
1067201 c.179G>A p.R60Q Chr.5: 2752443-2752443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924407122 Chr.5: 2752557-2752557 on Build GRCh38
rs764868038 Chr.5: 2752579-2752579 on Build GRCh38
rs749462969 Chr.5: 2752503-2752503 on Build GRCh38
rs991096506 Chr.5: 2752530-2752530 on Build GRCh38
rs755508252 Chr.5: 2752489-2752489 on Build GRCh38
rs914518863 Chr.5: 2752534-2752534 on Build GRCh38
rs745751690 Chr.5: 2752455-2752455 on Build GRCh38
rs113410512 Chr.5: 2752508-2752508 on Build GRCh38
rs562225418 Chr.5: 2752610-2752610 on Build GRCh38
rs575679851 Chr.5: 2752502-2752502 on Build GRCh38
rs540965910 Chr.5: 2752571-2752571 on Build GRCh38
rs958402538 Chr.5: 2752519-2752519 on Build GRCh38
rs751878228 Chr.5: 2752488-2752488 on Build GRCh38
rs764093877 Chr.5: 2752605-2752605 on Build GRCh38
rs370175635 Chr.5: 2752624-2752624 on Build GRCh38
rs992654889 Chr.5: 2752568-2752568 on Build GRCh38
rs779489339 Chr.5: 2752580-2752581 on Build GRCh38
rs980867138 Chr.5: 2752580-2752580 on Build GRCh38
rs952929985 Chr.5: 2752472-2752472 on Build GRCh38
rs771411660 Chr.5: 2752581-2752585 on Build GRCh38
rs779051555 Chr.5: 2752625-2752625 on Build GRCh38
rs749863869 Chr.5: 2752511-2752512 on Build GRCh38
rs945600986 Chr.5: 2752639-2752639 on Build GRCh38
rs1054045814 Chr.5: 2752615-2752615 on Build GRCh38
rs775606012 Chr.5: 2752573-2752573 on Build GRCh38
rs767970450 Chr.5: 2752490-2752490 on Build GRCh38
rs371083805 Chr.5: 2752595-2752595 on Build GRCh38
rs1016152608 Chr.5: 2752505-2752505 on Build GRCh38
rs147641225 Chr.5: 2752456-2752456 on Build GRCh38
rs768229034 Chr.5: 2752574-2752574 on Build GRCh38
rs758608640 Chr.5: 2752498-2752498 on Build GRCh38
rs567456947 Chr.5: 2752467-2752467 on Build GRCh38
rs947282282 Chr.5: 2752547-2752547 on Build GRCh38
rs756227510 Chr.5: 2752501-2752501 on Build GRCh38
rs746245907 Chr.5: 2752641-2752642 on Build GRCh38
rs557505885 Chr.5: 2752487-2752487 on Build GRCh38
rs188657405 Chr.5: 2752507-2752507 on Build GRCh38
rs376512087 Chr.5: 2752470-2752470 on Build GRCh38
rs553417965 Chr.5: 2752584-2752584 on Build GRCh38
rs752754017 Chr.5: 2752492-2752492 on Build GRCh38
rs926631970 Chr.5: 2752590-2752591 on Build GRCh38
rs574194556 Chr.5: 2752586-2752586 on Build GRCh38
rs756636370 Chr.5: 2752601-2752601 on Build GRCh38
rs573447040 Chr.5: 2752516-2752516 on Build GRCh38
rs1007305257 Chr.5: 2752482-2752482 on Build GRCh38
rs943878983 Chr.5: 2752612-2752612 on Build GRCh38
rs1041567068 Chr.5: 2752633-2752633 on Build GRCh38
rs772286114 Chr.5: 2752642-2752642 on Build GRCh38
rs555643866 Chr.5: 2752575-2752575 on Build GRCh38
rs775125061 Chr.5: 2752460-2752460 on Build GRCh38
rs543877982 Chr.5: 2752590-2752590 on Build GRCh38
rs1041555083 Chr.5: 2752549-2752549 on Build GRCh38
rs760307430 Chr.5: 2752466-2752466 on Build GRCh38

View Full Product Details