Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33676587-33676826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025963 c.1254C>T p.G418G Chr.20: 33676792-33676792 on Build GRCh38
3911137 c.1303C>T p.L435L Chr.20: 33676743-33676743 on Build GRCh38
2757806 c.1260C>T p.G420G Chr.20: 33676786-33676786 on Build GRCh38
1025964 c.1239C>T p.F413F Chr.20: 33676807-33676807 on Build GRCh38
1615482 c.1221G>A p.E407E Chr.20: 33676825-33676825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774912473 Chr.20: 33676794-33676794 on Build GRCh38
rs775233744 Chr.20: 33676692-33676692 on Build GRCh38
rs377471842 Chr.20: 33676719-33676719 on Build GRCh38
rs975204102 Chr.20: 33676619-33676619 on Build GRCh38
rs777420006 Chr.20: 33676770-33676770 on Build GRCh38
rs151195835 Chr.20: 33676748-33676748 on Build GRCh38
rs532185356 Chr.20: 33676656-33676656 on Build GRCh38
rs897107396 Chr.20: 33676759-33676759 on Build GRCh38
rs772350151 Chr.20: 33676803-33676803 on Build GRCh38
rs931137356 Chr.20: 33676638-33676638 on Build GRCh38
rs368965201 Chr.20: 33676744-33676744 on Build GRCh38
rs766184341 Chr.20: 33676732-33676732 on Build GRCh38
rs749840514 Chr.20: 33676787-33676787 on Build GRCh38
rs750410826 Chr.20: 33676726-33676726 on Build GRCh38
rs758107340 Chr.20: 33676760-33676760 on Build GRCh38
rs144921013 Chr.20: 33676721-33676721 on Build GRCh38
rs763256677 Chr.20: 33676718-33676718 on Build GRCh38
rs763187815 Chr.20: 33676716-33676716 on Build GRCh38
rs564748020 Chr.20: 33676640-33676640 on Build GRCh38
rs964030268 Chr.20: 33676613-33676613 on Build GRCh38
rs746377163 Chr.20: 33676690-33676690 on Build GRCh38
rs932094234 Chr.20: 33676667-33676668 on Build GRCh38
rs140266022 Chr.20: 33676774-33676774 on Build GRCh38
rs746393982 Chr.20: 33676667-33676667 on Build GRCh38
rs769138235 Chr.20: 33676792-33676792 on Build GRCh38
rs938654254 Chr.20: 33676679-33676679 on Build GRCh38
rs908369916 Chr.20: 33676657-33676657 on Build GRCh38
rs1040392274 Chr.20: 33676643-33676643 on Build GRCh38
rs3213178 Chr.20: 33676723-33676723 on Build GRCh38
rs147013405 Chr.20: 33676801-33676801 on Build GRCh38
rs935327063 Chr.20: 33676610-33676610 on Build GRCh38
rs770507779 Chr.20: 33676771-33676771 on Build GRCh38
rs139231439 Chr.20: 33676762-33676762 on Build GRCh38
rs3213177 Chr.20: 33676727-33676727 on Build GRCh38
rs267605891 Chr.20: 33676745-33676745 on Build GRCh38
rs767689297 Chr.20: 33676724-33676724 on Build GRCh38
rs919652683 Chr.20: 33676633-33676633 on Build GRCh38
rs772778710 Chr.20: 33676688-33676688 on Build GRCh38
rs535111025 Chr.20: 33676807-33676807 on Build GRCh38
rs566253357 Chr.20: 33676669-33676669 on Build GRCh38
rs547184326 Chr.20: 33676667-33676667 on Build GRCh38
rs536894303 Chr.20: 33676703-33676703 on Build GRCh38
rs752347181 Chr.20: 33676750-33676750 on Build GRCh38

View Full Product Details