Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100522602-100522837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD46
Entrez Gene ID
157567
Gene Name
ankyrin repeat domain 46
Gene Aliases
ANK-S, GENX-115279
UniGene
Hs.530199
Gene Chromosome Location
Chr.8: 100509752-100559786 on Build GRCh38
Transcripts
NM_001270377.1 NM_001270378.1 NM_001270379.1 NM_198401.3 XM_006716509.1 XM_017013106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
748087 c.637G>A p.V213M Chr.8: 100522605-100522605 on Build GRCh38
4860027 c.636+1G>A p.? Chr.8: 100522605-100522605 on Build GRCh38
3269157 c.526C>T p.L176F Chr.8: 100522716-100522716 on Build GRCh38
485877 c.605T>C p.I202T Chr.8: 100522637-100522637 on Build GRCh38
6581626 c.496A>C p.N166H Chr.8: 100522746-100522746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750961822 Chr.8: 100522803-100522803 on Build GRCh38
rs770864256 Chr.8: 100522712-100522712 on Build GRCh38
rs372353120 Chr.8: 100522732-100522732 on Build GRCh38
rs187920463 Chr.8: 100522681-100522681 on Build GRCh38
rs763761865 Chr.8: 100522796-100522796 on Build GRCh38
rs774739166 Chr.8: 100522722-100522724 on Build GRCh38
rs149022557 Chr.8: 100522686-100522686 on Build GRCh38
rs577749731 Chr.8: 100522703-100522703 on Build GRCh38
rs762285453 Chr.8: 100522746-100522747 on Build GRCh38
rs751985899 Chr.8: 100522642-100522642 on Build GRCh38
rs16898492 Chr.8: 100522807-100522807 on Build GRCh38
rs745791971 Chr.8: 100522739-100522739 on Build GRCh38
rs937747474 Chr.8: 100522759-100522759 on Build GRCh38
rs982092867 Chr.8: 100522745-100522745 on Build GRCh38
rs192644546 Chr.8: 100522756-100522756 on Build GRCh38
rs368028062 Chr.8: 100522779-100522779 on Build GRCh38
rs140128819 Chr.8: 100522802-100522803 on Build GRCh38
rs759003595 Chr.8: 100522636-100522636 on Build GRCh38
rs771921223 Chr.8: 100522743-100522743 on Build GRCh38
rs143395176 Chr.8: 100522758-100522758 on Build GRCh38
rs780640944 Chr.8: 100522641-100522641 on Build GRCh38
rs928568088 Chr.8: 100522752-100522752 on Build GRCh38
rs760295512 Chr.8: 100522789-100522789 on Build GRCh38
rs776008084 Chr.8: 100522797-100522797 on Build GRCh38
rs781612329 Chr.8: 100522651-100522651 on Build GRCh38
rs755447767 Chr.8: 100522647-100522647 on Build GRCh38
rs761348378 Chr.8: 100522798-100522798 on Build GRCh38
rs369691090 Chr.8: 100522737-100522737 on Build GRCh38
rs769205440 Chr.8: 100522696-100522696 on Build GRCh38
rs748356876 Chr.8: 100522672-100522672 on Build GRCh38

View Full Product Details