Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203703599-203703872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B4
Entrez Gene ID
493
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Gene Aliases
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
UniGene
Hs.343522
Gene Chromosome Location
Chr.1: 203626787-203744081 on Build GRCh38
Transcripts
NM_001001396.2 NM_001684.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3481888 c.991G>A p.G331R Chr.1: 203703705-203703705 on Build GRCh38
458306 c.951C>T p.D317D Chr.1: 203703665-203703665 on Build GRCh38
73777 c.1005G>C p.E335D Chr.1: 203703719-203703719 on Build GRCh38
1180137 c.999C>A p.D333E Chr.1: 203703713-203703713 on Build GRCh38
3664969 c.1062C>A p.G354G Chr.1: 203703776-203703776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554381106 Chr.1: 203703712-203703712 on Build GRCh38
rs182951975 Chr.1: 203703623-203703623 on Build GRCh38
rs202197624 Chr.1: 203703672-203703672 on Build GRCh38
rs200430665 Chr.1: 203703711-203703711 on Build GRCh38
rs771678625 Chr.1: 203703773-203703773 on Build GRCh38
rs576322841 Chr.1: 203703710-203703710 on Build GRCh38
rs193920934 Chr.1: 203703713-203703713 on Build GRCh38
rs768398809 Chr.1: 203703648-203703648 on Build GRCh38
rs771834143 Chr.1: 203703669-203703669 on Build GRCh38
rs757437651 Chr.1: 203703697-203703697 on Build GRCh38
rs775527046 Chr.1: 203703675-203703675 on Build GRCh38
rs748845883 Chr.1: 203703640-203703640 on Build GRCh38
rs145882632 Chr.1: 203703700-203703700 on Build GRCh38
rs199889328 Chr.1: 203703682-203703682 on Build GRCh38
rs145839216 Chr.1: 203703665-203703665 on Build GRCh38
rs754416213 Chr.1: 203703714-203703714 on Build GRCh38
rs762973524 Chr.1: 203703676-203703676 on Build GRCh38
rs369438674 Chr.1: 203703695-203703695 on Build GRCh38
rs537893117 Chr.1: 203703619-203703619 on Build GRCh38
rs776512538 Chr.1: 203703829-203703829 on Build GRCh38
rs756641165 Chr.1: 203703626-203703626 on Build GRCh38
rs759476576 Chr.1: 203703833-203703833 on Build GRCh38
rs201495785 Chr.1: 203703806-203703806 on Build GRCh38
rs750816064 Chr.1: 203703702-203703702 on Build GRCh38
rs770737574 Chr.1: 203703820-203703820 on Build GRCh38
rs747778799 Chr.1: 203703755-203703755 on Build GRCh38
rs746332372 Chr.1: 203703624-203703624 on Build GRCh38
rs79138871 Chr.1: 203703678-203703678 on Build GRCh38
rs778568395 Chr.1: 203703743-203703743 on Build GRCh38
rs187671253 Chr.1: 203703628-203703628 on Build GRCh38
rs756614300 Chr.1: 203703681-203703682 on Build GRCh38
rs768666859 Chr.1: 203703834-203703834 on Build GRCh38
rs747649058 Chr.1: 203703787-203703787 on Build GRCh38
rs753113482 Chr.1: 203703678-203703679 on Build GRCh38
rs772895722 Chr.1: 203703671-203703671 on Build GRCh38
rs145963279 Chr.1: 203703739-203703739 on Build GRCh38

View Full Product Details