Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118555167-118555400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT46
Entrez Gene ID
56912
Gene Name
intraflagellar transport 46
Gene Aliases
C11orf2, C11orf60, CFAP32
UniGene
Hs.533738
Gene Chromosome Location
Chr.11: 118544528-118575624 on Build GRCh38
Transcripts
NM_001168618.1 NM_020153.3 XM_011542905.2 XM_011542906.2 XM_017018017.1 XM_017018018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436009 c.370C>T p.P124S Chr.11: 118555291-118555291 on Build GRCh38
6268358 c.339-3C>A p.? Chr.11: 118555325-118555325 on Build GRCh38
4835477 c.377C>T p.S126F Chr.11: 118555284-118555284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781961185 Chr.11: 118555281-118555281 on Build GRCh38
rs782220462 Chr.11: 118555348-118555348 on Build GRCh38
rs760516828 Chr.11: 118555372-118555372 on Build GRCh38
rs782323249 Chr.11: 118555286-118555286 on Build GRCh38
rs77812853 Chr.11: 118555347-118555347 on Build GRCh38
rs782604953 Chr.11: 118555221-118555221 on Build GRCh38
rs781917504 Chr.11: 118555195-118555195 on Build GRCh38
rs782212290 Chr.11: 118555284-118555284 on Build GRCh38
rs782457822 Chr.11: 118555228-118555228 on Build GRCh38
rs944278383 Chr.11: 118555248-118555248 on Build GRCh38
rs782494445 Chr.11: 118555207-118555207 on Build GRCh38
rs781857636 Chr.11: 118555205-118555205 on Build GRCh38
rs904286287 Chr.11: 118555341-118555341 on Build GRCh38
rs781880943 Chr.11: 118555345-118555345 on Build GRCh38
rs782782834 Chr.11: 118555202-118555202 on Build GRCh38
rs573166702 Chr.11: 118555378-118555378 on Build GRCh38
rs143192297 Chr.11: 118555317-118555317 on Build GRCh38
rs376221796 Chr.11: 118555360-118555360 on Build GRCh38
rs782750393 Chr.11: 118555326-118555326 on Build GRCh38
rs557195802 Chr.11: 118555287-118555287 on Build GRCh38
rs781828935 Chr.11: 118555327-118555327 on Build GRCh38
rs1043102929 Chr.11: 118555311-118555311 on Build GRCh38
rs782228695 Chr.11: 118555211-118555212 on Build GRCh38
rs782199189 Chr.11: 118555225-118555225 on Build GRCh38
rs139391241 Chr.11: 118555251-118555251 on Build GRCh38
rs1041354477 Chr.11: 118555271-118555271 on Build GRCh38
rs908669819 Chr.11: 118555223-118555223 on Build GRCh38
rs782707333 Chr.11: 118555342-118555342 on Build GRCh38
rs375847066 Chr.11: 118555290-118555290 on Build GRCh38
rs782464871 Chr.11: 118555353-118555353 on Build GRCh38
rs554934951 Chr.11: 118555366-118555366 on Build GRCh38
rs782652017 Chr.11: 118555233-118555233 on Build GRCh38
rs782584112 Chr.11: 118555356-118555356 on Build GRCh38
rs782463624 Chr.11: 118555335-118555335 on Build GRCh38
rs372794442 Chr.11: 118555255-118555255 on Build GRCh38
rs782808904 Chr.11: 118555306-118555306 on Build GRCh38

View Full Product Details