Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58103922-58104189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNB
Entrez Gene ID
2317
Gene Name
filamin B
Gene Aliases
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP
UniGene
Hs.476448
Gene Chromosome Location
Chr.3: 58008400-58172255 on Build GRCh38
Transcripts
NM_001164317.1 NM_001164318.1 NM_001164319.1 NM_001457.3 XM_005264978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1593980 c.1538T>C p.F513S Chr.3: 58104013-58104013 on Build GRCh38
6393515 c.1597C>A p.H533N Chr.3: 58104072-58104072 on Build GRCh38
1149777 c.1492G>T p.E498* Chr.3: 58103967-58103967 on Build GRCh38
1593981 c.1513G>T p.D505Y Chr.3: 58103988-58103988 on Build GRCh38
3945640 c.1610+6G>T p.? Chr.3: 58104091-58104091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771808375 Chr.3: 58103991-58103991 on Build GRCh38
rs756540906 Chr.3: 58104125-58104125 on Build GRCh38
rs912934979 Chr.3: 58104104-58104104 on Build GRCh38
rs764967982 Chr.3: 58104014-58104014 on Build GRCh38
rs544138399 Chr.3: 58103997-58103997 on Build GRCh38
rs892239105 Chr.3: 58103982-58103982 on Build GRCh38
rs761624604 Chr.3: 58104070-58104070 on Build GRCh38
rs1005619387 Chr.3: 58104163-58104163 on Build GRCh38
rs763168530 Chr.3: 58104017-58104017 on Build GRCh38
rs766932995 Chr.3: 58104122-58104122 on Build GRCh38
rs987863606 Chr.3: 58104161-58104161 on Build GRCh38
rs758401425 Chr.3: 58104061-58104061 on Build GRCh38
rs750869078 Chr.3: 58104046-58104046 on Build GRCh38
rs757277059 Chr.3: 58104035-58104035 on Build GRCh38
rs762719838 Chr.3: 58104093-58104093 on Build GRCh38
rs201544295 Chr.3: 58104009-58104009 on Build GRCh38
rs761998967 Chr.3: 58104110-58104110 on Build GRCh38
rs745828853 Chr.3: 58104090-58104090 on Build GRCh38
rs764245609 Chr.3: 58104033-58104033 on Build GRCh38
rs751194469 Chr.3: 58104136-58104136 on Build GRCh38
rs964625043 Chr.3: 58104046-58104050 on Build GRCh38
rs145086495 Chr.3: 58104061-58104061 on Build GRCh38
rs767516205 Chr.3: 58104118-58104118 on Build GRCh38
rs143611997 Chr.3: 58104160-58104160 on Build GRCh38
rs773943113 Chr.3: 58104063-58104063 on Build GRCh38
rs528968388 Chr.3: 58104138-58104138 on Build GRCh38
rs772851411 Chr.3: 58103994-58103994 on Build GRCh38
rs376751278 Chr.3: 58103955-58103955 on Build GRCh38
rs147001986 Chr.3: 58104015-58104015 on Build GRCh38
rs770969895 Chr.3: 58104000-58104000 on Build GRCh38
rs1029216750 Chr.3: 58104044-58104044 on Build GRCh38
rs768105116 Chr.3: 58104043-58104043 on Build GRCh38
rs746105983 Chr.3: 58104060-58104061 on Build GRCh38
rs750474826 Chr.3: 58104120-58104120 on Build GRCh38
rs201241488 Chr.3: 58103960-58103960 on Build GRCh38
rs371251819 Chr.3: 58104008-58104008 on Build GRCh38
rs994704249 Chr.3: 58104147-58104147 on Build GRCh38
rs1522385 Chr.3: 58104124-58104124 on Build GRCh38
rs369252572 Chr.3: 58104128-58104128 on Build GRCh38
rs761089849 Chr.3: 58104121-58104121 on Build GRCh38
rs780705311 Chr.3: 58104083-58104083 on Build GRCh38
rs962189237 Chr.3: 58104074-58104074 on Build GRCh38
rs780235410 Chr.3: 58104134-58104134 on Build GRCh38
rs770325452 Chr.3: 58104065-58104065 on Build GRCh38
rs774520887 Chr.3: 58104100-58104100 on Build GRCh38
rs1522386 Chr.3: 58104150-58104150 on Build GRCh38
rs756594330 Chr.3: 58104056-58104056 on Build GRCh38
rs755365697 Chr.3: 58104127-58104127 on Build GRCh38
rs756897733 Chr.3: 58104137-58104137 on Build GRCh38
rs578032990 Chr.3: 58104016-58104016 on Build GRCh38
rs768930462 Chr.3: 58104097-58104097 on Build GRCh38
rs138220431 Chr.3: 58104034-58104034 on Build GRCh38
rs747825411 Chr.3: 58103961-58103961 on Build GRCh38
rs769707810 Chr.3: 58104091-58104091 on Build GRCh38

View Full Product Details