Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7626204-7626437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3074068 c.692T>A p.I231N Chr.5: 7626288-7626288 on Build GRCh38
3856364 c.651G>A p.Q217Q Chr.5: 7626247-7626247 on Build GRCh38
5351219 c.689G>T p.R230L Chr.5: 7626285-7626285 on Build GRCh38
3856365 c.687G>A p.S229S Chr.5: 7626283-7626283 on Build GRCh38
1567597 c.712C>T p.R238C Chr.5: 7626308-7626308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754055904 Chr.5: 7626308-7626308 on Build GRCh38
rs16878848 Chr.5: 7626395-7626395 on Build GRCh38
rs909098249 Chr.5: 7626275-7626275 on Build GRCh38
rs976062067 Chr.5: 7626226-7626226 on Build GRCh38
rs144846733 Chr.5: 7626309-7626309 on Build GRCh38
rs768172555 Chr.5: 7626366-7626366 on Build GRCh38
rs776045266 Chr.5: 7626368-7626368 on Build GRCh38
rs779908161 Chr.5: 7626359-7626359 on Build GRCh38
rs200404909 Chr.5: 7626286-7626286 on Build GRCh38
rs6865872 Chr.5: 7626289-7626289 on Build GRCh38
rs760109082 Chr.5: 7626259-7626259 on Build GRCh38
rs751401259 Chr.5: 7626344-7626344 on Build GRCh38
rs554318638 Chr.5: 7626277-7626277 on Build GRCh38
rs745728249 Chr.5: 7626331-7626331 on Build GRCh38
rs761404738 Chr.5: 7626233-7626233 on Build GRCh38
rs759387418 Chr.5: 7626356-7626358 on Build GRCh38
rs766735350 Chr.5: 7626235-7626235 on Build GRCh38
rs774906597 Chr.5: 7626238-7626238 on Build GRCh38
rs373621272 Chr.5: 7626229-7626229 on Build GRCh38
rs749648187 Chr.5: 7626258-7626258 on Build GRCh38
rs758193048 Chr.5: 7626357-7626357 on Build GRCh38
rs938422321 Chr.5: 7626390-7626390 on Build GRCh38
rs950769591 Chr.5: 7626228-7626228 on Build GRCh38
rs778663075 Chr.5: 7626324-7626324 on Build GRCh38
rs377677457 Chr.5: 7626276-7626276 on Build GRCh38
rs776298072 Chr.5: 7626223-7626223 on Build GRCh38
rs887540221 Chr.5: 7626399-7626399 on Build GRCh38
rs149422716 Chr.5: 7626283-7626283 on Build GRCh38
rs746942457 Chr.5: 7626362-7626362 on Build GRCh38

View Full Product Details