Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155239571-155239824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
414006 c.415G>T p.A139S Chr.1: 155239655-155239655 on Build GRCh38
5531620 c.369C>T p.A123A Chr.1: 155239701-155239701 on Build GRCh38
4022967 c.434A>G p.K145R Chr.1: 155239636-155239636 on Build GRCh38
5531619 c.368C>T p.A123V Chr.1: 155239702-155239702 on Build GRCh38
4022966 c.440A>G p.Y147C Chr.1: 155239630-155239630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878853314 Chr.1: 155239655-155239655 on Build GRCh38
rs121908312 Chr.1: 155239716-155239716 on Build GRCh38
rs368145008 Chr.1: 155239748-155239748 on Build GRCh38
rs756540913 Chr.1: 155239767-155239767 on Build GRCh38
rs749867889 Chr.1: 155239775-155239775 on Build GRCh38
rs79175920 Chr.1: 155239683-155239683 on Build GRCh38
rs758455177 Chr.1: 155239700-155239700 on Build GRCh38
rs878853316 Chr.1: 155239663-155239663 on Build GRCh38
rs753816998 Chr.1: 155239699-155239699 on Build GRCh38
rs767272691 Chr.1: 155239658-155239658 on Build GRCh38
rs794727708 Chr.1: 155239639-155239639 on Build GRCh38
rs758447515 Chr.1: 155239633-155239633 on Build GRCh38
rs916865472 Chr.1: 155239735-155239735 on Build GRCh38
rs372156961 Chr.1: 155239791-155239791 on Build GRCh38
rs781563451 Chr.1: 155239758-155239758 on Build GRCh38
rs770614424 Chr.1: 155239643-155239643 on Build GRCh38
rs752444566 Chr.1: 155239680-155239680 on Build GRCh38
rs767999479 Chr.1: 155239789-155239790 on Build GRCh38
rs377315750 Chr.1: 155239803-155239803 on Build GRCh38
rs779619231 Chr.1: 155239740-155239740 on Build GRCh38
rs770218957 Chr.1: 155239616-155239616 on Build GRCh38
rs759174705 Chr.1: 155239657-155239657 on Build GRCh38
rs770561256 Chr.1: 155239788-155239788 on Build GRCh38
rs1033293531 Chr.1: 155239609-155239609 on Build GRCh38
rs748485792 Chr.1: 155239762-155239762 on Build GRCh38
rs780169533 Chr.1: 155239746-155239746 on Build GRCh38
rs746684178 Chr.1: 155239778-155239778 on Build GRCh38
rs867024654 Chr.1: 155239709-155239709 on Build GRCh38
rs111857613 Chr.1: 155239773-155239773 on Build GRCh38
rs539166948 Chr.1: 155239615-155239615 on Build GRCh38
rs868211463 Chr.1: 155239705-155239705 on Build GRCh38
rs374003673 Chr.1: 155239648-155239648 on Build GRCh38
rs755161012 Chr.1: 155239752-155239752 on Build GRCh38
rs763972468 Chr.1: 155239687-155239687 on Build GRCh38
rs772806479 Chr.1: 155239623-155239623 on Build GRCh38

View Full Product Details