Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33676757-33676986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
UniGene
Hs.654393
Gene Chromosome Location
Chr.20: 33675486-33686404 on Build GRCh38
Transcripts
NM_005225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651005 c.1145C>T p.S382L Chr.20: 33676901-33676901 on Build GRCh38
4458312 c.1080C>T p.S360S Chr.20: 33676966-33676966 on Build GRCh38
2757806 c.1260C>T p.G420G Chr.20: 33676786-33676786 on Build GRCh38
4460037 c.1150C>T p.L384L Chr.20: 33676896-33676896 on Build GRCh38
4983088 c.1265G>C p.R422T Chr.20: 33676781-33676781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956866221 Chr.20: 33676826-33676826 on Build GRCh38
rs774912473 Chr.20: 33676794-33676794 on Build GRCh38
rs373587139 Chr.20: 33676935-33676935 on Build GRCh38
rs555876218 Chr.20: 33676906-33676906 on Build GRCh38
rs769226220 Chr.20: 33676883-33676883 on Build GRCh38
rs773280034 Chr.20: 33676901-33676901 on Build GRCh38
rs756700620 Chr.20: 33676861-33676861 on Build GRCh38
rs747864159 Chr.20: 33676870-33676872 on Build GRCh38
rs780453071 Chr.20: 33676860-33676862 on Build GRCh38
rs756991861 Chr.20: 33676939-33676939 on Build GRCh38
rs770915873 Chr.20: 33676907-33676907 on Build GRCh38
rs371082004 Chr.20: 33676825-33676825 on Build GRCh38
rs373228968 Chr.20: 33676830-33676830 on Build GRCh38
rs762689168 Chr.20: 33676933-33676933 on Build GRCh38
rs755713339 Chr.20: 33676949-33676949 on Build GRCh38
rs772350151 Chr.20: 33676803-33676803 on Build GRCh38
rs779509637 Chr.20: 33676950-33676950 on Build GRCh38
rs3213176 Chr.20: 33676869-33676869 on Build GRCh38
rs749840514 Chr.20: 33676787-33676787 on Build GRCh38
rs766526488 Chr.20: 33676819-33676819 on Build GRCh38
rs779566653 Chr.20: 33676884-33676884 on Build GRCh38
rs201607111 Chr.20: 33676909-33676909 on Build GRCh38
rs553240434 Chr.20: 33676828-33676828 on Build GRCh38
rs923288606 Chr.20: 33676888-33676888 on Build GRCh38
rs769138235 Chr.20: 33676792-33676792 on Build GRCh38
rs748480309 Chr.20: 33676891-33676891 on Build GRCh38
rs915788985 Chr.20: 33676929-33676929 on Build GRCh38
rs751141581 Chr.20: 33676936-33676936 on Build GRCh38
rs751247203 Chr.20: 33676854-33676854 on Build GRCh38
rs367723468 Chr.20: 33676941-33676941 on Build GRCh38
rs772585440 Chr.20: 33676900-33676900 on Build GRCh38
rs147013405 Chr.20: 33676801-33676801 on Build GRCh38
rs752664219 Chr.20: 33676918-33676918 on Build GRCh38
rs1012010770 Chr.20: 33676810-33676810 on Build GRCh38
rs371155232 Chr.20: 33676914-33676914 on Build GRCh38
rs758656204 Chr.20: 33676957-33676957 on Build GRCh38
rs535111025 Chr.20: 33676807-33676807 on Build GRCh38
rs748711291 Chr.20: 33676951-33676951 on Build GRCh38
rs778271263 Chr.20: 33676964-33676964 on Build GRCh38
rs747187347 Chr.20: 33676965-33676965 on Build GRCh38
rs560760846 Chr.20: 33676881-33676881 on Build GRCh38
rs760692854 Chr.20: 33676871-33676871 on Build GRCh38
rs760618502 Chr.20: 33676818-33676818 on Build GRCh38
rs749886199 Chr.20: 33676942-33676942 on Build GRCh38
rs142009843 Chr.20: 33676870-33676870 on Build GRCh38
rs796465642 Chr.20: 33676928-33676928 on Build GRCh38

View Full Product Details