Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197774042-197774313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYTTD1
Entrez Gene ID
84248
Gene Name
forty-two-three domain containing 1
Gene Aliases
UIF
UniGene
Hs.277533
Gene Chromosome Location
Chr.3: 197749553-197784446 on Build GRCh38
Transcripts
NM_001011537.2 NM_032288.6 NR_027840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3992992 c.651T>C p.T217T Chr.3: 197774205-197774205 on Build GRCh38
4116534 c.619C>G p.Q207E Chr.3: 197774173-197774173 on Build GRCh38
2948006 c.631G>A p.D211N Chr.3: 197774185-197774185 on Build GRCh38
1043195 c.647G>T p.R216I Chr.3: 197774201-197774201 on Build GRCh38
4439962 c.650C>A p.T217N Chr.3: 197774204-197774204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769838870 Chr.3: 197774205-197774205 on Build GRCh38
rs760034942 Chr.3: 197774169-197774169 on Build GRCh38
rs145575875 Chr.3: 197774243-197774243 on Build GRCh38
rs762275104 Chr.3: 197774130-197774130 on Build GRCh38
rs1001734368 Chr.3: 197774217-197774217 on Build GRCh38
rs867200339 Chr.3: 197774206-197774206 on Build GRCh38
rs369631633 Chr.3: 197774207-197774207 on Build GRCh38
rs569685873 Chr.3: 197774283-197774283 on Build GRCh38
rs750466107 Chr.3: 197774147-197774147 on Build GRCh38
rs762032490 Chr.3: 197774131-197774132 on Build GRCh38
rs765794105 Chr.3: 197774140-197774140 on Build GRCh38
rs933017869 Chr.3: 197774138-197774138 on Build GRCh38
rs754231043 Chr.3: 197774112-197774112 on Build GRCh38
rs768040827 Chr.3: 197774193-197774193 on Build GRCh38
rs138534165 Chr.3: 197774151-197774151 on Build GRCh38
rs368676118 Chr.3: 197774254-197774254 on Build GRCh38
rs765598012 Chr.3: 197774216-197774216 on Build GRCh38
rs776955759 Chr.3: 197774261-197774261 on Build GRCh38
rs559222677 Chr.3: 197774246-197774246 on Build GRCh38
rs933361363 Chr.3: 197774108-197774108 on Build GRCh38
rs749182919 Chr.3: 197774235-197774235 on Build GRCh38
rs906090975 Chr.3: 197774187-197774187 on Build GRCh38
rs781106842 Chr.3: 197774184-197774184 on Build GRCh38
rs901836519 Chr.3: 197774101-197774101 on Build GRCh38
rs374937231 Chr.3: 197774088-197774088 on Build GRCh38
rs775238841 Chr.3: 197774132-197774132 on Build GRCh38
rs770411979 Chr.3: 197774242-197774242 on Build GRCh38
rs536782175 Chr.3: 197774153-197774153 on Build GRCh38
rs748007922 Chr.3: 197774185-197774185 on Build GRCh38
rs554922150 Chr.3: 197774172-197774172 on Build GRCh38
rs376869371 Chr.3: 197774152-197774152 on Build GRCh38

View Full Product Details