Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30723178-30723431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
UniGene
Hs.234355
Gene Chromosome Location
Chr.12: 30709552-30754994 on Build GRCh38
Transcripts
NM_001002259.2 NM_001206856.2 NM_001319842.1 NM_001319843.1 NM_001319844.1 NM_001319845.1 NM_001319846.1 NM_023925.4 NM_032156.4 XM_006719138.3 XM_006719139.3 XM_006719140.3 XM_006719142.3 XM_006719144.3 XM_006719145.3 XM_006719146.3 XM_006719147.3 XM_017019851.1 XM_017019852.1 XM_017019853.1 XM_017019854.1 XM_017019855.1 XM_017019856.1 XM_017019857.1 XM_017019858.1 XM_017019859.1 XM_017019860.1 XM_017019861.1 XM_017019862.1 XM_017019863.1 XM_017019864.1 XM_017019865.1 XM_017019866.1 XM_017019867.1 XM_017019868.1 XM_017019869.1 XM_017019870.1 XM_017019871.1 XM_017019872.1 XM_017019873.1 XM_017019874.1 XM_017019875.1 XM_017019876.1 XM_017019877.1 XM_017019878.1 XM_017019879.1 XM_017019880.1 XM_017019881.1 XM_017019882.1 XM_017019883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1361184 c.2043+2T>A p.? Chr.12: 30723257-30723257 on Build GRCh38
299105 c.1996G>T p.E666* Chr.12: 30723306-30723306 on Build GRCh38
3871468 c.2009C>T p.S670F Chr.12: 30723293-30723293 on Build GRCh38
390053 c.2012G>C p.S671T Chr.12: 30723290-30723290 on Build GRCh38
5827178 c.2043+2T>A p.? Chr.12: 30723257-30723257 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375191805 Chr.12: 30723330-30723330 on Build GRCh38
rs373700141 Chr.12: 30723212-30723212 on Build GRCh38
rs760208096 Chr.12: 30723357-30723357 on Build GRCh38
rs749993923 Chr.12: 30723251-30723251 on Build GRCh38
rs779158421 Chr.12: 30723345-30723346 on Build GRCh38
rs767405989 Chr.12: 30723346-30723346 on Build GRCh38
rs76973125 Chr.12: 30723320-30723320 on Build GRCh38
rs747131353 Chr.12: 30723303-30723303 on Build GRCh38
rs754516169 Chr.12: 30723365-30723365 on Build GRCh38
rs761657640 Chr.12: 30723343-30723343 on Build GRCh38
rs111738878 Chr.12: 30723254-30723254 on Build GRCh38
rs757076548 Chr.12: 30723249-30723249 on Build GRCh38
rs574079766 Chr.12: 30723275-30723275 on Build GRCh38
rs779720040 Chr.12: 30723260-30723260 on Build GRCh38
rs750327533 Chr.12: 30723351-30723351 on Build GRCh38
rs753447503 Chr.12: 30723261-30723261 on Build GRCh38
rs776964840 Chr.12: 30723219-30723219 on Build GRCh38
rs755390955 Chr.12: 30723264-30723264 on Build GRCh38
rs796988450 Chr.12: 30723342-30723342 on Build GRCh38
rs771097053 Chr.12: 30723313-30723313 on Build GRCh38
rs982719871 Chr.12: 30723324-30723324 on Build GRCh38
rs748689164 Chr.12: 30723269-30723269 on Build GRCh38
rs534412750 Chr.12: 30723220-30723220 on Build GRCh38
rs545757771 Chr.12: 30723374-30723374 on Build GRCh38
rs751533296 Chr.12: 30723237-30723237 on Build GRCh38
rs139865209 Chr.12: 30723364-30723365 on Build GRCh38
rs74878752 Chr.12: 30723273-30723273 on Build GRCh38
rs368154769 Chr.12: 30723336-30723336 on Build GRCh38
rs767487061 Chr.12: 30723250-30723250 on Build GRCh38
rs746604246 Chr.12: 30723346-30723347 on Build GRCh38
rs983386955 Chr.12: 30723367-30723367 on Build GRCh38
rs182388119 Chr.12: 30723364-30723364 on Build GRCh38
rs778408732 Chr.12: 30723286-30723286 on Build GRCh38
rs765825205 Chr.12: 30723358-30723358 on Build GRCh38
rs763751400 Chr.12: 30723223-30723223 on Build GRCh38
rs3835092 Chr.12: 30723365-30723366 on Build GRCh38
rs751573910 Chr.12: 30723265-30723265 on Build GRCh38
rs201569358 Chr.12: 30723319-30723319 on Build GRCh38
rs79465544 Chr.12: 30723288-30723288 on Build GRCh38
rs759778253 Chr.12: 30723321-30723321 on Build GRCh38

View Full Product Details