Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76439679-76439912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRRB
Entrez Gene ID
2103
Gene Name
estrogen related receptor beta
Gene Aliases
DFNB35, ERR2, ERRb, ESRL2, NR3B2
UniGene
Hs.435845
Gene Chromosome Location
Chr.14: 76371347-76501839 on Build GRCh38
Transcripts
NM_004452.3 XM_011536547.2 XM_011536550.2 XM_011536553.2 XM_011536554.2 XM_017021085.1 XM_017021086.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4772655 c.336C>A p.G112G Chr.14: 76439689-76439689 on Build GRCh38
5634891 c.340C>A p.H114N Chr.14: 76439693-76439693 on Build GRCh38
122901 c.352G>A p.A118T Chr.14: 76439705-76439705 on Build GRCh38
7265214 c.346G>A p.G116S Chr.14: 76439699-76439699 on Build GRCh38
6226493 c.345C>T p.Y115Y Chr.14: 76439698-76439698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907963697 Chr.14: 76439809-76439809 on Build GRCh38
rs779422934 Chr.14: 76439799-76439799 on Build GRCh38
rs184363159 Chr.14: 76439826-76439826 on Build GRCh38
rs755439000 Chr.14: 76439769-76439769 on Build GRCh38
rs567671346 Chr.14: 76439698-76439698 on Build GRCh38
rs727503040 Chr.14: 76439754-76439754 on Build GRCh38
rs760684578 Chr.14: 76439737-76439737 on Build GRCh38
rs752984128 Chr.14: 76439776-76439776 on Build GRCh38
rs975224218 Chr.14: 76439881-76439881 on Build GRCh38
rs61413504 Chr.14: 76439815-76439815 on Build GRCh38
rs557867243 Chr.14: 76439808-76439808 on Build GRCh38
rs987101244 Chr.14: 76439868-76439868 on Build GRCh38
rs947223845 Chr.14: 76439859-76439859 on Build GRCh38
rs768452968 Chr.14: 76439801-76439801 on Build GRCh38
rs577847646 Chr.14: 76439810-76439810 on Build GRCh38
rs931917294 Chr.14: 76439838-76439838 on Build GRCh38
rs767674170 Chr.14: 76439715-76439715 on Build GRCh38
rs542941260 Chr.14: 76439875-76439875 on Build GRCh38
rs1035296646 Chr.14: 76439804-76439804 on Build GRCh38
rs555646531 Chr.14: 76439779-76439779 on Build GRCh38
rs765676575 Chr.14: 76439773-76439773 on Build GRCh38
rs141586518 Chr.14: 76439704-76439704 on Build GRCh38
rs773307234 Chr.14: 76439726-76439726 on Build GRCh38
rs138961914 Chr.14: 76439780-76439780 on Build GRCh38
rs371516512 Chr.14: 76439794-76439794 on Build GRCh38
rs749286499 Chr.14: 76439800-76439800 on Build GRCh38
rs45507197 Chr.14: 76439806-76439806 on Build GRCh38
rs113856973 Chr.14: 76439846-76439846 on Build GRCh38
rs963817204 Chr.14: 76439876-76439876 on Build GRCh38
rs876657808 Chr.14: 76439755-76439755 on Build GRCh38
rs923147422 Chr.14: 76439832-76439832 on Build GRCh38
rs776785034 Chr.14: 76439836-76439836 on Build GRCh38
rs956426462 Chr.14: 76439860-76439860 on Build GRCh38
rs755742203 Chr.14: 76439795-76439795 on Build GRCh38
rs754429624 Chr.14: 76439768-76439768 on Build GRCh38
rs12432875 Chr.14: 76439814-76439814 on Build GRCh38
rs45511205 Chr.14: 76439850-76439850 on Build GRCh38

View Full Product Details