Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44447431-44447688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD8
Entrez Gene ID
386617
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
UniGene
Hs.479644
Gene Chromosome Location
Chr.4: 44173903-44448885 on Build GRCh38
Transcripts
NM_198353.2 XM_011513690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5947573 c.841G>C p.D281H Chr.4: 44447683-44447683 on Build GRCh38
6236167 c.903C>T p.F301F Chr.4: 44447621-44447621 on Build GRCh38
5953764 c.961+6T>C p.? Chr.4: 44447557-44447557 on Build GRCh38
6692134 c.889G>A p.G297S Chr.4: 44447635-44447635 on Build GRCh38
5011275 c.922G>T p.D308Y Chr.4: 44447602-44447602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932339337 Chr.4: 44447669-44447669 on Build GRCh38
rs977643242 Chr.4: 44447555-44447555 on Build GRCh38
rs972839315 Chr.4: 44447648-44447648 on Build GRCh38
rs1001511874 Chr.4: 44447582-44447582 on Build GRCh38
rs747676954 Chr.4: 44447611-44447611 on Build GRCh38
rs924751109 Chr.4: 44447577-44447577 on Build GRCh38
rs765408963 Chr.4: 44447547-44447547 on Build GRCh38
rs754595486 Chr.4: 44447579-44447579 on Build GRCh38
rs775710206 Chr.4: 44447658-44447658 on Build GRCh38
rs372138598 Chr.4: 44447589-44447589 on Build GRCh38
rs765968801 Chr.4: 44447591-44447591 on Build GRCh38
rs771159710 Chr.4: 44447526-44447526 on Build GRCh38
rs935082580 Chr.4: 44447476-44447476 on Build GRCh38
rs769540960 Chr.4: 44447495-44447495 on Build GRCh38
rs921513700 Chr.4: 44447634-44447634 on Build GRCh38
rs778380899 Chr.4: 44447609-44447609 on Build GRCh38
rs761800922 Chr.4: 44447565-44447565 on Build GRCh38
rs779825338 Chr.4: 44447532-44447532 on Build GRCh38
rs750203965 Chr.4: 44447585-44447585 on Build GRCh38
rs117297857 Chr.4: 44447630-44447630 on Build GRCh38
rs759644507 Chr.4: 44447542-44447542 on Build GRCh38
rs778189286 Chr.4: 44447621-44447621 on Build GRCh38
rs753521049 Chr.4: 44447600-44447600 on Build GRCh38
rs776923781 Chr.4: 44447631-44447631 on Build GRCh38
rs774377016 Chr.4: 44447548-44447548 on Build GRCh38
rs868158851 Chr.4: 44447644-44447644 on Build GRCh38
rs754419042 Chr.4: 44447607-44447607 on Build GRCh38
rs1043515506 Chr.4: 44447469-44447469 on Build GRCh38
rs905889612 Chr.4: 44447576-44447576 on Build GRCh38
rs566017725 Chr.4: 44447475-44447475 on Build GRCh38
rs758905704 Chr.4: 44447617-44447617 on Build GRCh38
rs762985548 Chr.4: 44447666-44447666 on Build GRCh38
rs1012869015 Chr.4: 44447467-44447467 on Build GRCh38
rs747346931 Chr.4: 44447628-44447628 on Build GRCh38
rs745421635 Chr.4: 44447473-44447473 on Build GRCh38
rs1023475544 Chr.4: 44447575-44447575 on Build GRCh38
rs557968835 Chr.4: 44447497-44447497 on Build GRCh38
rs903623958 Chr.4: 44447470-44447470 on Build GRCh38
rs539725620 Chr.4: 44447482-44447482 on Build GRCh38
rs746110451 Chr.4: 44447649-44447649 on Build GRCh38
rs965670478 Chr.4: 44447601-44447601 on Build GRCh38
rs770203655 Chr.4: 44447656-44447656 on Build GRCh38
rs11944873 Chr.4: 44447518-44447518 on Build GRCh38
rs770727878 Chr.4: 44447586-44447586 on Build GRCh38
rs779207636 Chr.4: 44447551-44447551 on Build GRCh38
rs777013649 Chr.4: 44447530-44447530 on Build GRCh38

View Full Product Details