Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50513689-50513918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPDH
Entrez Gene ID
554235
Gene Name
aspartate dehydrogenase domain containing
Gene Aliases
-
UniGene
Hs.436338
Gene Chromosome Location
Chr.19: 50511600-50514690 on Build GRCh38
Transcripts
NM_001024656.2 NM_001114598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
999752 c.29G>A p.G10D Chr.19: 50513795-50513795 on Build GRCh38
2754871 c.28G>C p.G10R Chr.19: 50513796-50513796 on Build GRCh38
4599326 c.17C>G p.P6R Chr.19: 50513807-50513807 on Build GRCh38
3835457 c.52+1G>A p.? Chr.19: 50513771-50513771 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747302101 Chr.19: 50513783-50513783 on Build GRCh38
rs779819484 Chr.19: 50513726-50513726 on Build GRCh38
rs1010036087 Chr.19: 50513712-50513712 on Build GRCh38
rs1015968343 Chr.19: 50513778-50513778 on Build GRCh38
rs767251459 Chr.19: 50513808-50513808 on Build GRCh38
rs773392982 Chr.19: 50513773-50513773 on Build GRCh38
rs752606168 Chr.19: 50513810-50513810 on Build GRCh38
rs949383015 Chr.19: 50513888-50513888 on Build GRCh38
rs771154423 Chr.19: 50513859-50513860 on Build GRCh38
rs181814523 Chr.19: 50513896-50513896 on Build GRCh38
rs891672962 Chr.19: 50513867-50513867 on Build GRCh38
rs756820539 Chr.19: 50513856-50513856 on Build GRCh38
rs547346495 Chr.19: 50513801-50513801 on Build GRCh38
rs1017936937 Chr.19: 50513772-50513772 on Build GRCh38
rs770174125 Chr.19: 50513731-50513731 on Build GRCh38
rs904265223 Chr.19: 50513742-50513742 on Build GRCh38
rs77177254 Chr.19: 50513806-50513806 on Build GRCh38
rs1006999602 Chr.19: 50513754-50513754 on Build GRCh38
rs760879776 Chr.19: 50513762-50513763 on Build GRCh38
rs535109093 Chr.19: 50513796-50513796 on Build GRCh38
rs778711163 Chr.19: 50513863-50513863 on Build GRCh38
rs1046326777 Chr.19: 50513895-50513895 on Build GRCh38
rs748591566 Chr.19: 50513866-50513866 on Build GRCh38
rs576379216 Chr.19: 50513845-50513845 on Build GRCh38
rs990860403 Chr.19: 50513874-50513874 on Build GRCh38
rs1030555372 Chr.19: 50513802-50513802 on Build GRCh38
rs539778691 Chr.19: 50513822-50513822 on Build GRCh38
rs549312958 Chr.19: 50513729-50513729 on Build GRCh38
rs927314217 Chr.19: 50513830-50513830 on Build GRCh38
rs945905918 Chr.19: 50513878-50513878 on Build GRCh38
rs763913462 Chr.19: 50513817-50513817 on Build GRCh38
rs543057368 Chr.19: 50513873-50513873 on Build GRCh38
rs916570289 Chr.19: 50513880-50513880 on Build GRCh38
rs376542665 Chr.19: 50513835-50513835 on Build GRCh38
rs756116193 Chr.19: 50513812-50513812 on Build GRCh38
rs978216839 Chr.19: 50513841-50513841 on Build GRCh38
rs936732464 Chr.19: 50513868-50513868 on Build GRCh38
rs7248272 Chr.19: 50513807-50513807 on Build GRCh38
rs147876313 Chr.19: 50513794-50513794 on Build GRCh38

View Full Product Details