Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 103275886-103276159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf30
Entrez Gene ID
90355
Gene Name
chromosome 5 open reading frame 30
Gene Aliases
-
UniGene
Hs.482976
Gene Chromosome Location
Chr.5: 103258702-103278660 on Build GRCh38
Transcripts
NM_001316968.1 NM_001316969.1 NM_033211.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5041464 c.146C>G p.S49* Chr.5: 103276065-103276065 on Build GRCh38
5124821 c.104G>A p.R35H Chr.5: 103276023-103276023 on Build GRCh38
5687999 c.87G>A p.A29A Chr.5: 103276006-103276006 on Build GRCh38
1198870 c.134G>A p.R45Q Chr.5: 103276053-103276053 on Build GRCh38
5709417 c.215A>G p.K72R Chr.5: 103276134-103276134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782690747 Chr.5: 103276130-103276130 on Build GRCh38
rs782715851 Chr.5: 103275974-103275974 on Build GRCh38
rs565547139 Chr.5: 103275994-103275994 on Build GRCh38
rs369790097 Chr.5: 103276064-103276064 on Build GRCh38
rs782054406 Chr.5: 103275958-103275958 on Build GRCh38
rs1053074462 Chr.5: 103275973-103275973 on Build GRCh38
rs944061429 Chr.5: 103275950-103275950 on Build GRCh38
rs782476366 Chr.5: 103276136-103276136 on Build GRCh38
rs139004073 Chr.5: 103276048-103276048 on Build GRCh38
rs892371431 Chr.5: 103275978-103275978 on Build GRCh38
rs375435402 Chr.5: 103276050-103276050 on Build GRCh38
rs782756632 Chr.5: 103276114-103276114 on Build GRCh38
rs148596053 Chr.5: 103275982-103275982 on Build GRCh38
rs961611433 Chr.5: 103276073-103276073 on Build GRCh38
rs377719276 Chr.5: 103276133-103276133 on Build GRCh38
rs201922818 Chr.5: 103275996-103275996 on Build GRCh38
rs781879436 Chr.5: 103276092-103276092 on Build GRCh38
rs782244316 Chr.5: 103276132-103276132 on Build GRCh38
rs150195375 Chr.5: 103276123-103276123 on Build GRCh38
rs782720333 Chr.5: 103276078-103276078 on Build GRCh38
rs782521601 Chr.5: 103276060-103276060 on Build GRCh38
rs111728159 Chr.5: 103276005-103276005 on Build GRCh38
rs781976718 Chr.5: 103275956-103275956 on Build GRCh38
rs372789948 Chr.5: 103276113-103276113 on Build GRCh38
rs782133654 Chr.5: 103275949-103275949 on Build GRCh38
rs562216410 Chr.5: 103275951-103275951 on Build GRCh38
rs781789147 Chr.5: 103275997-103275997 on Build GRCh38
rs960377348 Chr.5: 103276025-103276025 on Build GRCh38
rs1015296666 Chr.5: 103276052-103276052 on Build GRCh38
rs538732104 Chr.5: 103276012-103276012 on Build GRCh38
rs146463444 Chr.5: 103276121-103276121 on Build GRCh38
rs782291611 Chr.5: 103276022-103276022 on Build GRCh38
rs372423937 Chr.5: 103276045-103276045 on Build GRCh38
rs782563123 Chr.5: 103275918-103275918 on Build GRCh38
rs115795390 Chr.5: 103276075-103276075 on Build GRCh38
rs920067089 Chr.5: 103276108-103276108 on Build GRCh38
rs201791651 Chr.5: 103275988-103275988 on Build GRCh38
rs954369299 Chr.5: 103276111-103276111 on Build GRCh38
rs781969121 Chr.5: 103276066-103276066 on Build GRCh38
rs782370807 Chr.5: 103276027-103276027 on Build GRCh38
rs147509179 Chr.5: 103275980-103275980 on Build GRCh38
rs781987839 Chr.5: 103276063-103276063 on Build GRCh38
rs189792579 Chr.5: 103275929-103275929 on Build GRCh38
rs782728831 Chr.5: 103276065-103276065 on Build GRCh38
rs548779142 Chr.5: 103275947-103275947 on Build GRCh38
rs114035466 Chr.5: 103276120-103276120 on Build GRCh38
rs377580691 Chr.5: 103276118-103276118 on Build GRCh38
rs782230140 Chr.5: 103276047-103276047 on Build GRCh38
rs782049974 Chr.5: 103275928-103275928 on Build GRCh38
rs782274203 Chr.5: 103276056-103276056 on Build GRCh38
rs782659787 Chr.5: 103276006-103276006 on Build GRCh38
rs782193473 Chr.5: 103275923-103275923 on Build GRCh38
rs781942521 Chr.5: 103276053-103276053 on Build GRCh38
rs142953265 Chr.5: 103275984-103275984 on Build GRCh38

View Full Product Details